Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 58564037-58564530 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM107A
Entrez Gene ID
11170
Gene Name
family with sequence similarity 107 member A
Gene Aliases
DRR1, TU3A
UniGene
Hs.506357
Gene Chromosome Location
Chr.3: 58564112-58627610 on Build GRCh38
Transcripts
NM_001076778.2 NM_001282713.1 NM_001282714.1 NM_007177.3

SNP Detail

SNP ID Chromosome Location
rs577259917 Chr.3: 58564468-58564468 on Build GRCh38
rs968464860 Chr.3: 58564356-58564356 on Build GRCh38
rs559531016 Chr.3: 58564165-58564165 on Build GRCh38
rs143377731 Chr.3: 58564483-58564483 on Build GRCh38
rs917188187 Chr.3: 58564493-58564493 on Build GRCh38
rs141068018 Chr.3: 58564371-58564371 on Build GRCh38
rs1026500677 Chr.3: 58564105-58564105 on Build GRCh38
rs904968062 Chr.3: 58564322-58564322 on Build GRCh38
rs868338059 Chr.3: 58564337-58564337 on Build GRCh38
rs187446495 Chr.3: 58564113-58564113 on Build GRCh38
rs1009156976 Chr.3: 58564169-58564169 on Build GRCh38
rs746086845 Chr.3: 58564197-58564197 on Build GRCh38
rs939063192 Chr.3: 58564325-58564325 on Build GRCh38
rs914917178 Chr.3: 58564211-58564211 on Build GRCh38
rs1023041005 Chr.3: 58564058-58564058 on Build GRCh38
rs1019661505 Chr.3: 58564235-58564235 on Build GRCh38
rs994984206 Chr.3: 58564088-58564088 on Build GRCh38
rs921001640 Chr.3: 58564328-58564328 on Build GRCh38
rs115364085 Chr.3: 58564085-58564085 on Build GRCh38
rs531011224 Chr.3: 58564429-58564429 on Build GRCh38
rs978542748 Chr.3: 58564366-58564366 on Build GRCh38
rs528634387 Chr.3: 58564250-58564250 on Build GRCh38
rs568493806 Chr.3: 58564319-58564319 on Build GRCh38
rs370433261 Chr.3: 58564275-58564275 on Build GRCh38
rs546079963 Chr.3: 58564124-58564124 on Build GRCh38
rs924461746 Chr.3: 58564401-58564401 on Build GRCh38
rs1018746272 Chr.3: 58564057-58564057 on Build GRCh38
rs562612144 Chr.3: 58564130-58564130 on Build GRCh38
rs897652906 Chr.3: 58564424-58564424 on Build GRCh38
rs373568117 Chr.3: 58564321-58564321 on Build GRCh38
rs935621411 Chr.3: 58564122-58564122 on Build GRCh38

View Full Product Details