Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184342178-184342681 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM131A
Entrez Gene ID
131408
Gene Name
family with sequence similarity 131 member A
Gene Aliases
C3orf40, FLAT715, PRO1378
UniGene
Hs.591307
Gene Chromosome Location
Chr.3: 184335924-184346275 on Build GRCh38
Transcripts
NM_001171093.1 NM_144635.4 XM_005247113.2 XM_005247114.3 XM_011512413.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6529138 c.388G>C p.G130R Chr.3: 184342221-184342221 on Build GRCh38
6097153 c.400G>C p.A134P Chr.3: 184342233-184342233 on Build GRCh38
6097152 c.493G>C p.A165P Chr.3: 184342233-184342233 on Build GRCh38
6529137 c.481G>C p.G161R Chr.3: 184342221-184342221 on Build GRCh38
5864041 c.497G>T p.R166L Chr.3: 184342237-184342237 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1043898372 Chr.3: 184342640-184342640 on Build GRCh38
rs529531031 Chr.3: 184342628-184342628 on Build GRCh38
rs566414808 Chr.3: 184342602-184342602 on Build GRCh38
rs749237896 Chr.3: 184342265-184342265 on Build GRCh38
rs909939654 Chr.3: 184342596-184342596 on Build GRCh38
rs372362074 Chr.3: 184342236-184342236 on Build GRCh38
rs891555658 Chr.3: 184342459-184342459 on Build GRCh38
rs545391860 Chr.3: 184342330-184342330 on Build GRCh38
rs939241780 Chr.3: 184342607-184342607 on Build GRCh38
rs112798724 Chr.3: 184342297-184342297 on Build GRCh38
rs906446665 Chr.3: 184342369-184342369 on Build GRCh38
rs982064488 Chr.3: 184342289-184342289 on Build GRCh38
rs543453994 Chr.3: 184342346-184342346 on Build GRCh38
rs532510517 Chr.3: 184342586-184342586 on Build GRCh38
rs984549018 Chr.3: 184342585-184342585 on Build GRCh38
rs757804140 Chr.3: 184342392-184342392 on Build GRCh38
rs1037285596 Chr.3: 184342336-184342336 on Build GRCh38
rs561417322 Chr.3: 184342440-184342440 on Build GRCh38
rs751229762 Chr.3: 184342294-184342297 on Build GRCh38
rs754348451 Chr.3: 184342316-184342317 on Build GRCh38
rs762382348 Chr.3: 184342363-184342363 on Build GRCh38
rs780215339 Chr.3: 184342274-184342274 on Build GRCh38
rs974921944 Chr.3: 184342480-184342480 on Build GRCh38
rs531847929 Chr.3: 184342488-184342488 on Build GRCh38
rs755898900 Chr.3: 184342241-184342241 on Build GRCh38
rs972575675 Chr.3: 184342637-184342637 on Build GRCh38
rs551596470 Chr.3: 184342569-184342569 on Build GRCh38
rs563965153 Chr.3: 184342382-184342382 on Build GRCh38
rs1019233997 Chr.3: 184342455-184342455 on Build GRCh38
rs940692802 Chr.3: 184342347-184342347 on Build GRCh38
rs1003354349 Chr.3: 184342370-184342370 on Build GRCh38
rs970933514 Chr.3: 184342514-184342514 on Build GRCh38
rs140044854 Chr.3: 184342221-184342221 on Build GRCh38
rs930694328 Chr.3: 184342328-184342328 on Build GRCh38
rs371092672 Chr.3: 184342280-184342280 on Build GRCh38
rs76640995 Chr.3: 184342281-184342281 on Build GRCh38
rs754589250 Chr.3: 184342224-184342224 on Build GRCh38
rs762436793 Chr.3: 184342205-184342205 on Build GRCh38
rs752518495 Chr.3: 184342237-184342237 on Build GRCh38
rs985320627 Chr.3: 184342523-184342523 on Build GRCh38
rs1036943157 Chr.3: 184342357-184342357 on Build GRCh38
rs1033903640 Chr.3: 184342558-184342558 on Build GRCh38
rs922060565 Chr.3: 184342316-184342316 on Build GRCh38
rs1009707263 Chr.3: 184342466-184342466 on Build GRCh38
rs367585415 Chr.3: 184342293-184342293 on Build GRCh38
rs909961412 Chr.3: 184342317-184342317 on Build GRCh38
rs777759456 Chr.3: 184342264-184342264 on Build GRCh38
rs1010735389 Chr.3: 184342624-184342624 on Build GRCh38
rs752668260 Chr.3: 184342609-184342609 on Build GRCh38
rs1011036609 Chr.3: 184342403-184342403 on Build GRCh38
rs577524867 Chr.3: 184342315-184342315 on Build GRCh38
rs766038750 Chr.3: 184342291-184342292 on Build GRCh38
rs894931526 Chr.3: 184342620-184342620 on Build GRCh38
rs961789750 Chr.3: 184342598-184342598 on Build GRCh38
rs762359380 Chr.3: 184342290-184342292 on Build GRCh38
rs1045689770 Chr.3: 184342591-184342591 on Build GRCh38
rs777228984 Chr.3: 184342360-184342360 on Build GRCh38
rs922061138 Chr.3: 184342494-184342494 on Build GRCh38
rs771002781 Chr.3: 184342272-184342272 on Build GRCh38
rs765749876 Chr.3: 184342371-184342371 on Build GRCh38
rs902247531 Chr.3: 184342643-184342643 on Build GRCh38
rs751077199 Chr.3: 184342223-184342223 on Build GRCh38
rs750808376 Chr.3: 184342380-184342380 on Build GRCh38
rs551415120 Chr.3: 184342601-184342601 on Build GRCh38
rs781091023 Chr.3: 184342581-184342581 on Build GRCh38
rs966992911 Chr.3: 184342217-184342217 on Build GRCh38
rs565153567 Chr.3: 184342567-184342567 on Build GRCh38
rs543860725 Chr.3: 184342566-184342566 on Build GRCh38
rs999180883 Chr.3: 184342453-184342453 on Build GRCh38
rs751675604 Chr.3: 184342508-184342508 on Build GRCh38
rs768948455 Chr.3: 184342287-184342287 on Build GRCh38
rs779104966 Chr.3: 184342627-184342627 on Build GRCh38
rs755104979 Chr.3: 184342551-184342551 on Build GRCh38
rs1021961928 Chr.3: 184342417-184342417 on Build GRCh38
rs543224776 Chr.3: 184342434-184342434 on Build GRCh38
rs966261895 Chr.3: 184342462-184342462 on Build GRCh38
rs572704158 Chr.3: 184342415-184342415 on Build GRCh38

View Full Product Details