Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 83460600-83461136 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM175A
Entrez Gene ID
84142
Gene Name
family with sequence similarity 175 member A
Gene Aliases
ABRA1, CCDC98
UniGene
Hs.334772
Gene Chromosome Location
Chr.4: 83459515-83485181 on Build GRCh38
Transcripts
NM_139076.2 XM_005263280.4 XM_017008694.1 NM_001297767.1 NM_001297769.1 NM_001297770.1 NM_016067.3

Gene Symbol
MRPS18C
Entrez Gene ID
51023
Gene Name
mitochondrial ribosomal protein S18C
Gene Aliases
CGI-134, MRP-S18-1, MRP-S18-c, MRPS18-1, S18mt-c, mrps18-c
UniGene
Hs.334772
Gene Chromosome Location
Chr.4: 83455932-83461723 on Build GRCh38
Transcripts
NM_139076.2 XM_005263280.4 XM_017008694.1 NM_001297767.1 NM_001297769.1 NM_001297770.1 NM_016067.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1495899 c.361C>T p.P121S Chr.4: 83461129-83461129 on Build GRCh38
262071 c.316G>T p.E106* Chr.4: 83460996-83460996 on Build GRCh38
3826345 c.340G>A p.A114T Chr.4: 83461020-83461020 on Build GRCh38
1310326 c.332T>A p.I111N Chr.4: 83461012-83461012 on Build GRCh38
269621 c.355T>G p.F119V Chr.4: 83461123-83461123 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs771159531 Chr.4: 83460945-83460945 on Build GRCh38
rs535536590 Chr.4: 83460643-83460643 on Build GRCh38
rs889218453 Chr.4: 83460713-83460713 on Build GRCh38
rs909372912 Chr.4: 83460854-83460854 on Build GRCh38
rs1033140980 Chr.4: 83460655-83460655 on Build GRCh38
rs752766600 Chr.4: 83460979-83460979 on Build GRCh38
rs79609149 Chr.4: 83460905-83460905 on Build GRCh38
rs762949273 Chr.4: 83460980-83460980 on Build GRCh38
rs763486674 Chr.4: 83460821-83460824 on Build GRCh38
rs751703470 Chr.4: 83461108-83461108 on Build GRCh38
rs754733491 Chr.4: 83460849-83460849 on Build GRCh38
rs1007584922 Chr.4: 83460989-83460989 on Build GRCh38
rs756109273 Chr.4: 83461044-83461044 on Build GRCh38
rs535542234 Chr.4: 83460821-83460821 on Build GRCh38
rs998244480 Chr.4: 83460789-83460789 on Build GRCh38
rs972120072 Chr.4: 83460717-83460717 on Build GRCh38
rs573173687 Chr.4: 83460860-83460860 on Build GRCh38
rs748866658 Chr.4: 83460927-83460927 on Build GRCh38
rs765788201 Chr.4: 83460975-83460975 on Build GRCh38
rs183008110 Chr.4: 83461059-83461059 on Build GRCh38
rs991712842 Chr.4: 83460685-83460685 on Build GRCh38
rs764957517 Chr.4: 83460859-83460859 on Build GRCh38
rs752066672 Chr.4: 83460721-83460721 on Build GRCh38
rs377575434 Chr.4: 83460855-83460855 on Build GRCh38
rs557033911 Chr.4: 83460661-83460661 on Build GRCh38
rs970689654 Chr.4: 83461092-83461092 on Build GRCh38
rs767385157 Chr.4: 83460991-83460992 on Build GRCh38
rs958837718 Chr.4: 83460882-83460882 on Build GRCh38
rs917524193 Chr.4: 83460698-83460698 on Build GRCh38
rs377194442 Chr.4: 83460771-83460772 on Build GRCh38
rs559047395 Chr.4: 83460934-83460934 on Build GRCh38
rs1041425183 Chr.4: 83460820-83460820 on Build GRCh38
rs958870305 Chr.4: 83460679-83460679 on Build GRCh38
rs865809205 Chr.4: 83460639-83460639 on Build GRCh38
rs751687532 Chr.4: 83461016-83461016 on Build GRCh38
rs774881598 Chr.4: 83460991-83460994 on Build GRCh38
rs142051764 Chr.4: 83461111-83461111 on Build GRCh38
rs1013116288 Chr.4: 83460917-83460917 on Build GRCh38
rs1052246937 Chr.4: 83460704-83460704 on Build GRCh38
rs377741260 Chr.4: 83461010-83461010 on Build GRCh38
rs750295282 Chr.4: 83461037-83461037 on Build GRCh38
rs759956917 Chr.4: 83461077-83461077 on Build GRCh38
rs575476249 Chr.4: 83460716-83460716 on Build GRCh38
rs190420900 Chr.4: 83460920-83460920 on Build GRCh38
rs780209189 Chr.4: 83461050-83461050 on Build GRCh38
rs1009365810 Chr.4: 83461038-83461038 on Build GRCh38
rs749242864 Chr.4: 83461052-83461052 on Build GRCh38
rs570659348 Chr.4: 83461019-83461019 on Build GRCh38
rs188443986 Chr.4: 83461088-83461088 on Build GRCh38
rs901111188 Chr.4: 83460766-83460766 on Build GRCh38
rs752577070 Chr.4: 83460888-83460888 on Build GRCh38
rs187948729 Chr.4: 83460723-83460723 on Build GRCh38
rs1036649813 Chr.4: 83460680-83460681 on Build GRCh38
rs1020832878 Chr.4: 83460803-83460803 on Build GRCh38
rs397733178 Chr.4: 83460916-83460916 on Build GRCh38
rs753564204 Chr.4: 83460837-83460837 on Build GRCh38
rs573367922 Chr.4: 83460906-83460907 on Build GRCh38
rs997529080 Chr.4: 83460694-83460694 on Build GRCh38
rs745790422 Chr.4: 83460959-83460960 on Build GRCh38
rs1003895186 Chr.4: 83461093-83461093 on Build GRCh38
rs376490450 Chr.4: 83460640-83460640 on Build GRCh38
rs761549292 Chr.4: 83460936-83460936 on Build GRCh38
rs760125948 Chr.4: 83460732-83460732 on Build GRCh38
rs776845603 Chr.4: 83461076-83461076 on Build GRCh38
rs780821291 Chr.4: 83461030-83461030 on Build GRCh38
rs765919213 Chr.4: 83460750-83460750 on Build GRCh38
rs770145230 Chr.4: 83461083-83461083 on Build GRCh38
rs1051789775 Chr.4: 83460915-83460915 on Build GRCh38
rs1039571064 Chr.4: 83460763-83460763 on Build GRCh38
rs777092043 Chr.4: 83460973-83460973 on Build GRCh38
rs747724744 Chr.4: 83461062-83461062 on Build GRCh38
rs763469011 Chr.4: 83461097-83461097 on Build GRCh38
rs34734728 Chr.4: 83460906-83460906 on Build GRCh38
rs111663085 Chr.4: 83460718-83460718 on Build GRCh38
rs746483791 Chr.4: 83461103-83461106 on Build GRCh38
rs771803901 Chr.4: 83461070-83461070 on Build GRCh38
rs370198625 Chr.4: 83460933-83460933 on Build GRCh38
rs970768387 Chr.4: 83460813-83460813 on Build GRCh38
rs758168197 Chr.4: 83460924-83460924 on Build GRCh38
rs764044193 Chr.4: 83461106-83461106 on Build GRCh38
rs919525568 Chr.4: 83460869-83460869 on Build GRCh38
rs779054803 Chr.4: 83460938-83460939 on Build GRCh38

View Full Product Details