Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 35045781-35046276 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM177A1
Entrez Gene ID
283635
Gene Name
family with sequence similarity 177 member A1
Gene Aliases
C14orf24
UniGene
Hs.446357
Gene Chromosome Location
Chr.14: 35044907-35083383 on Build GRCh38
Transcripts
NM_001079519.1 NM_001289022.1

SNP Detail

SNP ID Chromosome Location
rs990578610 Chr.14: 35046203-35046203 on Build GRCh38
rs536424377 Chr.14: 35046143-35046143 on Build GRCh38
rs780661610 Chr.14: 35046034-35046034 on Build GRCh38
rs921693221 Chr.14: 35046147-35046147 on Build GRCh38
rs909575903 Chr.14: 35046241-35046241 on Build GRCh38
rs1048917865 Chr.14: 35046014-35046014 on Build GRCh38
rs548799731 Chr.14: 35046155-35046164 on Build GRCh38
rs557378707 Chr.14: 35046119-35046119 on Build GRCh38
rs1036269452 Chr.14: 35046195-35046195 on Build GRCh38
rs1004088138 Chr.14: 35046186-35046186 on Build GRCh38
rs185805780 Chr.14: 35046162-35046162 on Build GRCh38
rs989032546 Chr.14: 35046135-35046135 on Build GRCh38
rs952174658 Chr.14: 35045934-35045934 on Build GRCh38
rs962351358 Chr.14: 35046242-35046242 on Build GRCh38
rs3809448 Chr.14: 35045890-35045890 on Build GRCh38
rs957543613 Chr.14: 35046129-35046129 on Build GRCh38
rs944591952 Chr.14: 35046232-35046232 on Build GRCh38
rs981903343 Chr.14: 35046007-35046007 on Build GRCh38
rs45447399 Chr.14: 35046121-35046121 on Build GRCh38
rs1046182501 Chr.14: 35046235-35046235 on Build GRCh38
rs1017864246 Chr.14: 35045829-35045829 on Build GRCh38
rs1028139372 Chr.14: 35045891-35045891 on Build GRCh38
rs905864731 Chr.14: 35046256-35046256 on Build GRCh38
rs546306248 Chr.14: 35046042-35046042 on Build GRCh38
rs180886022 Chr.14: 35046136-35046136 on Build GRCh38
rs918475944 Chr.14: 35046251-35046251 on Build GRCh38
rs1001627019 Chr.14: 35046115-35046115 on Build GRCh38
rs959079412 Chr.14: 35046030-35046030 on Build GRCh38
rs896418234 Chr.14: 35046224-35046233 on Build GRCh38
rs989063605 Chr.14: 35045929-35045929 on Build GRCh38
rs926190544 Chr.14: 35045881-35045881 on Build GRCh38
rs1036935918 Chr.14: 35046110-35046110 on Build GRCh38
rs376475543 Chr.14: 35046138-35046138 on Build GRCh38
rs1011138667 Chr.14: 35046166-35046166 on Build GRCh38
rs941293626 Chr.14: 35046105-35046105 on Build GRCh38
rs756855463 Chr.14: 35046017-35046017 on Build GRCh38
rs897126026 Chr.14: 35046047-35046047 on Build GRCh38
rs958243279 Chr.14: 35045856-35045856 on Build GRCh38
rs944384161 Chr.14: 35045938-35045938 on Build GRCh38
rs1036709095 Chr.14: 35046037-35046037 on Build GRCh38
rs867889547 Chr.14: 35046180-35046180 on Build GRCh38
rs1021139034 Chr.14: 35046169-35046169 on Build GRCh38
rs921699376 Chr.14: 35046004-35046004 on Build GRCh38
rs546101659 Chr.14: 35046122-35046122 on Build GRCh38
rs529668721 Chr.14: 35046190-35046190 on Build GRCh38
rs966918013 Chr.14: 35046173-35046173 on Build GRCh38
rs537788526 Chr.14: 35046025-35046025 on Build GRCh38
rs950078861 Chr.14: 35046225-35046225 on Build GRCh38
rs1007561596 Chr.14: 35046146-35046146 on Build GRCh38
rs1034780797 Chr.14: 35046188-35046188 on Build GRCh38
rs570246597 Chr.14: 35046191-35046191 on Build GRCh38
rs1024376490 Chr.14: 35046075-35046075 on Build GRCh38
rs972661601 Chr.14: 35046246-35046246 on Build GRCh38
rs1014699676 Chr.14: 35046066-35046066 on Build GRCh38
rs1021109242 Chr.14: 35045931-35045931 on Build GRCh38
rs1039048949 Chr.14: 35046155-35046155 on Build GRCh38
rs933792905 Chr.14: 35046131-35046131 on Build GRCh38
rs986342847 Chr.14: 35045962-35045962 on Build GRCh38
rs571838339 Chr.14: 35046137-35046137 on Build GRCh38
rs575026789 Chr.14: 35046021-35046021 on Build GRCh38
rs909547977 Chr.14: 35046096-35046096 on Build GRCh38
rs748760899 Chr.14: 35045998-35045998 on Build GRCh38
rs931812141 Chr.14: 35046010-35046010 on Build GRCh38
rs560591554 Chr.14: 35046163-35046163 on Build GRCh38

View Full Product Details