Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 45004063-45004609 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM179B
Entrez Gene ID
23116
Gene Name
family with sequence similarity 179 member B
Gene Aliases
KIAA0423
UniGene
Hs.371078
Gene Chromosome Location
Chr.14: 44962190-45074431 on Build GRCh38
Transcripts
NM_001308120.1 NM_015091.3 NR_131765.1 XM_011536571.1 XM_017021098.1 XM_017021099.1 XM_017021100.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6554337 c.2644G>T p.G882* Chr.14: 45004366-45004366 on Build GRCh38
2197278 c.2408C>T p.S803L Chr.14: 45004130-45004130 on Build GRCh38
3956036 c.2345C>T p.A782V Chr.14: 45004067-45004067 on Build GRCh38
2197281 c.2549C>T p.S850F Chr.14: 45004271-45004271 on Build GRCh38
5705704 c.2579C>G p.S860* Chr.14: 45004301-45004301 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749729265 Chr.14: 45004380-45004380 on Build GRCh38
rs746338197 Chr.14: 45004214-45004214 on Build GRCh38
rs762811903 Chr.14: 45004312-45004312 on Build GRCh38
rs910506446 Chr.14: 45004510-45004510 on Build GRCh38
rs200505398 Chr.14: 45004379-45004379 on Build GRCh38
rs751371979 Chr.14: 45004177-45004177 on Build GRCh38
rs774214182 Chr.14: 45004324-45004324 on Build GRCh38
rs377532752 Chr.14: 45004352-45004352 on Build GRCh38
rs776208820 Chr.14: 45004144-45004144 on Build GRCh38
rs898692227 Chr.14: 45004442-45004442 on Build GRCh38
rs151062820 Chr.14: 45004213-45004213 on Build GRCh38
rs559068906 Chr.14: 45004413-45004413 on Build GRCh38
rs551896855 Chr.14: 45004489-45004492 on Build GRCh38
rs995631047 Chr.14: 45004464-45004464 on Build GRCh38
rs1033459716 Chr.14: 45004135-45004135 on Build GRCh38
rs750846481 Chr.14: 45004106-45004106 on Build GRCh38
rs947191819 Chr.14: 45004290-45004290 on Build GRCh38
rs772517997 Chr.14: 45004110-45004110 on Build GRCh38
rs773108604 Chr.14: 45004162-45004162 on Build GRCh38
rs990541687 Chr.14: 45004198-45004198 on Build GRCh38
rs747762961 Chr.14: 45004364-45004364 on Build GRCh38
rs771859014 Chr.14: 45004406-45004406 on Build GRCh38
rs754064646 Chr.14: 45004337-45004337 on Build GRCh38
rs770627533 Chr.14: 45004090-45004091 on Build GRCh38
rs775956620 Chr.14: 45004120-45004120 on Build GRCh38
rs369978624 Chr.14: 45004353-45004353 on Build GRCh38
rs756231171 Chr.14: 45004178-45004178 on Build GRCh38
rs112125886 Chr.14: 45004402-45004402 on Build GRCh38
rs1040211527 Chr.14: 45004250-45004250 on Build GRCh38
rs929222457 Chr.14: 45004279-45004279 on Build GRCh38
rs748783894 Chr.14: 45004377-45004377 on Build GRCh38
rs139041622 Chr.14: 45004346-45004346 on Build GRCh38
rs757390148 Chr.14: 45004212-45004212 on Build GRCh38
rs561717208 Chr.14: 45004316-45004316 on Build GRCh38
rs189300616 Chr.14: 45004394-45004394 on Build GRCh38
rs371750726 Chr.14: 45004245-45004245 on Build GRCh38
rs772742155 Chr.14: 45004302-45004302 on Build GRCh38
rs192596591 Chr.14: 45004253-45004253 on Build GRCh38
rs746578668 Chr.14: 45004124-45004124 on Build GRCh38
rs912646952 Chr.14: 45004125-45004125 on Build GRCh38
rs146298146 Chr.14: 45004343-45004343 on Build GRCh38
rs940428351 Chr.14: 45004431-45004431 on Build GRCh38
rs143566568 Chr.14: 45004152-45004152 on Build GRCh38
rs759371161 Chr.14: 45004329-45004329 on Build GRCh38
rs924632227 Chr.14: 45004192-45004192 on Build GRCh38
rs1008650091 Chr.14: 45004509-45004509 on Build GRCh38
rs766119873 Chr.14: 45004173-45004173 on Build GRCh38
rs768370970 Chr.14: 45004133-45004133 on Build GRCh38
rs866396556 Chr.14: 45004321-45004321 on Build GRCh38
rs765339576 Chr.14: 45004341-45004341 on Build GRCh38
rs777209073 Chr.14: 45004367-45004367 on Build GRCh38
rs1031910923 Chr.14: 45004475-45004475 on Build GRCh38
rs1028567120 Chr.14: 45004497-45004497 on Build GRCh38
rs773727693 Chr.14: 45004099-45004100 on Build GRCh38
rs762897857 Chr.14: 45004169-45004169 on Build GRCh38
rs375509272 Chr.14: 45004424-45004424 on Build GRCh38
rs956334769 Chr.14: 45004558-45004558 on Build GRCh38
rs943477441 Chr.14: 45004517-45004517 on Build GRCh38
rs199830668 Chr.14: 45004271-45004271 on Build GRCh38
rs1019590451 Chr.14: 45004548-45004548 on Build GRCh38
rs369731949 Chr.14: 45004193-45004193 on Build GRCh38
rs267603991 Chr.14: 45004277-45004277 on Build GRCh38
rs977539107 Chr.14: 45004383-45004383 on Build GRCh38
rs754074614 Chr.14: 45004208-45004208 on Build GRCh38
rs551357816 Chr.14: 45004428-45004428 on Build GRCh38
rs746427722 Chr.14: 45004104-45004104 on Build GRCh38
rs201409623 Chr.14: 45004236-45004236 on Build GRCh38
rs750542373 Chr.14: 45004342-45004342 on Build GRCh38
rs780227952 Chr.14: 45004243-45004243 on Build GRCh38
rs769443390 Chr.14: 45004366-45004366 on Build GRCh38
rs890002959 Chr.14: 45004507-45004507 on Build GRCh38
rs759135714 Chr.14: 45004292-45004307 on Build GRCh38
rs769249015 Chr.14: 45004299-45004299 on Build GRCh38
rs1047656317 Chr.14: 45004328-45004328 on Build GRCh38
rs764762357 Chr.14: 45004160-45004160 on Build GRCh38
rs757469684 Chr.14: 45004089-45004089 on Build GRCh38
rs767265010 Chr.14: 45004332-45004332 on Build GRCh38
rs747851068 Chr.14: 45004278-45004278 on Build GRCh38
rs762067590 Chr.14: 45004416-45004416 on Build GRCh38
rs141013517 Chr.14: 45004340-45004340 on Build GRCh38
rs1037617684 Chr.14: 45004437-45004437 on Build GRCh38
rs779531910 Chr.14: 45004094-45004094 on Build GRCh38
rs914381399 Chr.14: 45004283-45004283 on Build GRCh38

View Full Product Details