Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150997189-150997698 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM63A
Entrez Gene ID
55793
Gene Name
family with sequence similarity 63 member A
Gene Aliases
-
UniGene
Hs.743952
Gene Chromosome Location
Chr.1: 150996737-151008378 on Build GRCh38
Transcripts
NM_001040217.2 NM_001163258.1 NM_001163259.1 NM_001163260.1 NM_001319998.1 NM_018379.4 XM_005245321.3 XM_005245325.3 XM_005245327.3 XM_005245328.3 XM_011509757.2 XM_011509759.2 XM_017001770.1 XM_017001771.1 XM_017001772.1 XM_017001773.1 XM_017001774.1 XM_017001775.1 XM_017001776.1 XM_017001777.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3801784 c.1273C>G p.Q425E Chr.1: 150997680-150997680 on Build GRCh38
5223437 c.1417C>G p.Q473E Chr.1: 150997680-150997680 on Build GRCh38
895663 c.1330-1G>A p.? Chr.1: 150997368-150997368 on Build GRCh38
3771447 c.1360G>T p.G454W Chr.1: 150997337-150997337 on Build GRCh38
895662 c.1370G>A p.R457Q Chr.1: 150997327-150997327 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780224057 Chr.1: 150997572-150997572 on Build GRCh38
rs754411788 Chr.1: 150997321-150997321 on Build GRCh38
rs774523136 Chr.1: 150997295-150997295 on Build GRCh38
rs904822989 Chr.1: 150997658-150997658 on Build GRCh38
rs376405297 Chr.1: 150997596-150997596 on Build GRCh38
rs753647884 Chr.1: 150997642-150997642 on Build GRCh38
rs781248774 Chr.1: 150997246-150997246 on Build GRCh38
rs754961919 Chr.1: 150997236-150997236 on Build GRCh38
rs940397456 Chr.1: 150997657-150997657 on Build GRCh38
rs768040007 Chr.1: 150997600-150997600 on Build GRCh38
rs753715264 Chr.1: 150997562-150997562 on Build GRCh38
rs770038628 Chr.1: 150997510-150997510 on Build GRCh38
rs948991565 Chr.1: 150997646-150997646 on Build GRCh38
rs770989794 Chr.1: 150997293-150997293 on Build GRCh38
rs41299641 Chr.1: 150997606-150997606 on Build GRCh38
rs2925740 Chr.1: 150997516-150997516 on Build GRCh38
rs369462765 Chr.1: 150997573-150997573 on Build GRCh38
rs746735245 Chr.1: 150997581-150997581 on Build GRCh38
rs370021640 Chr.1: 150997318-150997318 on Build GRCh38
rs781517420 Chr.1: 150997503-150997503 on Build GRCh38
rs748609243 Chr.1: 150997507-150997507 on Build GRCh38
rs779194906 Chr.1: 150997248-150997248 on Build GRCh38
rs758561849 Chr.1: 150997484-150997484 on Build GRCh38
rs587682805 Chr.1: 150997385-150997385 on Build GRCh38
rs587715402 Chr.1: 150997571-150997571 on Build GRCh38
rs911946033 Chr.1: 150997222-150997222 on Build GRCh38
rs745888286 Chr.1: 150997299-150997299 on Build GRCh38
rs774237923 Chr.1: 150997555-150997555 on Build GRCh38
rs771026745 Chr.1: 150997436-150997436 on Build GRCh38
rs770653066 Chr.1: 150997360-150997360 on Build GRCh38
rs752668722 Chr.1: 150997326-150997326 on Build GRCh38
rs994991817 Chr.1: 150997455-150997455 on Build GRCh38
rs587770895 Chr.1: 150997593-150997593 on Build GRCh38
rs770154486 Chr.1: 150997550-150997550 on Build GRCh38
rs373353013 Chr.1: 150997594-150997594 on Build GRCh38
rs750600438 Chr.1: 150997468-150997468 on Build GRCh38
rs761827412 Chr.1: 150997653-150997653 on Build GRCh38
rs769197920 Chr.1: 150997496-150997496 on Build GRCh38
rs202059139 Chr.1: 150997313-150997313 on Build GRCh38
rs778198227 Chr.1: 150997280-150997280 on Build GRCh38
rs1015017543 Chr.1: 150997384-150997384 on Build GRCh38
rs776152857 Chr.1: 150997602-150997602 on Build GRCh38
rs146551660 Chr.1: 150997643-150997643 on Build GRCh38
rs143194224 Chr.1: 150997663-150997663 on Build GRCh38
rs947534164 Chr.1: 150997219-150997219 on Build GRCh38
rs587622796 Chr.1: 150997375-150997375 on Build GRCh38
rs926048788 Chr.1: 150997613-150997613 on Build GRCh38
rs776448856 Chr.1: 150997220-150997220 on Build GRCh38
rs766652221 Chr.1: 150997640-150997640 on Build GRCh38
rs587714270 Chr.1: 150997286-150997286 on Build GRCh38
rs775945770 Chr.1: 150997308-150997308 on Build GRCh38
rs373831013 Chr.1: 150997652-150997652 on Build GRCh38
rs528686852 Chr.1: 150997311-150997311 on Build GRCh38
rs771808299 Chr.1: 150997519-150997519 on Build GRCh38
rs759143563 Chr.1: 150997374-150997374 on Build GRCh38
rs760724246 Chr.1: 150997425-150997425 on Build GRCh38
rs746035331 Chr.1: 150997530-150997530 on Build GRCh38
rs773866607 Chr.1: 150997361-150997361 on Build GRCh38
rs779910860 Chr.1: 150997666-150997666 on Build GRCh38
rs762058839 Chr.1: 150997350-150997350 on Build GRCh38
rs761664475 Chr.1: 150997605-150997605 on Build GRCh38
rs760596095 Chr.1: 150997641-150997641 on Build GRCh38
rs746585268 Chr.1: 150997599-150997599 on Build GRCh38
rs1057374052 Chr.1: 150997328-150997328 on Build GRCh38
rs780114306 Chr.1: 150997485-150997485 on Build GRCh38
rs778982251 Chr.1: 150997334-150997334 on Build GRCh38
rs140375829 Chr.1: 150997338-150997338 on Build GRCh38
rs747054097 Chr.1: 150997489-150997489 on Build GRCh38
rs773244809 Chr.1: 150997488-150997488 on Build GRCh38
rs201983888 Chr.1: 150997346-150997346 on Build GRCh38
rs764959774 Chr.1: 150997568-150997568 on Build GRCh38
rs773189956 Chr.1: 150997618-150997618 on Build GRCh38
rs757524284 Chr.1: 150997331-150997331 on Build GRCh38
rs775160219 Chr.1: 150997537-150997537 on Build GRCh38
rs201143690 Chr.1: 150997609-150997609 on Build GRCh38
rs374833584 Chr.1: 150997539-150997539 on Build GRCh38
rs866033020 Chr.1: 150997343-150997343 on Build GRCh38
rs767625772 Chr.1: 150997325-150997325 on Build GRCh38
rs749712491 Chr.1: 150997659-150997659 on Build GRCh38
rs765896143 Chr.1: 150997317-150997317 on Build GRCh38
rs587667595 Chr.1: 150997339-150997339 on Build GRCh38
rs587622055 Chr.1: 150997598-150997598 on Build GRCh38
rs772684343 Chr.1: 150997302-150997302 on Build GRCh38
rs374453452 Chr.1: 150997327-150997327 on Build GRCh38
rs587724501 Chr.1: 150997337-150997337 on Build GRCh38
rs755969150 Chr.1: 150997278-150997278 on Build GRCh38
rs747516882 Chr.1: 150997412-150997412 on Build GRCh38
rs748614766 Chr.1: 150997585-150997585 on Build GRCh38
rs138121177 Chr.1: 150997633-150997633 on Build GRCh38
rs761099204 Chr.1: 150997310-150997310 on Build GRCh38
rs769641959 Chr.1: 150997454-150997455 on Build GRCh38
rs763306295 Chr.1: 150997505-150997505 on Build GRCh38
rs759266306 Chr.1: 150997517-150997517 on Build GRCh38
rs747991776 Chr.1: 150997269-150997269 on Build GRCh38
rs756972205 Chr.1: 150997442-150997442 on Build GRCh38
rs781750145 Chr.1: 150997579-150997579 on Build GRCh38
rs779263570 Chr.1: 150997457-150997457 on Build GRCh38
rs764076257 Chr.1: 150997429-150997429 on Build GRCh38
rs370795454 Chr.1: 150997647-150997647 on Build GRCh38

View Full Product Details