Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 27761469-27762016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM76A
Entrez Gene ID
199870
Gene Name
family with sequence similarity 76 member A
Gene Aliases
-
UniGene
Hs.469359
Gene Chromosome Location
Chr.1: 27725979-27762915 on Build GRCh38
Transcripts
NM_001143912.1 NM_001143913.1 NM_001143914.1 NM_001143915.1 NM_152660.2

SNP Detail

SNP ID Chromosome Location
rs751156187 Chr.1: 27761821-27761821 on Build GRCh38
rs1014325349 Chr.1: 27761600-27761600 on Build GRCh38
rs561640028 Chr.1: 27761809-27761809 on Build GRCh38
rs949809159 Chr.1: 27761759-27761759 on Build GRCh38
rs995640691 Chr.1: 27761810-27761823 on Build GRCh38
rs949880877 Chr.1: 27761981-27761981 on Build GRCh38
rs975752495 Chr.1: 27761964-27761964 on Build GRCh38
rs1046858613 Chr.1: 27761787-27761787 on Build GRCh38
rs1032814244 Chr.1: 27761688-27761688 on Build GRCh38
rs777270957 Chr.1: 27761820-27761820 on Build GRCh38
rs886748796 Chr.1: 27761799-27761799 on Build GRCh38
rs759706638 Chr.1: 27761690-27761690 on Build GRCh38
rs955480593 Chr.1: 27761543-27761543 on Build GRCh38
rs1020998123 Chr.1: 27761862-27761862 on Build GRCh38
rs900123512 Chr.1: 27761816-27761816 on Build GRCh38
rs1045386122 Chr.1: 27761965-27761968 on Build GRCh38
rs937162965 Chr.1: 27761714-27761714 on Build GRCh38
rs917056703 Chr.1: 27761924-27761924 on Build GRCh38
rs553121806 Chr.1: 27761621-27761621 on Build GRCh38
rs541026619 Chr.1: 27761597-27761597 on Build GRCh38
rs138139515 Chr.1: 27761640-27761640 on Build GRCh38
rs1055671005 Chr.1: 27761732-27761732 on Build GRCh38
rs755947488 Chr.1: 27761964-27761965 on Build GRCh38
rs979047086 Chr.1: 27761704-27761704 on Build GRCh38
rs896757112 Chr.1: 27761517-27761517 on Build GRCh38
rs752434064 Chr.1: 27761594-27761594 on Build GRCh38
rs979490520 Chr.1: 27761883-27761883 on Build GRCh38
rs765446847 Chr.1: 27761799-27761800 on Build GRCh38
rs867902430 Chr.1: 27761801-27761801 on Build GRCh38
rs968040375 Chr.1: 27761689-27761689 on Build GRCh38
rs780728841 Chr.1: 27761890-27761890 on Build GRCh38
rs925900361 Chr.1: 27761706-27761706 on Build GRCh38
rs79541154 Chr.1: 27761965-27761965 on Build GRCh38
rs185312813 Chr.1: 27761834-27761834 on Build GRCh38
rs1029816050 Chr.1: 27761823-27761823 on Build GRCh38
rs968419284 Chr.1: 27761863-27761863 on Build GRCh38
rs370731107 Chr.1: 27761889-27761889 on Build GRCh38
rs1001238074 Chr.1: 27761987-27761987 on Build GRCh38
rs905535747 Chr.1: 27761968-27761968 on Build GRCh38
rs376304703 Chr.1: 27761882-27761882 on Build GRCh38
rs192524571 Chr.1: 27761592-27761592 on Build GRCh38
rs909339722 Chr.1: 27761833-27761833 on Build GRCh38
rs913876891 Chr.1: 27761646-27761646 on Build GRCh38
rs976463873 Chr.1: 27761783-27761783 on Build GRCh38
rs1009583546 Chr.1: 27761846-27761846 on Build GRCh38
rs1045439548 Chr.1: 27761637-27761637 on Build GRCh38
rs114586119 Chr.1: 27761907-27761907 on Build GRCh38
rs957426894 Chr.1: 27761699-27761699 on Build GRCh38
rs758129603 Chr.1: 27761683-27761683 on Build GRCh38
rs916982678 Chr.1: 27761733-27761733 on Build GRCh38
rs1036560568 Chr.1: 27761894-27761894 on Build GRCh38
rs763253518 Chr.1: 27761659-27761659 on Build GRCh38
rs958672674 Chr.1: 27761906-27761906 on Build GRCh38
rs949681533 Chr.1: 27761934-27761934 on Build GRCh38

View Full Product Details