Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 30012741-30013256 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKBP14
Entrez Gene ID
55033
Gene Name
FK506 binding protein 14
Gene Aliases
EDSKMH, FKBP22, IPBP12
UniGene
Hs.390838
Gene Chromosome Location
Chr.7: 30010583-30026801 on Build GRCh38
Transcripts
NM_017946.3 NR_046478.1 NR_046479.1

SNP Detail

SNP ID Chromosome Location
rs965733610 Chr.7: 30013186-30013186 on Build GRCh38
rs527830842 Chr.7: 30012920-30012920 on Build GRCh38
rs941801701 Chr.7: 30012832-30012832 on Build GRCh38
rs375404260 Chr.7: 30012856-30012856 on Build GRCh38
rs906163020 Chr.7: 30013150-30013150 on Build GRCh38
rs1047562766 Chr.7: 30012798-30012798 on Build GRCh38
rs947767926 Chr.7: 30013113-30013113 on Build GRCh38
rs914814671 Chr.7: 30012875-30012875 on Build GRCh38
rs535406089 Chr.7: 30012781-30012781 on Build GRCh38
rs990253525 Chr.7: 30013230-30013230 on Build GRCh38
rs997630997 Chr.7: 30013190-30013190 on Build GRCh38
rs1007486192 Chr.7: 30013098-30013098 on Build GRCh38
rs1037113705 Chr.7: 30013026-30013026 on Build GRCh38
rs956013387 Chr.7: 30013221-30013221 on Build GRCh38
rs1011061634 Chr.7: 30012993-30012993 on Build GRCh38
rs906786664 Chr.7: 30012905-30012905 on Build GRCh38
rs938714838 Chr.7: 30012985-30012985 on Build GRCh38
rs973201398 Chr.7: 30013092-30013092 on Build GRCh38
rs116582283 Chr.7: 30013065-30013065 on Build GRCh38
rs897253499 Chr.7: 30013050-30013050 on Build GRCh38
rs79160784 Chr.7: 30013044-30013044 on Build GRCh38
rs191020625 Chr.7: 30013070-30013070 on Build GRCh38
rs924390656 Chr.7: 30012765-30012765 on Build GRCh38
rs894051432 Chr.7: 30013189-30013189 on Build GRCh38
rs907777187 Chr.7: 30013066-30013066 on Build GRCh38
rs550866227 Chr.7: 30013188-30013188 on Build GRCh38
rs1018401437 Chr.7: 30013003-30013003 on Build GRCh38
rs777197249 Chr.7: 30012896-30012896 on Build GRCh38
rs562856101 Chr.7: 30012812-30012812 on Build GRCh38
rs907670034 Chr.7: 30012820-30012820 on Build GRCh38
rs1046484593 Chr.7: 30012898-30012898 on Build GRCh38
rs990302589 Chr.7: 30012844-30012844 on Build GRCh38
rs185579614 Chr.7: 30012843-30012843 on Build GRCh38
rs964118638 Chr.7: 30013004-30013004 on Build GRCh38
rs1044335143 Chr.7: 30012918-30012918 on Build GRCh38
rs899923519 Chr.7: 30013200-30013200 on Build GRCh38
rs887671288 Chr.7: 30013072-30013072 on Build GRCh38
rs976821193 Chr.7: 30013008-30013008 on Build GRCh38
rs947756574 Chr.7: 30012895-30012895 on Build GRCh38
rs893955563 Chr.7: 30012986-30012986 on Build GRCh38
rs538260831 Chr.7: 30013102-30013102 on Build GRCh38
rs554904998 Chr.7: 30012864-30012864 on Build GRCh38
rs560591872 Chr.7: 30012914-30012914 on Build GRCh38
rs1030040299 Chr.7: 30013013-30013013 on Build GRCh38
rs1046072406 Chr.7: 30013141-30013142 on Build GRCh38
rs1017657336 Chr.7: 30013153-30013153 on Build GRCh38
rs763106845 Chr.7: 30013192-30013192 on Build GRCh38
rs954661221 Chr.7: 30013058-30013058 on Build GRCh38

View Full Product Details