Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32868193-32868693 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FNDC5
Entrez Gene ID
252995
Gene Name
fibronectin type III domain containing 5
Gene Aliases
FRCP2, irisin
UniGene
Hs.524234
Gene Chromosome Location
Chr.1: 32862268-32872492 on Build GRCh38
Transcripts
NM_001171940.1 NM_001171941.2 NM_153756.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6665914 c.71C>T p.T24M Chr.1: 32868303-32868303 on Build GRCh38
1197152 c.19G>A p.E7K Chr.1: 32868355-32868355 on Build GRCh38
426114 c.99C>T p.I33I Chr.1: 32868275-32868275 on Build GRCh38
3488237 c.127G>A p.E43K Chr.1: 32868247-32868247 on Build GRCh38
908365 c.72G>A p.T24T Chr.1: 32868302-32868302 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775897042 Chr.1: 32868383-32868385 on Build GRCh38
rs763883177 Chr.1: 32868436-32868436 on Build GRCh38
rs201963189 Chr.1: 32868224-32868224 on Build GRCh38
rs765656287 Chr.1: 32868339-32868339 on Build GRCh38
rs1017230244 Chr.1: 32868632-32868633 on Build GRCh38
rs376090198 Chr.1: 32868352-32868352 on Build GRCh38
rs1031880380 Chr.1: 32868367-32868367 on Build GRCh38
rs769640098 Chr.1: 32868247-32868247 on Build GRCh38
rs1031372438 Chr.1: 32868522-32868522 on Build GRCh38
rs537650655 Chr.1: 32868446-32868446 on Build GRCh38
rs760769158 Chr.1: 32868400-32868400 on Build GRCh38
rs1057382663 Chr.1: 32868289-32868289 on Build GRCh38
rs185406506 Chr.1: 32868401-32868401 on Build GRCh38
rs140092865 Chr.1: 32868366-32868366 on Build GRCh38
rs373917435 Chr.1: 32868375-32868375 on Build GRCh38
rs777659320 Chr.1: 32868378-32868378 on Build GRCh38
rs780955257 Chr.1: 32868223-32868223 on Build GRCh38
rs369474610 Chr.1: 32868222-32868222 on Build GRCh38
rs750850228 Chr.1: 32868422-32868422 on Build GRCh38
rs993583717 Chr.1: 32868454-32868454 on Build GRCh38
rs578047041 Chr.1: 32868617-32868617 on Build GRCh38
rs898810264 Chr.1: 32868449-32868449 on Build GRCh38
rs754092673 Chr.1: 32868303-32868303 on Build GRCh38
rs938634301 Chr.1: 32868442-32868442 on Build GRCh38
rs533821354 Chr.1: 32868423-32868423 on Build GRCh38
rs747854483 Chr.1: 32868370-32868370 on Build GRCh38
rs772243605 Chr.1: 32868287-32868287 on Build GRCh38
rs973339695 Chr.1: 32868665-32868665 on Build GRCh38
rs567452472 Chr.1: 32868437-32868437 on Build GRCh38
rs1021798225 Chr.1: 32868337-32868337 on Build GRCh38
rs1008495904 Chr.1: 32868609-32868609 on Build GRCh38
rs779570641 Chr.1: 32868215-32868215 on Build GRCh38
rs563454230 Chr.1: 32868479-32868479 on Build GRCh38
rs765488784 Chr.1: 32868411-32868411 on Build GRCh38
rs775706100 Chr.1: 32868396-32868396 on Build GRCh38
rs149496621 Chr.1: 32868326-32868326 on Build GRCh38
rs771047914 Chr.1: 32868394-32868394 on Build GRCh38
rs529163611 Chr.1: 32868504-32868504 on Build GRCh38
rs776667620 Chr.1: 32868403-32868403 on Build GRCh38
rs377691602 Chr.1: 32868248-32868248 on Build GRCh38
rs768347509 Chr.1: 32868219-32868219 on Build GRCh38
rs929167213 Chr.1: 32868431-32868431 on Build GRCh38
rs1047622425 Chr.1: 32868244-32868244 on Build GRCh38
rs922689293 Chr.1: 32868409-32868409 on Build GRCh38
rs374085063 Chr.1: 32868342-32868342 on Build GRCh38
rs370511876 Chr.1: 32868369-32868369 on Build GRCh38
rs370602401 Chr.1: 32868410-32868410 on Build GRCh38
rs746635434 Chr.1: 32868217-32868217 on Build GRCh38
rs375151828 Chr.1: 32868361-32868361 on Build GRCh38
rs964378297 Chr.1: 32868660-32868660 on Build GRCh38
rs758944873 Chr.1: 32868348-32868348 on Build GRCh38
rs1026029820 Chr.1: 32868455-32868455 on Build GRCh38
rs772354615 Chr.1: 32868299-32868299 on Build GRCh38
rs371060777 Chr.1: 32868302-32868302 on Build GRCh38
rs1033468585 Chr.1: 32868666-32868666 on Build GRCh38
rs532898911 Chr.1: 32868294-32868294 on Build GRCh38
rs761156654 Chr.1: 32868399-32868400 on Build GRCh38
rs890218748 Chr.1: 32868525-32868525 on Build GRCh38
rs555282437 Chr.1: 32868424-32868424 on Build GRCh38
rs190082581 Chr.1: 32868495-32868495 on Build GRCh38
rs1003335089 Chr.1: 32868642-32868642 on Build GRCh38
rs753519854 Chr.1: 32868453-32868453 on Build GRCh38
rs762976068 Chr.1: 32868286-32868286 on Build GRCh38
rs999456378 Chr.1: 32868515-32868515 on Build GRCh38
rs368098828 Chr.1: 32868385-32868385 on Build GRCh38

View Full Product Details