Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 55625162-55625660 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXR2
Entrez Gene ID
139628
Gene Name
forkhead box R2
Gene Aliases
FOXN6
UniGene
Hs.364260
Gene Chromosome Location
Chr.X: 55623400-55626188 on Build GRCh38
Transcripts
NM_198451.3

SNP Detail

SNP ID Chromosome Location
rs987463472 Chr.X: 55625207-55625207 on Build GRCh38
rs147042315 Chr.X: 55625371-55625371 on Build GRCh38
rs889869713 Chr.X: 55625622-55625622 on Build GRCh38
rs749019104 Chr.X: 55625253-55625253 on Build GRCh38
rs770019507 Chr.X: 55625609-55625609 on Build GRCh38
rs761473834 Chr.X: 55625238-55625238 on Build GRCh38
rs1019227682 Chr.X: 55625613-55625613 on Build GRCh38
rs901734667 Chr.X: 55625442-55625442 on Build GRCh38
rs935829005 Chr.X: 55625356-55625356 on Build GRCh38
rs913140563 Chr.X: 55625372-55625372 on Build GRCh38
rs1018925688 Chr.X: 55625263-55625263 on Build GRCh38
rs946011426 Chr.X: 55625416-55625416 on Build GRCh38
rs764826929 Chr.X: 55625276-55625276 on Build GRCh38
rs1054332319 Chr.X: 55625362-55625362 on Build GRCh38
rs1047961207 Chr.X: 55625591-55625591 on Build GRCh38
rs944227729 Chr.X: 55625390-55625390 on Build GRCh38
rs780521741 Chr.X: 55625312-55625312 on Build GRCh38
rs995014382 Chr.X: 55625582-55625582 on Build GRCh38
rs777729231 Chr.X: 55625201-55625201 on Build GRCh38
rs988642077 Chr.X: 55625357-55625357 on Build GRCh38
rs973383038 Chr.X: 55625217-55625217 on Build GRCh38
rs781561084 Chr.X: 55625522-55625522 on Build GRCh38
rs1040674768 Chr.X: 55625428-55625428 on Build GRCh38
rs949772264 Chr.X: 55625246-55625246 on Build GRCh38
rs749901548 Chr.X: 55625301-55625301 on Build GRCh38
rs926936942 Chr.X: 55625420-55625421 on Build GRCh38
rs886617814 Chr.X: 55625603-55625603 on Build GRCh38
rs191026427 Chr.X: 55625564-55625564 on Build GRCh38
rs998605878 Chr.X: 55625443-55625443 on Build GRCh38
rs755443206 Chr.X: 55625408-55625408 on Build GRCh38
rs926056759 Chr.X: 55625315-55625315 on Build GRCh38
rs937017052 Chr.X: 55625424-55625424 on Build GRCh38
rs73489626 Chr.X: 55625311-55625311 on Build GRCh38
rs898263082 Chr.X: 55625557-55625558 on Build GRCh38

View Full Product Details