Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154533089-154533596 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
G6PD
Entrez Gene ID
2539
Gene Name
glucose-6-phosphate dehydrogenase
Gene Aliases
G6PD1
UniGene
Hs.461047
Gene Chromosome Location
Chr.X: 154531390-154547586 on Build GRCh38
Transcripts
NM_000402.4 NM_001042351.2

SNP Detail

SNP ID Chromosome Location
rs373339066 Chr.X: 154533170-154533170 on Build GRCh38
rs781997386 Chr.X: 154533146-154533146 on Build GRCh38
rs947021060 Chr.X: 154533519-154533519 on Build GRCh38
rs1014919446 Chr.X: 154533392-154533392 on Build GRCh38
rs183394670 Chr.X: 154533300-154533300 on Build GRCh38
rs1049521635 Chr.X: 154533218-154533218 on Build GRCh38
rs782416820 Chr.X: 154533562-154533562 on Build GRCh38
rs188061086 Chr.X: 154533350-154533350 on Build GRCh38
rs782012531 Chr.X: 154533471-154533471 on Build GRCh38
rs782017285 Chr.X: 154533568-154533568 on Build GRCh38
rs781963210 Chr.X: 154533572-154533572 on Build GRCh38
rs199970830 Chr.X: 154533144-154533144 on Build GRCh38
rs782256985 Chr.X: 154533569-154533569 on Build GRCh38
rs782640481 Chr.X: 154533535-154533535 on Build GRCh38
rs782743045 Chr.X: 154533167-154533167 on Build GRCh38
rs1052746376 Chr.X: 154533284-154533284 on Build GRCh38
rs372876649 Chr.X: 154533571-154533571 on Build GRCh38
rs781934049 Chr.X: 154533528-154533528 on Build GRCh38
rs781813390 Chr.X: 154533178-154533178 on Build GRCh38
rs912881178 Chr.X: 154533518-154533518 on Build GRCh38
rs377041776 Chr.X: 154533559-154533559 on Build GRCh38
rs73573478 Chr.X: 154533349-154533349 on Build GRCh38
rs782794403 Chr.X: 154533145-154533145 on Build GRCh38
rs922152752 Chr.X: 154533435-154533435 on Build GRCh38
rs191030887 Chr.X: 154533382-154533382 on Build GRCh38
rs782236330 Chr.X: 154533390-154533390 on Build GRCh38
rs782440230 Chr.X: 154533180-154533180 on Build GRCh38
rs200071953 Chr.X: 154533531-154533531 on Build GRCh38
rs782114455 Chr.X: 154533160-154533160 on Build GRCh38
rs137852327 Chr.X: 154533122-154533122 on Build GRCh38
rs953537892 Chr.X: 154533436-154533436 on Build GRCh38
rs782157687 Chr.X: 154533473-154533473 on Build GRCh38
rs782192379 Chr.X: 154533295-154533295 on Build GRCh38
rs782476171 Chr.X: 154533555-154533555 on Build GRCh38
rs1035747058 Chr.X: 154533375-154533375 on Build GRCh38
rs782326802 Chr.X: 154533126-154533126 on Build GRCh38
rs782714017 Chr.X: 154533567-154533568 on Build GRCh38
rs987601506 Chr.X: 154533472-154533472 on Build GRCh38
rs973662474 Chr.X: 154533415-154533415 on Build GRCh38
rs912286263 Chr.X: 154533220-154533220 on Build GRCh38
rs782339848 Chr.X: 154533380-154533380 on Build GRCh38
rs187815238 Chr.X: 154533488-154533488 on Build GRCh38
rs1004880303 Chr.X: 154533391-154533391 on Build GRCh38
rs943691141 Chr.X: 154533281-154533281 on Build GRCh38
rs782643504 Chr.X: 154533296-154533296 on Build GRCh38
rs201958777 Chr.X: 154533536-154533536 on Build GRCh38
rs782698698 Chr.X: 154533525-154533525 on Build GRCh38
rs1001296301 Chr.X: 154533354-154533354 on Build GRCh38
rs370066745 Chr.X: 154533560-154533560 on Build GRCh38
rs1042993417 Chr.X: 154533550-154533550 on Build GRCh38
rs5986990 Chr.X: 154533413-154533413 on Build GRCh38
rs943764534 Chr.X: 154533487-154533487 on Build GRCh38
rs781920344 Chr.X: 154533133-154533133 on Build GRCh38
rs782206909 Chr.X: 154533353-154533353 on Build GRCh38

View Full Product Details