Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 46925361-46925896 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRA4
Entrez Gene ID
2557
Gene Name
gamma-aminobutyric acid type A receptor alpha4 subunit
Gene Aliases
-
UniGene
Hs.248112
Gene Chromosome Location
Chr.4: 46918900-46994407 on Build GRCh38
Transcripts
NM_000809.3 NM_001204266.1 NM_001204267.1

SNP Detail

SNP ID Chromosome Location
rs936079656 Chr.4: 46925746-46925746 on Build GRCh38
rs200592932 Chr.4: 46925752-46925760 on Build GRCh38
rs1024269826 Chr.4: 46925583-46925583 on Build GRCh38
rs749993675 Chr.4: 46925715-46925720 on Build GRCh38
rs944989801 Chr.4: 46925739-46925739 on Build GRCh38
rs372102773 Chr.4: 46925834-46925834 on Build GRCh38
rs191949116 Chr.4: 46925549-46925549 on Build GRCh38
rs58229734 Chr.4: 46925763-46925765 on Build GRCh38
rs543152729 Chr.4: 46925486-46925486 on Build GRCh38
rs528848022 Chr.4: 46925551-46925551 on Build GRCh38
rs920244747 Chr.4: 46925645-46925645 on Build GRCh38
rs940379048 Chr.4: 46925840-46925840 on Build GRCh38
rs1051462700 Chr.4: 46925815-46925815 on Build GRCh38
rs981912854 Chr.4: 46925530-46925530 on Build GRCh38
rs549999235 Chr.4: 46925850-46925850 on Build GRCh38
rs921678822 Chr.4: 46925749-46925763 on Build GRCh38
rs753953804 Chr.4: 46925715-46925717 on Build GRCh38
rs1027365925 Chr.4: 46925813-46925813 on Build GRCh38
rs10004905 Chr.4: 46925869-46925869 on Build GRCh38
rs959224574 Chr.4: 46925793-46925793 on Build GRCh38
rs184086427 Chr.4: 46925568-46925568 on Build GRCh38
rs12643517 Chr.4: 46925760-46925760 on Build GRCh38
rs1032972874 Chr.4: 46925425-46925425 on Build GRCh38
rs558083648 Chr.4: 46925426-46925426 on Build GRCh38
rs906435758 Chr.4: 46925763-46925763 on Build GRCh38
rs1033540853 Chr.4: 46925539-46925539 on Build GRCh38
rs1014260652 Chr.4: 46925567-46925567 on Build GRCh38
rs922145639 Chr.4: 46925745-46925745 on Build GRCh38
rs200726027 Chr.4: 46925757-46925757 on Build GRCh38
rs71611987 Chr.4: 46925754-46925754 on Build GRCh38
rs770093380 Chr.4: 46925558-46925558 on Build GRCh38
rs912120361 Chr.4: 46925677-46925677 on Build GRCh38
rs995437186 Chr.4: 46925494-46925494 on Build GRCh38
rs891830565 Chr.4: 46925449-46925449 on Build GRCh38
rs561866022 Chr.4: 46925498-46925498 on Build GRCh38
rs971070610 Chr.4: 46925527-46925527 on Build GRCh38
rs146047945 Chr.4: 46925844-46925844 on Build GRCh38
rs28524855 Chr.4: 46925516-46925516 on Build GRCh38
rs576043843 Chr.4: 46925446-46925446 on Build GRCh38
rs533075645 Chr.4: 46925675-46925675 on Build GRCh38
rs115835211 Chr.4: 46925386-46925386 on Build GRCh38
rs973187201 Chr.4: 46925614-46925614 on Build GRCh38
rs1034424577 Chr.4: 46925526-46925526 on Build GRCh38
rs928951443 Chr.4: 46925657-46925657 on Build GRCh38
rs753643936 Chr.4: 46925754-46925755 on Build GRCh38
rs967377480 Chr.4: 46925603-46925604 on Build GRCh38
rs1039163345 Chr.4: 46925742-46925742 on Build GRCh38
rs188423139 Chr.4: 46925817-46925817 on Build GRCh38
rs891878904 Chr.4: 46925751-46925751 on Build GRCh38
rs958810442 Chr.4: 46925550-46925550 on Build GRCh38
rs34991134 Chr.4: 46925757-46925758 on Build GRCh38
rs868582638 Chr.4: 46925748-46925748 on Build GRCh38
rs754203959 Chr.4: 46925718-46925720 on Build GRCh38
rs959092612 Chr.4: 46925611-46925611 on Build GRCh38
rs994680075 Chr.4: 46925796-46925796 on Build GRCh38
rs180932227 Chr.4: 46925856-46925856 on Build GRCh38
rs750051703 Chr.4: 46925758-46925760 on Build GRCh38
rs978007914 Chr.4: 46925655-46925655 on Build GRCh38
rs1003484865 Chr.4: 46925766-46925766 on Build GRCh38
rs957879369 Chr.4: 46925681-46925681 on Build GRCh38
rs1033508243 Chr.4: 46925768-46925768 on Build GRCh38
rs931772816 Chr.4: 46925749-46925749 on Build GRCh38
rs559195565 Chr.4: 46925581-46925581 on Build GRCh38
rs546390863 Chr.4: 46925392-46925392 on Build GRCh38
rs551515697 Chr.4: 46925701-46925701 on Build GRCh38
rs397993248 Chr.4: 46925754-46925759 on Build GRCh38
rs760544331 Chr.4: 46925746-46925760 on Build GRCh38
rs58632284 Chr.4: 46925757-46925762 on Build GRCh38
rs928918943 Chr.4: 46925836-46925836 on Build GRCh38
rs138704036 Chr.4: 46925755-46925760 on Build GRCh38
rs1000262299 Chr.4: 46925415-46925415 on Build GRCh38
rs1015605328 Chr.4: 46925482-46925482 on Build GRCh38

View Full Product Details