Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 152645315-152645723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRQ
Entrez Gene ID
55879
Gene Name
gamma-aminobutyric acid type A receptor theta subunit
Gene Aliases
THETA
UniGene
Hs.283081
Gene Chromosome Location
Chr.X: 152637895-152659827 on Build GRCh38
Transcripts
NM_018558.3 XM_011531184.2

SNP Detail

SNP ID Chromosome Location
rs190746951 Chr.X: 152645526-152645526 on Build GRCh38
rs782197823 Chr.X: 152645640-152645640 on Build GRCh38
rs782590606 Chr.X: 152645534-152645534 on Build GRCh38
rs782655114 Chr.X: 152645566-152645566 on Build GRCh38
rs782810481 Chr.X: 152645439-152645439 on Build GRCh38
rs782059013 Chr.X: 152645429-152645429 on Build GRCh38
rs781839250 Chr.X: 152645512-152645512 on Build GRCh38
rs929942438 Chr.X: 152645448-152645448 on Build GRCh38
rs781976474 Chr.X: 152645356-152645356 on Build GRCh38
rs57920078 Chr.X: 152645432-152645432 on Build GRCh38
rs782234412 Chr.X: 152645591-152645591 on Build GRCh38
rs782032322 Chr.X: 152645579-152645579 on Build GRCh38
rs187433627 Chr.X: 152645693-152645693 on Build GRCh38
rs781889580 Chr.X: 152645482-152645482 on Build GRCh38
rs370082575 Chr.X: 152645485-152645485 on Build GRCh38
rs782548151 Chr.X: 152645554-152645554 on Build GRCh38
rs974721158 Chr.X: 152645352-152645352 on Build GRCh38
rs782595019 Chr.X: 152645607-152645607 on Build GRCh38
rs183267300 Chr.X: 152645647-152645647 on Build GRCh38
rs781799277 Chr.X: 152645538-152645538 on Build GRCh38
rs782755909 Chr.X: 152645500-152645500 on Build GRCh38
rs918629450 Chr.X: 152645408-152645408 on Build GRCh38
rs782350782 Chr.X: 152645603-152645603 on Build GRCh38
rs374323478 Chr.X: 152645499-152645499 on Build GRCh38

View Full Product Details