Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 113821846-113822346 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAS6
Entrez Gene ID
2621
Gene Name
growth arrest specific 6
Gene Aliases
AXLLG, AXSF
UniGene
Hs.646346
Gene Chromosome Location
Chr.13: 113820549-113864103 on Build GRCh38
Transcripts
NM_000820.3 NR_044995.2

Gene Symbol
GAS6-AS1
Entrez Gene ID
650669
Gene Name
GAS6 antisense RNA 1
Gene Aliases
-
UniGene
Hs.369201
Gene Chromosome Location
Chr.13: 113815610-113842841 on Build GRCh38
Transcripts
NM_000820.3 NR_044995.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4213294 c.1551C>T p.D517D Chr.13: 113822127-113822127 on Build GRCh38
4046235 c.1710C>T p.C570C Chr.13: 113822130-113822130 on Build GRCh38
5182463 c.1792G>A p.E598K Chr.13: 113822048-113822048 on Build GRCh38
6406228 c.1810G>T p.A604S Chr.13: 113822159-113822159 on Build GRCh38
7233976 c.1863C>T p.T621T Chr.13: 113822106-113822106 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576092871 Chr.13: 113821932-113821932 on Build GRCh38
rs776530935 Chr.13: 113822251-113822251 on Build GRCh38
rs1010187128 Chr.13: 113822297-113822297 on Build GRCh38
rs372042104 Chr.13: 113822091-113822091 on Build GRCh38
rs997282555 Chr.13: 113822281-113822281 on Build GRCh38
rs796413690 Chr.13: 113822002-113822002 on Build GRCh38
rs769949299 Chr.13: 113822078-113822078 on Build GRCh38
rs772789572 Chr.13: 113822188-113822188 on Build GRCh38
rs751493237 Chr.13: 113822125-113822125 on Build GRCh38
rs369772467 Chr.13: 113822201-113822201 on Build GRCh38
rs769422804 Chr.13: 113822184-113822184 on Build GRCh38
rs374910916 Chr.13: 113822114-113822114 on Build GRCh38
rs747132469 Chr.13: 113821943-113821943 on Build GRCh38
rs1010087610 Chr.13: 113822071-113822071 on Build GRCh38
rs371536576 Chr.13: 113822012-113822012 on Build GRCh38
rs1021197379 Chr.13: 113822298-113822298 on Build GRCh38
rs375892497 Chr.13: 113822171-113822171 on Build GRCh38
rs887499688 Chr.13: 113821945-113821945 on Build GRCh38
rs201844484 Chr.13: 113821938-113821938 on Build GRCh38
rs143209378 Chr.13: 113822139-113822139 on Build GRCh38
rs368109588 Chr.13: 113822129-113822129 on Build GRCh38
rs73583241 Chr.13: 113822006-113822006 on Build GRCh38
rs11619074 Chr.13: 113822288-113822288 on Build GRCh38
rs557741567 Chr.13: 113822153-113822153 on Build GRCh38
rs764747945 Chr.13: 113822035-113822035 on Build GRCh38
rs1025238448 Chr.13: 113822286-113822286 on Build GRCh38
rs148773342 Chr.13: 113822039-113822039 on Build GRCh38
rs909157428 Chr.13: 113822256-113822256 on Build GRCh38
rs773443142 Chr.13: 113822079-113822079 on Build GRCh38
rs537195128 Chr.13: 113822259-113822259 on Build GRCh38
rs370222268 Chr.13: 113821939-113821939 on Build GRCh38
rs144262744 Chr.13: 113822036-113822036 on Build GRCh38
rs781534168 Chr.13: 113822066-113822066 on Build GRCh38
rs1017389995 Chr.13: 113821968-113821968 on Build GRCh38
rs910912705 Chr.13: 113821878-113821878 on Build GRCh38
rs993536277 Chr.13: 113821930-113821930 on Build GRCh38
rs566401340 Chr.13: 113822127-113822127 on Build GRCh38
rs768916777 Chr.13: 113821962-113821962 on Build GRCh38
rs879218442 Chr.13: 113822219-113822219 on Build GRCh38
rs371231729 Chr.13: 113822136-113822136 on Build GRCh38
rs1000779799 Chr.13: 113822323-113822323 on Build GRCh38
rs763543156 Chr.13: 113821994-113821994 on Build GRCh38
rs762426948 Chr.13: 113822115-113822115 on Build GRCh38
rs752540081 Chr.13: 113822231-113822231 on Build GRCh38
rs7400002 Chr.13: 113821971-113821971 on Build GRCh38
rs761466951 Chr.13: 113822105-113822105 on Build GRCh38
rs141503833 Chr.13: 113822067-113822067 on Build GRCh38
rs1051980657 Chr.13: 113822284-113822284 on Build GRCh38
rs199700915 Chr.13: 113821993-113821993 on Build GRCh38
rs979434166 Chr.13: 113822172-113822172 on Build GRCh38
rs200023591 Chr.13: 113822070-113822070 on Build GRCh38
rs752912172 Chr.13: 113822130-113822130 on Build GRCh38
rs929158952 Chr.13: 113822244-113822244 on Build GRCh38
rs761095019 Chr.13: 113822180-113822180 on Build GRCh38
rs766320081 Chr.13: 113822118-113822118 on Build GRCh38
rs745832801 Chr.13: 113821931-113821931 on Build GRCh38
rs758358413 Chr.13: 113822037-113822037 on Build GRCh38
rs900231184 Chr.13: 113822280-113822280 on Build GRCh38
rs1009810756 Chr.13: 113822120-113822120 on Build GRCh38
rs146661095 Chr.13: 113822224-113822224 on Build GRCh38
rs1056306286 Chr.13: 113821869-113821869 on Build GRCh38
rs1005987192 Chr.13: 113821952-113821952 on Build GRCh38
rs746808423 Chr.13: 113822040-113822040 on Build GRCh38
rs746380608 Chr.13: 113822165-113822165 on Build GRCh38
rs975612741 Chr.13: 113821972-113821972 on Build GRCh38
rs762493549 Chr.13: 113822199-113822199 on Build GRCh38
rs770035055 Chr.13: 113821982-113821982 on Build GRCh38
rs568667737 Chr.13: 113822090-113822090 on Build GRCh38
rs774786719 Chr.13: 113822007-113822007 on Build GRCh38
rs556822784 Chr.13: 113821965-113821965 on Build GRCh38
rs532835258 Chr.13: 113821947-113821949 on Build GRCh38
rs769241728 Chr.13: 113821953-113821953 on Build GRCh38
rs772364543 Chr.13: 113822168-113822168 on Build GRCh38
rs139044158 Chr.13: 113822160-113822160 on Build GRCh38
rs900260679 Chr.13: 113822082-113822082 on Build GRCh38
rs983913008 Chr.13: 113822308-113822308 on Build GRCh38
rs779434253 Chr.13: 113822161-113822161 on Build GRCh38
rs41307142 Chr.13: 113821870-113821870 on Build GRCh38
rs968276475 Chr.13: 113822320-113822320 on Build GRCh38
rs149588435 Chr.13: 113822034-113822034 on Build GRCh38
rs1030293838 Chr.13: 113821988-113821988 on Build GRCh38
rs146664338 Chr.13: 113822150-113822150 on Build GRCh38
rs892573530 Chr.13: 113822166-113822166 on Build GRCh38
rs1038722615 Chr.13: 113822261-113822261 on Build GRCh38
rs79807310 Chr.13: 113822101-113822101 on Build GRCh38
rs929257792 Chr.13: 113822048-113822048 on Build GRCh38
rs1038795104 Chr.13: 113822075-113822075 on Build GRCh38
rs772453968 Chr.13: 113822100-113822100 on Build GRCh38
rs74663534 Chr.13: 113822049-113822049 on Build GRCh38
rs1018192470 Chr.13: 113822236-113822236 on Build GRCh38
rs1052408040 Chr.13: 113822163-113822163 on Build GRCh38
rs756034822 Chr.13: 113822232-113822232 on Build GRCh38
rs368450891 Chr.13: 113822018-113822018 on Build GRCh38
rs1044187556 Chr.13: 113822195-113822195 on Build GRCh38
rs759562748 Chr.13: 113822221-113822221 on Build GRCh38
rs373917784 Chr.13: 113822220-113822220 on Build GRCh38
rs374150220 Chr.13: 113822013-113822013 on Build GRCh38
rs34975371 Chr.13: 113822106-113822106 on Build GRCh38

View Full Product Details