Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37813201-37813604 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCAT
Entrez Gene ID
23464
Gene Name
glycine C-acetyltransferase
Gene Aliases
KBL
UniGene
Hs.54609
Gene Chromosome Location
Chr.22: 37807893-37817177 on Build GRCh38
Transcripts
NM_001171690.1 NM_014291.3 XM_005261409.3 XM_005261410.3 XM_005261411.1 XM_011530023.1 XM_017028674.1 XM_017028675.1 XM_017028676.1 XM_017028677.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3150862 c.493G>A p.G165S Chr.22: 37813526-37813526 on Build GRCh38
1632590 c.544G>A p.A182T Chr.22: 37813577-37813577 on Build GRCh38
3554273 c.529C>T p.R177C Chr.22: 37813562-37813562 on Build GRCh38
166580 c.492C>T p.D164D Chr.22: 37813525-37813525 on Build GRCh38
3727043 c.527A>G p.Y176C Chr.22: 37813560-37813560 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779032221 Chr.22: 37813373-37813373 on Build GRCh38
rs376802314 Chr.22: 37813444-37813444 on Build GRCh38
rs765485148 Chr.22: 37813547-37813547 on Build GRCh38
rs377504565 Chr.22: 37813266-37813266 on Build GRCh38
rs765429002 Chr.22: 37813494-37813494 on Build GRCh38
rs758410854 Chr.22: 37813498-37813498 on Build GRCh38
rs765428447 Chr.22: 37813295-37813295 on Build GRCh38
rs780257773 Chr.22: 37813346-37813346 on Build GRCh38
rs997412864 Chr.22: 37813510-37813510 on Build GRCh38
rs776812283 Chr.22: 37813544-37813544 on Build GRCh38
rs776004379 Chr.22: 37813227-37813227 on Build GRCh38
rs746747398 Chr.22: 37813409-37813409 on Build GRCh38
rs757053146 Chr.22: 37813563-37813563 on Build GRCh38
rs767850425 Chr.22: 37813321-37813321 on Build GRCh38
rs558384612 Chr.22: 37813273-37813273 on Build GRCh38
rs143093958 Chr.22: 37813297-37813297 on Build GRCh38
rs543986521 Chr.22: 37813329-37813329 on Build GRCh38
rs527590625 Chr.22: 37813487-37813487 on Build GRCh38
rs748730448 Chr.22: 37813527-37813527 on Build GRCh38
rs917601860 Chr.22: 37813387-37813387 on Build GRCh38
rs369469907 Chr.22: 37813422-37813422 on Build GRCh38
rs761044481 Chr.22: 37813483-37813483 on Build GRCh38
rs777739256 Chr.22: 37813403-37813403 on Build GRCh38
rs779625361 Chr.22: 37813576-37813576 on Build GRCh38
rs142027407 Chr.22: 37813525-37813525 on Build GRCh38
rs894198549 Chr.22: 37813475-37813475 on Build GRCh38
rs775013323 Chr.22: 37813526-37813526 on Build GRCh38
rs777706135 Chr.22: 37813503-37813503 on Build GRCh38
rs568423076 Chr.22: 37813580-37813580 on Build GRCh38
rs990492758 Chr.22: 37813385-37813385 on Build GRCh38
rs755016091 Chr.22: 37813532-37813532 on Build GRCh38
rs562487535 Chr.22: 37813340-37813340 on Build GRCh38
rs752834253 Chr.22: 37813556-37813556 on Build GRCh38
rs1027079673 Chr.22: 37813263-37813265 on Build GRCh38
rs867355707 Chr.22: 37813578-37813578 on Build GRCh38
rs572665229 Chr.22: 37813223-37813223 on Build GRCh38
rs776330604 Chr.22: 37813439-37813439 on Build GRCh38
rs931855485 Chr.22: 37813351-37813351 on Build GRCh38
rs181758632 Chr.22: 37813523-37813523 on Build GRCh38
rs202168968 Chr.22: 37813495-37813495 on Build GRCh38
rs759037953 Chr.22: 37813440-37813440 on Build GRCh38
rs921333011 Chr.22: 37813348-37813348 on Build GRCh38
rs949081920 Chr.22: 37813554-37813554 on Build GRCh38
rs753879134 Chr.22: 37813361-37813361 on Build GRCh38
rs938389839 Chr.22: 37813582-37813582 on Build GRCh38
rs979114938 Chr.22: 37813360-37813360 on Build GRCh38
rs755769004 Chr.22: 37813575-37813575 on Build GRCh38
rs748075631 Chr.22: 37813377-37813377 on Build GRCh38
rs780750004 Chr.22: 37813522-37813522 on Build GRCh38
rs762209720 Chr.22: 37813311-37813311 on Build GRCh38
rs896732372 Chr.22: 37813226-37813226 on Build GRCh38
rs878929926 Chr.22: 37813232-37813232 on Build GRCh38
rs61737836 Chr.22: 37813579-37813579 on Build GRCh38
rs750751950 Chr.22: 37813323-37813323 on Build GRCh38
rs775538334 Chr.22: 37813442-37813447 on Build GRCh38
rs144700850 Chr.22: 37813402-37813402 on Build GRCh38
rs781067171 Chr.22: 37813567-37813567 on Build GRCh38
rs754057883 Chr.22: 37813485-37813485 on Build GRCh38
rs1019595268 Chr.22: 37813486-37813486 on Build GRCh38
rs766741339 Chr.22: 37813534-37813534 on Build GRCh38
rs767973127 Chr.22: 37813459-37813459 on Build GRCh38
rs1020069024 Chr.22: 37813284-37813284 on Build GRCh38
rs763133882 Chr.22: 37813455-37813475 on Build GRCh38
rs768801166 Chr.22: 37813550-37813550 on Build GRCh38
rs374476684 Chr.22: 37813477-37813477 on Build GRCh38
rs1006892829 Chr.22: 37813281-37813281 on Build GRCh38
rs574377558 Chr.22: 37813362-37813362 on Build GRCh38
rs759867789 Chr.22: 37813546-37813546 on Build GRCh38
rs995563529 Chr.22: 37813250-37813250 on Build GRCh38
rs1032170268 Chr.22: 37813530-37813530 on Build GRCh38
rs539995753 Chr.22: 37813252-37813252 on Build GRCh38
rs982781647 Chr.22: 37813238-37813238 on Build GRCh38
rs745626193 Chr.22: 37813571-37813571 on Build GRCh38
rs773756902 Chr.22: 37813533-37813533 on Build GRCh38
rs889751819 Chr.22: 37813274-37813274 on Build GRCh38
rs372831337 Chr.22: 37813443-37813443 on Build GRCh38
rs367782960 Chr.22: 37813484-37813484 on Build GRCh38
rs758537417 Chr.22: 37813557-37813557 on Build GRCh38
rs995672095 Chr.22: 37813481-37813481 on Build GRCh38
rs764001412 Chr.22: 37813560-37813560 on Build GRCh38
rs746834386 Chr.22: 37813513-37813513 on Build GRCh38
rs762334184 Chr.22: 37813453-37813453 on Build GRCh38
rs764614970 Chr.22: 37813476-37813476 on Build GRCh38
rs201266387 Chr.22: 37813562-37813562 on Build GRCh38
rs190407011 Chr.22: 37813520-37813520 on Build GRCh38
rs945911859 Chr.22: 37813392-37813392 on Build GRCh38
rs750758530 Chr.22: 37813469-37813469 on Build GRCh38
rs1037188341 Chr.22: 37813231-37813231 on Build GRCh38

View Full Product Details