Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 117867216-117867751 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDAP2
Entrez Gene ID
54834
Gene Name
ganglioside induced differentiation associated protein 2
Gene Aliases
MACROD3
UniGene
Hs.310809
Gene Chromosome Location
Chr.1: 117860227-117930042 on Build GRCh38
Transcripts
NM_017686.3 XM_005270959.2 XM_011541623.2 XM_017001531.1 XM_017001532.1

SNP Detail

SNP ID Chromosome Location
rs570979393 Chr.1: 117867453-117867453 on Build GRCh38
rs191385237 Chr.1: 117867458-117867458 on Build GRCh38
rs138378554 Chr.1: 117867663-117867663 on Build GRCh38
rs1013586326 Chr.1: 117867377-117867377 on Build GRCh38
rs146393941 Chr.1: 117867463-117867463 on Build GRCh38
rs972287534 Chr.1: 117867634-117867634 on Build GRCh38
rs1009337149 Chr.1: 117867563-117867563 on Build GRCh38
rs866186924 Chr.1: 117867413-117867413 on Build GRCh38
rs1042822781 Chr.1: 117867245-117867245 on Build GRCh38
rs991605802 Chr.1: 117867410-117867410 on Build GRCh38
rs752329398 Chr.1: 117867281-117867281 on Build GRCh38
rs1046240529 Chr.1: 117867567-117867567 on Build GRCh38
rs551401909 Chr.1: 117867417-117867417 on Build GRCh38
rs555799066 Chr.1: 117867650-117867650 on Build GRCh38
rs572753170 Chr.1: 117867635-117867635 on Build GRCh38
rs947024825 Chr.1: 117867619-117867619 on Build GRCh38
rs780864168 Chr.1: 117867273-117867273 on Build GRCh38
rs149356083 Chr.1: 117867522-117867522 on Build GRCh38
rs903132003 Chr.1: 117867451-117867451 on Build GRCh38
rs949786397 Chr.1: 117867416-117867416 on Build GRCh38
rs12141200 Chr.1: 117867496-117867496 on Build GRCh38
rs1045368698 Chr.1: 117867445-117867445 on Build GRCh38
rs962270744 Chr.1: 117867295-117867295 on Build GRCh38
rs532444147 Chr.1: 117867543-117867544 on Build GRCh38
rs1005536934 Chr.1: 117867422-117867422 on Build GRCh38
rs557832814 Chr.1: 117867597-117867597 on Build GRCh38
rs1057486658 Chr.1: 117867329-117867329 on Build GRCh38
rs112586804 Chr.1: 117867688-117867688 on Build GRCh38
rs12141108 Chr.1: 117867240-117867240 on Build GRCh38
rs920734050 Chr.1: 117867345-117867345 on Build GRCh38
rs533300886 Chr.1: 117867691-117867691 on Build GRCh38
rs896034638 Chr.1: 117867364-117867364 on Build GRCh38
rs981101932 Chr.1: 117867675-117867675 on Build GRCh38
rs575443476 Chr.1: 117867561-117867561 on Build GRCh38
rs952275661 Chr.1: 117867637-117867637 on Build GRCh38
rs752313294 Chr.1: 117867557-117867557 on Build GRCh38
rs1005484383 Chr.1: 117867724-117867724 on Build GRCh38
rs866663572 Chr.1: 117867440-117867440 on Build GRCh38
rs80342357 Chr.1: 117867243-117867243 on Build GRCh38
rs562672177 Chr.1: 117867307-117867307 on Build GRCh38
rs535309888 Chr.1: 117867543-117867543 on Build GRCh38
rs781046641 Chr.1: 117867614-117867614 on Build GRCh38
rs200807475 Chr.1: 117867546-117867546 on Build GRCh38
rs1029526340 Chr.1: 117867547-117867547 on Build GRCh38
rs1045029126 Chr.1: 117867717-117867717 on Build GRCh38
rs780702482 Chr.1: 117867518-117867518 on Build GRCh38
rs544438655 Chr.1: 117867671-117867671 on Build GRCh38
rs1018945557 Chr.1: 117867576-117867576 on Build GRCh38
rs201890334 Chr.1: 117867701-117867701 on Build GRCh38
rs184417023 Chr.1: 117867579-117867579 on Build GRCh38
rs923106192 Chr.1: 117867528-117867528 on Build GRCh38
rs745964438 Chr.1: 117867527-117867528 on Build GRCh38
rs12141153 Chr.1: 117867353-117867353 on Build GRCh38
rs550734971 Chr.1: 117867263-117867263 on Build GRCh38
rs534151994 Chr.1: 117867455-117867455 on Build GRCh38
rs540185027 Chr.1: 117867648-117867648 on Build GRCh38
rs934595884 Chr.1: 117867694-117867694 on Build GRCh38
rs757987093 Chr.1: 117867577-117867577 on Build GRCh38
rs888720770 Chr.1: 117867718-117867718 on Build GRCh38
rs972256902 Chr.1: 117867318-117867318 on Build GRCh38
rs533237218 Chr.1: 117867411-117867411 on Build GRCh38
rs930431096 Chr.1: 117867352-117867352 on Build GRCh38
rs902966064 Chr.1: 117867262-117867262 on Build GRCh38

View Full Product Details