Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 59257345-59257853 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDPD1
Entrez Gene ID
284161
Gene Name
glycerophosphodiester phosphodiesterase domain containing 1
Gene Aliases
GDE4
UniGene
Hs.631744
Gene Chromosome Location
Chr.17: 59220467-59275970 on Build GRCh38
Transcripts
NM_001165993.1 NM_001165994.1 NM_182569.3 XM_017024521.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7164062 c.548A>T p.Y183F Chr.17: 59257812-59257812 on Build GRCh38
7164064 c.548A>T p.Y183F Chr.17: 59257812-59257812 on Build GRCh38
5429946 c.574G>C p.E192Q Chr.17: 59257838-59257838 on Build GRCh38
982050 c.551A>G p.E184G Chr.17: 59257815-59257815 on Build GRCh38
4878583 c.500T>C p.V167A Chr.17: 59257764-59257764 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777461328 Chr.17: 59257795-59257795 on Build GRCh38
rs765609705 Chr.17: 59257745-59257745 on Build GRCh38
rs757792388 Chr.17: 59257733-59257733 on Build GRCh38
rs984930022 Chr.17: 59257769-59257769 on Build GRCh38
rs778649129 Chr.17: 59257816-59257816 on Build GRCh38
rs777757321 Chr.17: 59257809-59257810 on Build GRCh38
rs763210126 Chr.17: 59257424-59257424 on Build GRCh38
rs568257070 Chr.17: 59257442-59257442 on Build GRCh38
rs925325428 Chr.17: 59257755-59257755 on Build GRCh38
rs745743817 Chr.17: 59257820-59257820 on Build GRCh38
rs754919843 Chr.17: 59257399-59257399 on Build GRCh38
rs550160566 Chr.17: 59257429-59257429 on Build GRCh38
rs965373677 Chr.17: 59257808-59257808 on Build GRCh38
rs371799165 Chr.17: 59257724-59257724 on Build GRCh38
rs200841211 Chr.17: 59257770-59257770 on Build GRCh38
rs1042956007 Chr.17: 59257646-59257646 on Build GRCh38
rs78486108 Chr.17: 59257694-59257694 on Build GRCh38
rs867178472 Chr.17: 59257789-59257789 on Build GRCh38
rs565914568 Chr.17: 59257641-59257641 on Build GRCh38
rs1016635734 Chr.17: 59257583-59257583 on Build GRCh38
rs752759599 Chr.17: 59257730-59257731 on Build GRCh38
rs764672680 Chr.17: 59257425-59257425 on Build GRCh38
rs367660324 Chr.17: 59257637-59257637 on Build GRCh38
rs531563585 Chr.17: 59257428-59257428 on Build GRCh38
rs182563068 Chr.17: 59257496-59257496 on Build GRCh38
rs979373594 Chr.17: 59257392-59257392 on Build GRCh38
rs1006873644 Chr.17: 59257532-59257532 on Build GRCh38
rs891606920 Chr.17: 59257613-59257613 on Build GRCh38
rs201436934 Chr.17: 59257721-59257721 on Build GRCh38
rs996635282 Chr.17: 59257629-59257629 on Build GRCh38
rs771752237 Chr.17: 59257823-59257823 on Build GRCh38
rs938120488 Chr.17: 59257418-59257418 on Build GRCh38
rs986628312 Chr.17: 59257716-59257716 on Build GRCh38
rs754382256 Chr.17: 59257730-59257730 on Build GRCh38
rs540282856 Chr.17: 59257529-59257529 on Build GRCh38
rs775572615 Chr.17: 59257715-59257715 on Build GRCh38
rs867371060 Chr.17: 59257689-59257689 on Build GRCh38
rs1011434910 Chr.17: 59257645-59257645 on Build GRCh38
rs1033735990 Chr.17: 59257378-59257378 on Build GRCh38
rs966679769 Chr.17: 59257744-59257744 on Build GRCh38
rs767785935 Chr.17: 59257704-59257704 on Build GRCh38
rs750809246 Chr.17: 59257746-59257746 on Build GRCh38
rs896680963 Chr.17: 59257499-59257499 on Build GRCh38
rs774650668 Chr.17: 59257611-59257611 on Build GRCh38
rs984982491 Chr.17: 59257487-59257487 on Build GRCh38
rs886668414 Chr.17: 59257531-59257531 on Build GRCh38
rs185365386 Chr.17: 59257747-59257747 on Build GRCh38
rs992321083 Chr.17: 59257405-59257405 on Build GRCh38
rs1039146325 Chr.17: 59257504-59257504 on Build GRCh38
rs149556412 Chr.17: 59257515-59257515 on Build GRCh38
rs748762189 Chr.17: 59257812-59257812 on Build GRCh38
rs974419941 Chr.17: 59257809-59257809 on Build GRCh38
rs945388526 Chr.17: 59257371-59257371 on Build GRCh38
rs898162878 Chr.17: 59257592-59257592 on Build GRCh38
rs756165527 Chr.17: 59257808-59257809 on Build GRCh38

View Full Product Details