Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 85545648-85546164 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGCX
Entrez Gene ID
2677
Gene Name
gamma-glutamyl carboxylase
Gene Aliases
VKCFD1
UniGene
Hs.77719
Gene Chromosome Location
Chr.2: 85544720-85561534 on Build GRCh38
Transcripts
NM_000821.6 NM_001142269.3

SNP Detail

SNP ID Chromosome Location
rs575467767 Chr.2: 85545964-85545964 on Build GRCh38
rs886056359 Chr.2: 85546010-85546010 on Build GRCh38
rs1046049435 Chr.2: 85546004-85546004 on Build GRCh38
rs532034095 Chr.2: 85546142-85546142 on Build GRCh38
rs575928369 Chr.2: 85545729-85545729 on Build GRCh38
rs754152731 Chr.2: 85545692-85545692 on Build GRCh38
rs539831975 Chr.2: 85546124-85546124 on Build GRCh38
rs548192918 Chr.2: 85545874-85545874 on Build GRCh38
rs935697867 Chr.2: 85545734-85545734 on Build GRCh38
rs986672098 Chr.2: 85546093-85546093 on Build GRCh38
rs953939458 Chr.2: 85545901-85545901 on Build GRCh38
rs910760515 Chr.2: 85546012-85546012 on Build GRCh38
rs939882552 Chr.2: 85545919-85545919 on Build GRCh38
rs1036429957 Chr.2: 85545930-85545930 on Build GRCh38
rs536985458 Chr.2: 85545739-85545739 on Build GRCh38
rs972950148 Chr.2: 85546133-85546133 on Build GRCh38
rs538307032 Chr.2: 85545937-85545937 on Build GRCh38
rs995734071 Chr.2: 85545962-85545962 on Build GRCh38
rs922911293 Chr.2: 85545998-85545998 on Build GRCh38
rs920941193 Chr.2: 85545889-85545889 on Build GRCh38
rs912503842 Chr.2: 85546102-85546102 on Build GRCh38
rs746006060 Chr.2: 85545866-85545866 on Build GRCh38
rs962060806 Chr.2: 85545674-85545674 on Build GRCh38
rs953721814 Chr.2: 85546091-85546091 on Build GRCh38
rs1034227829 Chr.2: 85546123-85546123 on Build GRCh38
rs957318099 Chr.2: 85546040-85546040 on Build GRCh38
rs894702615 Chr.2: 85545781-85545781 on Build GRCh38
rs1031404800 Chr.2: 85546036-85546036 on Build GRCh38
rs907368890 Chr.2: 85546010-85546010 on Build GRCh38
rs747381327 Chr.2: 85545969-85545969 on Build GRCh38
rs971566477 Chr.2: 85545820-85545820 on Build GRCh38
rs779065176 Chr.2: 85545749-85545749 on Build GRCh38
rs907097281 Chr.2: 85545895-85545895 on Build GRCh38
rs12473819 Chr.2: 85545938-85545938 on Build GRCh38
rs58540395 Chr.2: 85546059-85546059 on Build GRCh38
rs886056358 Chr.2: 85545719-85545719 on Build GRCh38
rs897821999 Chr.2: 85545827-85545827 on Build GRCh38
rs894483657 Chr.2: 85545970-85545970 on Build GRCh38
rs897882406 Chr.2: 85546026-85546026 on Build GRCh38
rs1002780624 Chr.2: 85546096-85546096 on Build GRCh38
rs183528132 Chr.2: 85545846-85545846 on Build GRCh38
rs912361114 Chr.2: 85545905-85545905 on Build GRCh38
rs944769399 Chr.2: 85545753-85545753 on Build GRCh38
rs927066864 Chr.2: 85545735-85545735 on Build GRCh38
rs762242192 Chr.2: 85546141-85546141 on Build GRCh38
rs999451513 Chr.2: 85546013-85546013 on Build GRCh38
rs971114580 Chr.2: 85545709-85545709 on Build GRCh38
rs1037784960 Chr.2: 85545826-85545826 on Build GRCh38
rs945730741 Chr.2: 85545755-85545755 on Build GRCh38
rs1031875553 Chr.2: 85545876-85545876 on Build GRCh38
rs1049305909 Chr.2: 85545777-85545777 on Build GRCh38
rs897823494 Chr.2: 85545766-85545766 on Build GRCh38
rs147318802 Chr.2: 85545999-85545999 on Build GRCh38
rs933616992 Chr.2: 85546001-85546001 on Build GRCh38
rs1036862377 Chr.2: 85545760-85545760 on Build GRCh38
rs533843636 Chr.2: 85545997-85545997 on Build GRCh38
rs767489176 Chr.2: 85545803-85545804 on Build GRCh38
rs80350214 Chr.2: 85545915-85545915 on Build GRCh38
rs932471497 Chr.2: 85545740-85545740 on Build GRCh38
rs570833364 Chr.2: 85545923-85545923 on Build GRCh38
rs189173899 Chr.2: 85545705-85545705 on Build GRCh38
rs977578767 Chr.2: 85545727-85545727 on Build GRCh38
rs189391524 Chr.2: 85545887-85545887 on Build GRCh38
rs1028362012 Chr.2: 85545679-85545679 on Build GRCh38
rs369547539 Chr.2: 85545744-85545744 on Build GRCh38
rs989898236 Chr.2: 85545947-85545947 on Build GRCh38
rs181889577 Chr.2: 85545815-85545815 on Build GRCh38
rs1034175211 Chr.2: 85545926-85545926 on Build GRCh38
rs897439627 Chr.2: 85545950-85545950 on Build GRCh38
rs990077957 Chr.2: 85545738-85545738 on Build GRCh38
rs576926970 Chr.2: 85546112-85546113 on Build GRCh38
rs991686706 Chr.2: 85545704-85545704 on Build GRCh38
rs542845489 Chr.2: 85545965-85545965 on Build GRCh38
rs981271664 Chr.2: 85545720-85545720 on Build GRCh38
rs768855729 Chr.2: 85546056-85546056 on Build GRCh38
rs995270414 Chr.2: 85545776-85545776 on Build GRCh38
rs957040111 Chr.2: 85545881-85545881 on Build GRCh38
rs12470957 Chr.2: 85546129-85546129 on Build GRCh38
rs558953551 Chr.2: 85545902-85545902 on Build GRCh38
rs1013142583 Chr.2: 85546003-85546003 on Build GRCh38
rs866533240 Chr.2: 85545856-85545856 on Build GRCh38
rs886056360 Chr.2: 85546013-85546013 on Build GRCh38
rs1016299860 Chr.2: 85545672-85545672 on Build GRCh38
rs1013194898 Chr.2: 85545805-85545805 on Build GRCh38

View Full Product Details