Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 85549283-85549794 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGCX
Entrez Gene ID
2677
Gene Name
gamma-glutamyl carboxylase
Gene Aliases
VKCFD1
UniGene
Hs.77719
Gene Chromosome Location
Chr.2: 85544720-85561534 on Build GRCh38
Transcripts
NM_000821.6 NM_001142269.3 XM_005264259.4 XM_011532764.2 XM_011532765.2 XM_017003803.1

SNP Detail

SNP ID Chromosome Location
rs538636383 Chr.2: 85549483-85549483 on Build GRCh38
rs868235570 Chr.2: 85549481-85549481 on Build GRCh38
rs1010215575 Chr.2: 85549456-85549456 on Build GRCh38
rs141959654 Chr.2: 85549375-85549376 on Build GRCh38
rs928737982 Chr.2: 85549424-85549424 on Build GRCh38
rs537175451 Chr.2: 85549388-85549388 on Build GRCh38
rs906037047 Chr.2: 85549428-85549428 on Build GRCh38
rs996606781 Chr.2: 85549654-85549654 on Build GRCh38
rs1041564315 Chr.2: 85549423-85549423 on Build GRCh38
rs533283843 Chr.2: 85549710-85549710 on Build GRCh38
rs80309281 Chr.2: 85549493-85549493 on Build GRCh38
rs987217859 Chr.2: 85549322-85549322 on Build GRCh38
rs893789215 Chr.2: 85549649-85549649 on Build GRCh38
rs45475792 Chr.2: 85549425-85549425 on Build GRCh38
rs1009452924 Chr.2: 85549466-85549466 on Build GRCh38
rs922417113 Chr.2: 85549325-85549325 on Build GRCh38
rs762881732 Chr.2: 85549372-85549373 on Build GRCh38
rs541260755 Chr.2: 85549699-85549699 on Build GRCh38
rs181342571 Chr.2: 85549546-85549546 on Build GRCh38
rs542684610 Chr.2: 85549606-85549606 on Build GRCh38
rs868228293 Chr.2: 85549435-85549435 on Build GRCh38
rs757691991 Chr.2: 85549305-85549305 on Build GRCh38
rs973157941 Chr.2: 85549482-85549482 on Build GRCh38
rs1036683638 Chr.2: 85549643-85549643 on Build GRCh38
rs767804895 Chr.2: 85549439-85549439 on Build GRCh38
rs941086289 Chr.2: 85549623-85549623 on Build GRCh38
rs549113214 Chr.2: 85549411-85549411 on Build GRCh38
rs991404628 Chr.2: 85549568-85549568 on Build GRCh38
rs557756406 Chr.2: 85549465-85549465 on Build GRCh38
rs746577234 Chr.2: 85549478-85549478 on Build GRCh38
rs973043510 Chr.2: 85549364-85549364 on Build GRCh38
rs370169388 Chr.2: 85549743-85549743 on Build GRCh38
rs764210722 Chr.2: 85549313-85549313 on Build GRCh38
rs367980433 Chr.2: 85549327-85549327 on Build GRCh38
rs955084031 Chr.2: 85549321-85549321 on Build GRCh38
rs567829249 Chr.2: 85549430-85549430 on Build GRCh38
rs190583105 Chr.2: 85549692-85549692 on Build GRCh38
rs539761806 Chr.2: 85549596-85549596 on Build GRCh38
rs1009156939 Chr.2: 85549688-85549688 on Build GRCh38
rs532781606 Chr.2: 85549324-85549324 on Build GRCh38
rs551441214 Chr.2: 85549334-85549334 on Build GRCh38
rs772911409 Chr.2: 85549716-85549716 on Build GRCh38
rs1015069135 Chr.2: 85549694-85549694 on Build GRCh38
rs776385209 Chr.2: 85549578-85549578 on Build GRCh38
rs890680499 Chr.2: 85549669-85549669 on Build GRCh38
rs1041492025 Chr.2: 85549590-85549590 on Build GRCh38
rs569744167 Chr.2: 85549360-85549360 on Build GRCh38
rs536803952 Chr.2: 85549650-85549650 on Build GRCh38
rs1053834442 Chr.2: 85549627-85549627 on Build GRCh38
rs5832649 Chr.2: 85549367-85549368 on Build GRCh38
rs937496327 Chr.2: 85549693-85549693 on Build GRCh38
rs553605433 Chr.2: 85549336-85549336 on Build GRCh38
rs1050464340 Chr.2: 85549664-85549664 on Build GRCh38
rs45560635 Chr.2: 85549589-85549589 on Build GRCh38
rs181695165 Chr.2: 85549769-85549769 on Build GRCh38
rs551976064 Chr.2: 85549511-85549512 on Build GRCh38
rs578230076 Chr.2: 85549480-85549480 on Build GRCh38
rs886056376 Chr.2: 85549625-85549625 on Build GRCh38
rs1056150620 Chr.2: 85549398-85549398 on Build GRCh38
rs906033996 Chr.2: 85549672-85549672 on Build GRCh38
rs996439254 Chr.2: 85549462-85549462 on Build GRCh38
rs950014768 Chr.2: 85549588-85549588 on Build GRCh38
rs935408648 Chr.2: 85549597-85549597 on Build GRCh38
rs941004431 Chr.2: 85549358-85549358 on Build GRCh38
rs72843839 Chr.2: 85549709-85549709 on Build GRCh38
rs575253890 Chr.2: 85549550-85549550 on Build GRCh38
rs886056377 Chr.2: 85549664-85549664 on Build GRCh38
rs768419574 Chr.2: 85549517-85549517 on Build GRCh38
rs917087399 Chr.2: 85549400-85549400 on Build GRCh38
rs561050384 Chr.2: 85549668-85549668 on Build GRCh38
rs746348970 Chr.2: 85549464-85549464 on Build GRCh38
rs36102060 Chr.2: 85549368-85549369 on Build GRCh38
rs775090081 Chr.2: 85549372-85549372 on Build GRCh38
rs937207922 Chr.2: 85549389-85549389 on Build GRCh38
rs553383801 Chr.2: 85549471-85549471 on Build GRCh38

View Full Product Details