Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4556820-4557316 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGT6
Entrez Gene ID
124975
Gene Name
gamma-glutamyltransferase 6
Gene Aliases
-
UniGene
Hs.130749
Gene Chromosome Location
Chr.17: 4556927-4560596 on Build GRCh38
Transcripts
NM_001122890.2 NM_001288702.1 NM_001288703.1 NM_001288704.1 NM_153338.3

SNP Detail

SNP ID Chromosome Location
rs183459477 Chr.17: 4557184-4557184 on Build GRCh38
rs778902462 Chr.17: 4557241-4557241 on Build GRCh38
rs3760193 Chr.17: 4557150-4557150 on Build GRCh38
rs751297674 Chr.17: 4556988-4556988 on Build GRCh38
rs1022002526 Chr.17: 4557206-4557206 on Build GRCh38
rs992342328 Chr.17: 4556937-4556937 on Build GRCh38
rs1016423178 Chr.17: 4557222-4557222 on Build GRCh38
rs962319247 Chr.17: 4557226-4557226 on Build GRCh38
rs918123895 Chr.17: 4557287-4557287 on Build GRCh38
rs901933924 Chr.17: 4557158-4557158 on Build GRCh38
rs1010094474 Chr.17: 4557107-4557107 on Build GRCh38
rs997600906 Chr.17: 4556870-4556870 on Build GRCh38
rs893076091 Chr.17: 4557106-4557106 on Build GRCh38
rs962265834 Chr.17: 4556975-4556975 on Build GRCh38
rs933350644 Chr.17: 4557178-4557178 on Build GRCh38
rs528306357 Chr.17: 4557101-4557101 on Build GRCh38
rs74602457 Chr.17: 4556962-4556962 on Build GRCh38
rs368721164 Chr.17: 4557096-4557096 on Build GRCh38
rs750342526 Chr.17: 4556906-4556906 on Build GRCh38
rs1029616401 Chr.17: 4556895-4556895 on Build GRCh38
rs966554596 Chr.17: 4556868-4556868 on Build GRCh38
rs985474323 Chr.17: 4557082-4557082 on Build GRCh38
rs554938962 Chr.17: 4557042-4557042 on Build GRCh38
rs1010209499 Chr.17: 4556856-4556856 on Build GRCh38
rs959800693 Chr.17: 4557267-4557267 on Build GRCh38
rs901842522 Chr.17: 4557152-4557152 on Build GRCh38
rs569082889 Chr.17: 4556952-4556952 on Build GRCh38
rs928423341 Chr.17: 4556882-4556882 on Build GRCh38
rs545528349 Chr.17: 4557199-4557199 on Build GRCh38
rs574802863 Chr.17: 4557089-4557089 on Build GRCh38
rs777963434 Chr.17: 4557054-4557054 on Build GRCh38
rs979736742 Chr.17: 4556860-4556860 on Build GRCh38
rs969720780 Chr.17: 4556847-4556847 on Build GRCh38
rs893026895 Chr.17: 4556854-4556854 on Build GRCh38
rs972302316 Chr.17: 4557275-4557275 on Build GRCh38
rs997887811 Chr.17: 4557169-4557169 on Build GRCh38
rs1020221192 Chr.17: 4556864-4556864 on Build GRCh38
rs927000357 Chr.17: 4557059-4557059 on Build GRCh38
rs953560023 Chr.17: 4557218-4557218 on Build GRCh38
rs552077450 Chr.17: 4556936-4556936 on Build GRCh38
rs757335692 Chr.17: 4557157-4557157 on Build GRCh38
rs1052973835 Chr.17: 4557186-4557186 on Build GRCh38
rs937202066 Chr.17: 4557073-4557073 on Build GRCh38
rs891654072 Chr.17: 4557194-4557194 on Build GRCh38
rs188695802 Chr.17: 4557221-4557221 on Build GRCh38
rs766324992 Chr.17: 4556957-4556957 on Build GRCh38
rs762757660 Chr.17: 4556947-4556947 on Build GRCh38
rs764714480 Chr.17: 4556904-4556904 on Build GRCh38
rs949877875 Chr.17: 4557050-4557050 on Build GRCh38
rs754768848 Chr.17: 4557049-4557049 on Build GRCh38

View Full Product Details