Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3592018-3592511 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIPC3
Entrez Gene ID
126326
Gene Name
GIPC PDZ domain containing family member 3
Gene Aliases
C19orf64, DFNB15, DFNB72, DFNB95
UniGene
Hs.266873
Gene Chromosome Location
Chr.19: 3585531-3593541 on Build GRCh38
Transcripts
NM_133261.2 XM_005259492.3

SNP Detail

SNP ID Chromosome Location
rs959738000 Chr.19: 3592339-3592339 on Build GRCh38
rs983425170 Chr.19: 3592124-3592124 on Build GRCh38
rs562704971 Chr.19: 3592123-3592123 on Build GRCh38
rs16991953 Chr.19: 3592135-3592135 on Build GRCh38
rs968787121 Chr.19: 3592398-3592398 on Build GRCh38
rs558978587 Chr.19: 3592433-3592433 on Build GRCh38
rs942095971 Chr.19: 3592163-3592163 on Build GRCh38
rs946316270 Chr.19: 3592207-3592207 on Build GRCh38
rs570473894 Chr.19: 3592142-3592142 on Build GRCh38
rs746238944 Chr.19: 3592314-3592314 on Build GRCh38
rs1014054733 Chr.19: 3592321-3592321 on Build GRCh38
rs569031408 Chr.19: 3592487-3592487 on Build GRCh38
rs890685801 Chr.19: 3592206-3592206 on Build GRCh38
rs755816324 Chr.19: 3592187-3592187 on Build GRCh38
rs8105780 Chr.19: 3592173-3592173 on Build GRCh38
rs112193642 Chr.19: 3592354-3592354 on Build GRCh38
rs530431888 Chr.19: 3592120-3592120 on Build GRCh38
rs963449608 Chr.19: 3592384-3592384 on Build GRCh38
rs1000353076 Chr.19: 3592257-3592257 on Build GRCh38
rs758831103 Chr.19: 3592327-3592327 on Build GRCh38
rs934926891 Chr.19: 3592415-3592415 on Build GRCh38
rs1051857335 Chr.19: 3592435-3592435 on Build GRCh38
rs768575534 Chr.19: 3592442-3592442 on Build GRCh38
rs1024192748 Chr.19: 3592119-3592119 on Build GRCh38
rs879083887 Chr.19: 3592302-3592302 on Build GRCh38
rs991145952 Chr.19: 3592114-3592114 on Build GRCh38
rs912092796 Chr.19: 3592426-3592426 on Build GRCh38
rs1034139678 Chr.19: 3592271-3592271 on Build GRCh38
rs935644646 Chr.19: 3592472-3592472 on Build GRCh38
rs987572022 Chr.19: 3592419-3592419 on Build GRCh38
rs922090714 Chr.19: 3592191-3592191 on Build GRCh38
rs994884314 Chr.19: 3592283-3592283 on Build GRCh38
rs999978232 Chr.19: 3592085-3592085 on Build GRCh38
rs567549760 Chr.19: 3592180-3592180 on Build GRCh38
rs951247187 Chr.19: 3592332-3592332 on Build GRCh38
rs771267484 Chr.19: 3592255-3592255 on Build GRCh38
rs566303767 Chr.19: 3592411-3592411 on Build GRCh38
rs528215794 Chr.19: 3592147-3592147 on Build GRCh38
rs530054328 Chr.19: 3592249-3592249 on Build GRCh38
rs1024161840 Chr.19: 3592328-3592328 on Build GRCh38
rs894221487 Chr.19: 3592273-3592273 on Build GRCh38
rs73529965 Chr.19: 3592414-3592414 on Build GRCh38
rs925604592 Chr.19: 3592443-3592443 on Build GRCh38
rs1041353376 Chr.19: 3592229-3592229 on Build GRCh38
rs1034581715 Chr.19: 3592097-3592097 on Build GRCh38
rs1009820151 Chr.19: 3592048-3592048 on Build GRCh38
rs766413683 Chr.19: 3592060-3592060 on Build GRCh38
rs959807744 Chr.19: 3592110-3592110 on Build GRCh38
rs139969029 Chr.19: 3592317-3592317 on Build GRCh38
rs1015320883 Chr.19: 3592365-3592365 on Build GRCh38
rs779192934 Chr.19: 3592243-3592243 on Build GRCh38
rs958607225 Chr.19: 3592100-3592100 on Build GRCh38
rs904244593 Chr.19: 3592068-3592068 on Build GRCh38
rs973820385 Chr.19: 3592386-3592386 on Build GRCh38
rs932136094 Chr.19: 3592192-3592192 on Build GRCh38
rs113699849 Chr.19: 3592102-3592102 on Build GRCh38
rs1051882948 Chr.19: 3592193-3592193 on Build GRCh38
rs78989071 Chr.19: 3592047-3592047 on Build GRCh38
rs1004037767 Chr.19: 3592329-3592329 on Build GRCh38

View Full Product Details