Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 58708878-58709427 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLYAT
Entrez Gene ID
10249
Gene Name
glycine-N-acyltransferase
Gene Aliases
ACGNAT, CAT, GAT
UniGene
Hs.145384
Gene Chromosome Location
Chr.11: 58708757-58731974 on Build GRCh38
Transcripts
NM_201648.2 XM_017017087.1

SNP Detail

SNP ID Chromosome Location
rs974165252 Chr.11: 58709208-58709208 on Build GRCh38
rs977203959 Chr.11: 58709265-58709265 on Build GRCh38
rs949588887 Chr.11: 58709331-58709331 on Build GRCh38
rs539222730 Chr.11: 58708993-58708993 on Build GRCh38
rs557952770 Chr.11: 58709029-58709029 on Build GRCh38
rs1013727163 Chr.11: 58709062-58709062 on Build GRCh38
rs72514068 Chr.11: 58709375-58709376 on Build GRCh38
rs973816363 Chr.11: 58708973-58708973 on Build GRCh38
rs1000916051 Chr.11: 58708908-58708908 on Build GRCh38
rs924205106 Chr.11: 58709368-58709368 on Build GRCh38
rs999430129 Chr.11: 58709129-58709129 on Build GRCh38
rs996651923 Chr.11: 58708927-58708927 on Build GRCh38
rs114299324 Chr.11: 58709071-58709071 on Build GRCh38
rs5792123 Chr.11: 58709371-58709372 on Build GRCh38
rs201462888 Chr.11: 58709370-58709371 on Build GRCh38
rs546035815 Chr.11: 58709210-58709210 on Build GRCh38
rs34833934 Chr.11: 58709372-58709373 on Build GRCh38
rs118119913 Chr.11: 58709318-58709318 on Build GRCh38
rs887481230 Chr.11: 58708987-58708987 on Build GRCh38
rs77337461 Chr.11: 58708982-58708982 on Build GRCh38
rs930120990 Chr.11: 58709308-58709308 on Build GRCh38
rs1032847608 Chr.11: 58708918-58708918 on Build GRCh38
rs192675385 Chr.11: 58709390-58709390 on Build GRCh38
rs936960306 Chr.11: 58709387-58709387 on Build GRCh38
rs957170238 Chr.11: 58708920-58708920 on Build GRCh38
rs562412280 Chr.11: 58709143-58709143 on Build GRCh38
rs573006032 Chr.11: 58709030-58709030 on Build GRCh38
rs961338176 Chr.11: 58708965-58708965 on Build GRCh38
rs143895466 Chr.11: 58708911-58708911 on Build GRCh38
rs900575984 Chr.11: 58709073-58709073 on Build GRCh38
rs3833749 Chr.11: 58709376-58709377 on Build GRCh38
rs911186857 Chr.11: 58709235-58709235 on Build GRCh38
rs546479315 Chr.11: 58709096-58709096 on Build GRCh38
rs959099348 Chr.11: 58709211-58709211 on Build GRCh38
rs1032219670 Chr.11: 58709133-58709133 on Build GRCh38
rs955278111 Chr.11: 58708938-58708938 on Build GRCh38
rs949655564 Chr.11: 58709004-58709004 on Build GRCh38
rs546884390 Chr.11: 58709388-58709388 on Build GRCh38
rs564714566 Chr.11: 58709243-58709243 on Build GRCh38
rs894907851 Chr.11: 58709021-58709021 on Build GRCh38
rs756876305 Chr.11: 58708906-58708906 on Build GRCh38
rs1043033467 Chr.11: 58709007-58709007 on Build GRCh38
rs542374726 Chr.11: 58708935-58708935 on Build GRCh38
rs188047991 Chr.11: 58709093-58709093 on Build GRCh38
rs557011297 Chr.11: 58708970-58708970 on Build GRCh38
rs142476210 Chr.11: 58709403-58709403 on Build GRCh38
rs918728376 Chr.11: 58708958-58708958 on Build GRCh38
rs937560455 Chr.11: 58709360-58709360 on Build GRCh38
rs535500567 Chr.11: 58708955-58708955 on Build GRCh38
rs528699138 Chr.11: 58709359-58709359 on Build GRCh38
rs140899585 Chr.11: 58709072-58709072 on Build GRCh38
rs1013343688 Chr.11: 58709392-58709392 on Build GRCh38
rs1041232216 Chr.11: 58708914-58708914 on Build GRCh38
rs1054816744 Chr.11: 58709386-58709386 on Build GRCh38
rs76461290 Chr.11: 58709371-58709371 on Build GRCh38
rs544259856 Chr.11: 58709141-58709141 on Build GRCh38
rs1048700810 Chr.11: 58709326-58709326 on Build GRCh38
rs867631947 Chr.11: 58709377-58709377 on Build GRCh38
rs1028105615 Chr.11: 58709101-58709101 on Build GRCh38
rs866404912 Chr.11: 58709378-58709378 on Build GRCh38
rs938138970 Chr.11: 58708986-58708986 on Build GRCh38
rs1055311105 Chr.11: 58709019-58709019 on Build GRCh38
rs533072153 Chr.11: 58709154-58709154 on Build GRCh38
rs987155188 Chr.11: 58708948-58708948 on Build GRCh38
rs1008107561 Chr.11: 58709146-58709146 on Build GRCh38
rs961044451 Chr.11: 58709177-58709177 on Build GRCh38
rs918766993 Chr.11: 58709295-58709295 on Build GRCh38
rs902475733 Chr.11: 58709057-58709057 on Build GRCh38
rs1005872146 Chr.11: 58708998-58708998 on Build GRCh38
rs1055283198 Chr.11: 58708992-58708992 on Build GRCh38
rs749165444 Chr.11: 58709245-58709245 on Build GRCh38
rs568102282 Chr.11: 58708923-58708923 on Build GRCh38
rs918089667 Chr.11: 58709311-58709311 on Build GRCh38
rs756547316 Chr.11: 58709231-58709231 on Build GRCh38

View Full Product Details