Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 49511947-49512496 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLYATL3
Entrez Gene ID
389396
Gene Name
glycine-N-acyltransferase like 3
Gene Aliases
C6orf140, bA28H17.2
UniGene
Hs.585172
Gene Chromosome Location
Chr.6: 49499958-49528064 on Build GRCh38
Transcripts
NM_001010904.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3949302 c.78+4C>G p.? Chr.6: 49512072-49512072 on Build GRCh38
3949301 c.64C>A p.P22T Chr.6: 49512054-49512054 on Build GRCh38
7251689 c.75C>A p.L25L Chr.6: 49512065-49512065 on Build GRCh38
1137536 c.47T>C p.M16T Chr.6: 49512037-49512037 on Build GRCh38
3762114 c.72A>T p.S24S Chr.6: 49512062-49512062 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs964030427 Chr.6: 49512154-49512154 on Build GRCh38
rs1047182572 Chr.6: 49512203-49512203 on Build GRCh38
rs141201277 Chr.6: 49511981-49511981 on Build GRCh38
rs557296573 Chr.6: 49512190-49512190 on Build GRCh38
rs546566247 Chr.6: 49512462-49512462 on Build GRCh38
rs1057138792 Chr.6: 49512032-49512032 on Build GRCh38
rs546529523 Chr.6: 49512043-49512043 on Build GRCh38
rs36125184 Chr.6: 49512053-49512054 on Build GRCh38
rs756675540 Chr.6: 49512173-49512173 on Build GRCh38
rs6927790 Chr.6: 49512337-49512337 on Build GRCh38
rs941692748 Chr.6: 49512163-49512163 on Build GRCh38
rs760307636 Chr.6: 49512105-49512105 on Build GRCh38
rs759132418 Chr.6: 49512046-49512046 on Build GRCh38
rs534613072 Chr.6: 49512038-49512038 on Build GRCh38
rs896731675 Chr.6: 49512402-49512402 on Build GRCh38
rs1021703069 Chr.6: 49512034-49512034 on Build GRCh38
rs13193160 Chr.6: 49512108-49512108 on Build GRCh38
rs538484418 Chr.6: 49512085-49512085 on Build GRCh38
rs1005297040 Chr.6: 49512140-49512140 on Build GRCh38
rs1016711067 Chr.6: 49512461-49512461 on Build GRCh38
rs929570248 Chr.6: 49512413-49512413 on Build GRCh38
rs9395500 Chr.6: 49512217-49512217 on Build GRCh38
rs1056432672 Chr.6: 49512368-49512368 on Build GRCh38
rs559843093 Chr.6: 49512274-49512274 on Build GRCh38
rs7450503 Chr.6: 49512279-49512279 on Build GRCh38
rs946832275 Chr.6: 49512231-49512231 on Build GRCh38
rs903236130 Chr.6: 49512344-49512344 on Build GRCh38
rs201450544 Chr.6: 49512280-49512280 on Build GRCh38
rs913731208 Chr.6: 49512218-49512218 on Build GRCh38
rs1016637512 Chr.6: 49512153-49512153 on Build GRCh38
rs1043742293 Chr.6: 49512000-49512000 on Build GRCh38
rs7453050 Chr.6: 49512259-49512259 on Build GRCh38
rs979469863 Chr.6: 49512253-49512253 on Build GRCh38
rs1030211756 Chr.6: 49512405-49512405 on Build GRCh38
rs572838252 Chr.6: 49512246-49512246 on Build GRCh38
rs575261053 Chr.6: 49512186-49512186 on Build GRCh38
rs370134760 Chr.6: 49512266-49512266 on Build GRCh38
rs563875801 Chr.6: 49512333-49512333 on Build GRCh38
rs11358799 Chr.6: 49512267-49512267 on Build GRCh38
rs1480615 Chr.6: 49512062-49512062 on Build GRCh38
rs530909114 Chr.6: 49512378-49512378 on Build GRCh38
rs189790075 Chr.6: 49512327-49512327 on Build GRCh38
rs954749618 Chr.6: 49512090-49512090 on Build GRCh38
rs894494437 Chr.6: 49512257-49512257 on Build GRCh38
rs112642871 Chr.6: 49512453-49512453 on Build GRCh38
rs751124999 Chr.6: 49512414-49512414 on Build GRCh38
rs578181589 Chr.6: 49512288-49512288 on Build GRCh38
rs910245392 Chr.6: 49512150-49512150 on Build GRCh38
rs557104763 Chr.6: 49512087-49512087 on Build GRCh38
rs571486855 Chr.6: 49512472-49512472 on Build GRCh38
rs9381792 Chr.6: 49512449-49512449 on Build GRCh38
rs927436005 Chr.6: 49512266-49512266 on Build GRCh38
rs986063674 Chr.6: 49512121-49512121 on Build GRCh38
rs1030006135 Chr.6: 49512180-49512180 on Build GRCh38
rs938025335 Chr.6: 49512359-49512359 on Build GRCh38
rs369016092 Chr.6: 49512398-49512398 on Build GRCh38
rs571101787 Chr.6: 49512352-49512352 on Build GRCh38
rs542670390 Chr.6: 49512396-49512396 on Build GRCh38
rs560399915 Chr.6: 49512028-49512028 on Build GRCh38
rs879094725 Chr.6: 49512438-49512438 on Build GRCh38
rs905191475 Chr.6: 49512007-49512007 on Build GRCh38
rs930121179 Chr.6: 49512303-49512303 on Build GRCh38
rs1003574231 Chr.6: 49512215-49512215 on Build GRCh38
rs867744887 Chr.6: 49512458-49512458 on Build GRCh38
rs746650375 Chr.6: 49512436-49512436 on Build GRCh38
rs199836267 Chr.6: 49512284-49512284 on Build GRCh38
rs907467884 Chr.6: 49512205-49512205 on Build GRCh38
rs13193063 Chr.6: 49512037-49512037 on Build GRCh38

View Full Product Details