Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 72377421-72377971 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEXA-AS1
Entrez Gene ID
80072
Gene Name
HEXA antisense RNA 1
Gene Aliases
C15orf34
UniGene
Hs.591113
Gene Chromosome Location
Chr.15: 72376113-72378788 on Build GRCh38
Transcripts
NR_027262.1

SNP Detail

SNP ID Chromosome Location
rs569605344 Chr.15: 72377884-72377884 on Build GRCh38
rs748991841 Chr.15: 72377545-72377545 on Build GRCh38
rs577099815 Chr.15: 72377916-72377916 on Build GRCh38
rs528793209 Chr.15: 72377679-72377679 on Build GRCh38
rs1003726728 Chr.15: 72377737-72377737 on Build GRCh38
rs146129399 Chr.15: 72377574-72377574 on Build GRCh38
rs915468127 Chr.15: 72377571-72377571 on Build GRCh38
rs531135093 Chr.15: 72377546-72377546 on Build GRCh38
rs746030933 Chr.15: 72377894-72377895 on Build GRCh38
rs140143493 Chr.15: 72377843-72377843 on Build GRCh38
rs1035636926 Chr.15: 72377493-72377493 on Build GRCh38
rs1051994476 Chr.15: 72377944-72377944 on Build GRCh38
rs1040460555 Chr.15: 72377656-72377656 on Build GRCh38
rs989421734 Chr.15: 72377807-72377807 on Build GRCh38
rs534946676 Chr.15: 72377919-72377919 on Build GRCh38
rs759175550 Chr.15: 72377617-72377617 on Build GRCh38
rs771706097 Chr.15: 72377650-72377650 on Build GRCh38
rs944829179 Chr.15: 72377652-72377652 on Build GRCh38
rs558750963 Chr.15: 72377915-72377915 on Build GRCh38
rs4777507 Chr.15: 72377730-72377730 on Build GRCh38
rs988826019 Chr.15: 72377542-72377542 on Build GRCh38
rs1046857982 Chr.15: 72377926-72377926 on Build GRCh38
rs888323699 Chr.15: 72377688-72377688 on Build GRCh38
rs111279699 Chr.15: 72377870-72377870 on Build GRCh38
rs1034761142 Chr.15: 72377768-72377768 on Build GRCh38
rs4777506 Chr.15: 72377539-72377539 on Build GRCh38
rs760475189 Chr.15: 72377805-72377805 on Build GRCh38
rs1023337815 Chr.15: 72377722-72377722 on Build GRCh38
rs981849196 Chr.15: 72377448-72377448 on Build GRCh38
rs78192684 Chr.15: 72377832-72377832 on Build GRCh38
rs774125250 Chr.15: 72377593-72377593 on Build GRCh38
rs929814612 Chr.15: 72377677-72377677 on Build GRCh38
rs564452257 Chr.15: 72377595-72377595 on Build GRCh38
rs1005619770 Chr.15: 72377589-72377589 on Build GRCh38
rs996466663 Chr.15: 72377640-72377640 on Build GRCh38
rs1047438979 Chr.15: 72377682-72377682 on Build GRCh38
rs1005228190 Chr.15: 72377706-72377706 on Build GRCh38
rs532988489 Chr.15: 72377934-72377934 on Build GRCh38
rs901026790 Chr.15: 72377664-72377664 on Build GRCh38
rs796239251 Chr.15: 72377446-72377448 on Build GRCh38
rs556798828 Chr.15: 72377892-72377892 on Build GRCh38
rs1031475813 Chr.15: 72377790-72377790 on Build GRCh38
rs537274382 Chr.15: 72377913-72377913 on Build GRCh38
rs1046211206 Chr.15: 72377447-72377447 on Build GRCh38
rs988304641 Chr.15: 72377810-72377810 on Build GRCh38
rs986338386 Chr.15: 72377857-72377857 on Build GRCh38
rs899466941 Chr.15: 72377928-72377928 on Build GRCh38

View Full Product Details