Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 8893027-8893566 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERI1
Entrez Gene ID
90459
Gene Name
exoribonuclease 1
Gene Aliases
3'HEXO, HEXO, THEX1
UniGene
Hs.20000
Gene Chromosome Location
Chr.8: 8892225-9100262 on Build GRCh38
Transcripts
XM_017013949.1 NM_004225.2 XM_011543852.2

Gene Symbol
MFHAS1
Entrez Gene ID
9258
Gene Name
malignant fibrous histiocytoma amplified sequence 1
Gene Aliases
LRRC65, MASL1
UniGene
Hs.379414
Gene Chromosome Location
Chr.8: 8783354-8893621 on Build GRCh38
Transcripts
XM_017013949.1 NM_004225.2 XM_011543852.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6385950 c.14A>G p.D5G Chr.8: 8893045-8893045 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1005690707 Chr.8: 8893341-8893341 on Build GRCh38
rs772964514 Chr.8: 8893062-8893062 on Build GRCh38
rs764833947 Chr.8: 8893098-8893098 on Build GRCh38
rs3110411 Chr.8: 8893391-8893391 on Build GRCh38
rs1033553032 Chr.8: 8893432-8893432 on Build GRCh38
rs992094061 Chr.8: 8893439-8893439 on Build GRCh38
rs960906426 Chr.8: 8893438-8893438 on Build GRCh38
rs943149919 Chr.8: 8893534-8893534 on Build GRCh38
rs571965955 Chr.8: 8893296-8893296 on Build GRCh38
rs540970630 Chr.8: 8893305-8893305 on Build GRCh38
rs1030073191 Chr.8: 8893389-8893389 on Build GRCh38
rs988263111 Chr.8: 8893262-8893262 on Build GRCh38
rs918970293 Chr.8: 8893310-8893310 on Build GRCh38
rs1048807370 Chr.8: 8893330-8893330 on Build GRCh38
rs926515749 Chr.8: 8893295-8893295 on Build GRCh38
rs576766983 Chr.8: 8893066-8893066 on Build GRCh38
rs772196002 Chr.8: 8893050-8893050 on Build GRCh38
rs918892015 Chr.8: 8893504-8893504 on Build GRCh38
rs1018239004 Chr.8: 8893105-8893105 on Build GRCh38
rs1025798370 Chr.8: 8893091-8893091 on Build GRCh38
rs915038467 Chr.8: 8893267-8893267 on Build GRCh38
rs539518189 Chr.8: 8893224-8893224 on Build GRCh38
rs532441026 Chr.8: 8893396-8893396 on Build GRCh38
rs998480644 Chr.8: 8893375-8893375 on Build GRCh38
rs984479743 Chr.8: 8893519-8893519 on Build GRCh38
rs908945966 Chr.8: 8893528-8893528 on Build GRCh38
rs1045006738 Chr.8: 8893173-8893173 on Build GRCh38
rs956834641 Chr.8: 8893117-8893117 on Build GRCh38
rs976832653 Chr.8: 8893111-8893112 on Build GRCh38
rs939033053 Chr.8: 8893183-8893183 on Build GRCh38
rs900068458 Chr.8: 8893373-8893373 on Build GRCh38
rs563000367 Chr.8: 8893542-8893542 on Build GRCh38
rs561559305 Chr.8: 8893285-8893285 on Build GRCh38
rs987958667 Chr.8: 8893130-8893130 on Build GRCh38
rs922695667 Chr.8: 8893115-8893115 on Build GRCh38
rs879781814 Chr.8: 8893482-8893482 on Build GRCh38
rs964052665 Chr.8: 8893227-8893227 on Build GRCh38
rs981001572 Chr.8: 8893431-8893431 on Build GRCh38
rs897557291 Chr.8: 8893199-8893199 on Build GRCh38
rs928990292 Chr.8: 8893517-8893517 on Build GRCh38
rs760674079 Chr.8: 8893064-8893064 on Build GRCh38
rs753682212 Chr.8: 8893071-8893071 on Build GRCh38
rs915110650 Chr.8: 8893143-8893143 on Build GRCh38
rs993243041 Chr.8: 8893200-8893200 on Build GRCh38
rs994278400 Chr.8: 8893076-8893076 on Build GRCh38
rs759349620 Chr.8: 8893094-8893094 on Build GRCh38
rs1029691146 Chr.8: 8893248-8893248 on Build GRCh38
rs552623930 Chr.8: 8893421-8893421 on Build GRCh38
rs1056157913 Chr.8: 8893197-8893197 on Build GRCh38
rs888619282 Chr.8: 8893204-8893204 on Build GRCh38
rs929021314 Chr.8: 8893316-8893316 on Build GRCh38
rs1039894840 Chr.8: 8893355-8893355 on Build GRCh38
rs575809107 Chr.8: 8893238-8893238 on Build GRCh38
rs888583600 Chr.8: 8893338-8893338 on Build GRCh38
rs968148623 Chr.8: 8893426-8893426 on Build GRCh38
rs1022407135 Chr.8: 8893424-8893424 on Build GRCh38
rs926280049 Chr.8: 8893174-8893174 on Build GRCh38
rs1047219155 Chr.8: 8893203-8893203 on Build GRCh38
rs1005727991 Chr.8: 8893206-8893206 on Build GRCh38
rs766132366 Chr.8: 8893068-8893068 on Build GRCh38
rs964396654 Chr.8: 8893108-8893108 on Build GRCh38
rs967887367 Chr.8: 8893286-8893286 on Build GRCh38
rs752402309 Chr.8: 8893110-8893110 on Build GRCh38
rs541594536 Chr.8: 8893258-8893258 on Build GRCh38
rs556027176 Chr.8: 8893232-8893232 on Build GRCh38
rs1018952926 Chr.8: 8893216-8893216 on Build GRCh38
rs1005800104 Chr.8: 8893099-8893099 on Build GRCh38
rs980661617 Chr.8: 8893289-8893289 on Build GRCh38

View Full Product Details