Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 205601534-205602083 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELK4
Entrez Gene ID
2005
Gene Name
ELK4, ETS transcription factor
Gene Aliases
SAP1
UniGene
Hs.497520
Gene Chromosome Location
Chr.1: 205597774-205634077 on Build GRCh38
Transcripts
NM_181644.4 XM_011509229.2

Gene Symbol
MFSD4A
Entrez Gene ID
148808
Gene Name
major facilitator superfamily domain containing 4A
Gene Aliases
MFSD4, UNQ3064
UniGene
Hs.567714
Gene Chromosome Location
Chr.1: 205568984-205602918 on Build GRCh38
Transcripts
NM_181644.4 XM_011509229.2

SNP Detail

SNP ID Chromosome Location
rs934569374 Chr.1: 205601981-205601981 on Build GRCh38
rs148519076 Chr.1: 205601983-205601983 on Build GRCh38
rs985022814 Chr.1: 205601591-205601591 on Build GRCh38
rs770494316 Chr.1: 205601937-205601941 on Build GRCh38
rs1048852382 Chr.1: 205601578-205601578 on Build GRCh38
rs951914407 Chr.1: 205601917-205601920 on Build GRCh38
rs539673707 Chr.1: 205601573-205601573 on Build GRCh38
rs560306430 Chr.1: 205601794-205601794 on Build GRCh38
rs769479993 Chr.1: 205601855-205601855 on Build GRCh38
rs549529834 Chr.1: 205601900-205601900 on Build GRCh38
rs1042955416 Chr.1: 205601886-205601886 on Build GRCh38
rs903030721 Chr.1: 205601678-205601678 on Build GRCh38
rs750946293 Chr.1: 205601630-205601630 on Build GRCh38
rs1014104800 Chr.1: 205602051-205602051 on Build GRCh38
rs146871671 Chr.1: 205601601-205601601 on Build GRCh38
rs910401948 Chr.1: 205601954-205601954 on Build GRCh38
rs1052696326 Chr.1: 205601984-205601984 on Build GRCh38
rs947326253 Chr.1: 205601653-205601653 on Build GRCh38
rs544216318 Chr.1: 205601682-205601682 on Build GRCh38
rs971811658 Chr.1: 205601624-205601624 on Build GRCh38
rs146232060 Chr.1: 205601669-205601669 on Build GRCh38
rs559368314 Chr.1: 205601774-205601774 on Build GRCh38
rs888826192 Chr.1: 205601579-205601580 on Build GRCh38
rs943142531 Chr.1: 205601758-205601758 on Build GRCh38
rs1040416302 Chr.1: 205601582-205601585 on Build GRCh38
rs531846758 Chr.1: 205601968-205601968 on Build GRCh38
rs910225720 Chr.1: 205601862-205601862 on Build GRCh38
rs919144388 Chr.1: 205601716-205601716 on Build GRCh38
rs1024079392 Chr.1: 205601773-205601773 on Build GRCh38
rs923053590 Chr.1: 205601625-205601625 on Build GRCh38
rs12041432 Chr.1: 205601642-205601642 on Build GRCh38
rs533693175 Chr.1: 205602023-205602023 on Build GRCh38
rs1035773552 Chr.1: 205601884-205601884 on Build GRCh38
rs575355362 Chr.1: 205601681-205601681 on Build GRCh38
rs985875866 Chr.1: 205601856-205601856 on Build GRCh38
rs747696242 Chr.1: 205601928-205601928 on Build GRCh38
rs529494401 Chr.1: 205601896-205601896 on Build GRCh38
rs961380125 Chr.1: 205601893-205601893 on Build GRCh38
rs947653832 Chr.1: 205602036-205602036 on Build GRCh38
rs961349072 Chr.1: 205601688-205601688 on Build GRCh38
rs541967570 Chr.1: 205601829-205601829 on Build GRCh38
rs1007244858 Chr.1: 205601580-205601585 on Build GRCh38
rs895632169 Chr.1: 205602025-205602025 on Build GRCh38
rs1049345895 Chr.1: 205601735-205601735 on Build GRCh38
rs1050545554 Chr.1: 205601635-205601635 on Build GRCh38
rs578069219 Chr.1: 205601802-205601802 on Build GRCh38
rs909397767 Chr.1: 205601598-205601598 on Build GRCh38
rs7555438 Chr.1: 205601831-205601831 on Build GRCh38
rs1035524108 Chr.1: 205601693-205601693 on Build GRCh38
rs191341055 Chr.1: 205602056-205602056 on Build GRCh38
rs901497370 Chr.1: 205601585-205601585 on Build GRCh38
rs970072498 Chr.1: 205601589-205601589 on Build GRCh38
rs947018532 Chr.1: 205601863-205601863 on Build GRCh38
rs972691315 Chr.1: 205601902-205601902 on Build GRCh38
rs933091138 Chr.1: 205601849-205601849 on Build GRCh38
rs140087789 Chr.1: 205601728-205601728 on Build GRCh38
rs113980058 Chr.1: 205601903-205601903 on Build GRCh38
rs998604697 Chr.1: 205601686-205601686 on Build GRCh38
rs1023690650 Chr.1: 205602053-205602053 on Build GRCh38
rs1013657669 Chr.1: 205601729-205601729 on Build GRCh38
rs535552031 Chr.1: 205601644-205601644 on Build GRCh38
rs972010005 Chr.1: 205601710-205601710 on Build GRCh38
rs747793307 Chr.1: 205601599-205601604 on Build GRCh38
rs12408345 Chr.1: 205601835-205601835 on Build GRCh38
rs370399603 Chr.1: 205601740-205601740 on Build GRCh38
rs943215676 Chr.1: 205601955-205601955 on Build GRCh38

View Full Product Details