Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 56476801-56477322 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BBS2
Entrez Gene ID
583
Gene Name
Bardet-Biedl syndrome 2
Gene Aliases
BBS, RP74
UniGene
Hs.333738
Gene Chromosome Location
Chr.16: 56470403-56520096 on Build GRCh38
Transcripts
XM_005256080.2 NM_001324357.1 NM_001324358.1 NM_001324359.1 NM_001324360.1 NM_001324361.1 NM_001324362.1 NM_001324363.1 NM_018233.3

Gene Symbol
OGFOD1
Entrez Gene ID
55239
Gene Name
2-oxoglutarate and iron dependent oxygenase domain containing 1
Gene Aliases
TPA1
UniGene
Hs.231883
Gene Chromosome Location
Chr.16: 56451480-56477495 on Build GRCh38
Transcripts
XM_005256080.2 NM_001324357.1 NM_001324358.1 NM_001324359.1 NM_001324360.1 NM_001324361.1 NM_001324362.1 NM_001324363.1 NM_018233.3

SNP Detail

SNP ID Chromosome Location
rs9939035 Chr.16: 56476933-56476933 on Build GRCh38
rs757428512 Chr.16: 56477174-56477174 on Build GRCh38
rs35091365 Chr.16: 56476947-56476948 on Build GRCh38
rs1001246685 Chr.16: 56477142-56477142 on Build GRCh38
rs147387538 Chr.16: 56476858-56476858 on Build GRCh38
rs369051216 Chr.16: 56476854-56476854 on Build GRCh38
rs964217444 Chr.16: 56477216-56477216 on Build GRCh38
rs545122632 Chr.16: 56476944-56476944 on Build GRCh38
rs555656224 Chr.16: 56477130-56477130 on Build GRCh38
rs1039463166 Chr.16: 56476990-56476990 on Build GRCh38
rs979690110 Chr.16: 56476917-56476917 on Build GRCh38
rs553938377 Chr.16: 56476973-56476973 on Build GRCh38
rs761503296 Chr.16: 56476846-56476846 on Build GRCh38
rs112162270 Chr.16: 56477114-56477114 on Build GRCh38
rs1023635433 Chr.16: 56476904-56476904 on Build GRCh38
rs995426605 Chr.16: 56476840-56476840 on Build GRCh38
rs538926822 Chr.16: 56476866-56476866 on Build GRCh38
rs530010352 Chr.16: 56477257-56477257 on Build GRCh38
rs538001219 Chr.16: 56476921-56476921 on Build GRCh38
rs193063311 Chr.16: 56477258-56477258 on Build GRCh38
rs112571594 Chr.16: 56477077-56477077 on Build GRCh38
rs1012335184 Chr.16: 56477122-56477122 on Build GRCh38
rs567473261 Chr.16: 56476905-56476905 on Build GRCh38
rs572259744 Chr.16: 56477154-56477154 on Build GRCh38
rs528535963 Chr.16: 56477140-56477142 on Build GRCh38
rs898163223 Chr.16: 56476826-56476826 on Build GRCh38
rs567514056 Chr.16: 56477268-56477268 on Build GRCh38
rs190285592 Chr.16: 56476853-56476853 on Build GRCh38
rs960744311 Chr.16: 56476869-56476869 on Build GRCh38
rs9938494 Chr.16: 56477033-56477033 on Build GRCh38
rs1012348052 Chr.16: 56476884-56476884 on Build GRCh38
rs370174161 Chr.16: 56476981-56476981 on Build GRCh38
rs532766730 Chr.16: 56477139-56477139 on Build GRCh38
rs867492792 Chr.16: 56477061-56477061 on Build GRCh38
rs919697784 Chr.16: 56477262-56477262 on Build GRCh38
rs561518043 Chr.16: 56477071-56477071 on Build GRCh38
rs779043392 Chr.16: 56477219-56477219 on Build GRCh38
rs796316650 Chr.16: 56477129-56477130 on Build GRCh38
rs111625702 Chr.16: 56476930-56476930 on Build GRCh38
rs1035542960 Chr.16: 56476864-56476864 on Build GRCh38
rs10438565 Chr.16: 56477154-56477154 on Build GRCh38
rs1056954582 Chr.16: 56477099-56477099 on Build GRCh38
rs559437375 Chr.16: 56477193-56477193 on Build GRCh38
rs182487482 Chr.16: 56476868-56476868 on Build GRCh38
rs376333831 Chr.16: 56477141-56477141 on Build GRCh38
rs542665657 Chr.16: 56477046-56477046 on Build GRCh38
rs764822838 Chr.16: 56476859-56476859 on Build GRCh38
rs956912997 Chr.16: 56477166-56477166 on Build GRCh38
rs919597632 Chr.16: 56477020-56477020 on Build GRCh38

View Full Product Details