Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 68271348-68271804 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARSG
Entrez Gene ID
22901
Gene Name
arylsulfatase G
Gene Aliases
-
UniGene
Hs.437249
Gene Chromosome Location
Chr.17: 68259089-68452000 on Build GRCh38
Transcripts
NM_014960.4 XM_005257170.3 XM_005257172.3 XM_006721777.3 XM_006721779.3 XM_011524535.2 XM_011524538.2 XM_011524540.2 XM_011524541.2 XM_011524542.2 XM_011524543.2 XM_011524544.2 XM_011524545.2 XM_017024361.1 XM_017024366.1 NM_001174166.1 NM_004694.4 XM_005257789.3 XM_011525461.2 XM_017025291.1 XM_017025292.1

Gene Symbol
SLC16A6
Entrez Gene ID
9120
Gene Name
solute carrier family 16 member 6
Gene Aliases
MCT6, MCT7
UniGene
Hs.42645
Gene Chromosome Location
Chr.17: 68267026-68291264 on Build GRCh38
Transcripts
NM_014960.4 XM_005257170.3 XM_005257172.3 XM_006721777.3 XM_006721779.3 XM_011524535.2 XM_011524538.2 XM_011524540.2 XM_011524541.2 XM_011524542.2 XM_011524543.2 XM_011524544.2 XM_011524545.2 XM_017024361.1 XM_017024366.1 NM_001174166.1 NM_004694.4 XM_005257789.3 XM_011525461.2 XM_017025291.1 XM_017025292.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5418235 c.563T>C p.L188P Chr.17: 68271597-68271597 on Build GRCh38
1303246 c.519G>A p.L173L Chr.17: 68271641-68271641 on Build GRCh38
3717559 c.692C>T p.T231I Chr.17: 68271468-68271468 on Build GRCh38
6779848 c.632C>T p.P211L Chr.17: 68271528-68271528 on Build GRCh38
1265709 c.538A>G p.R180G Chr.17: 68271622-68271622 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs7222013 Chr.17: 68271550-68271550 on Build GRCh38
rs9910901 Chr.17: 68271695-68271695 on Build GRCh38
rs193189838 Chr.17: 68271378-68271378 on Build GRCh38
rs377560800 Chr.17: 68271504-68271504 on Build GRCh38
rs782358914 Chr.17: 68271372-68271372 on Build GRCh38
rs781907337 Chr.17: 68271528-68271528 on Build GRCh38
rs374993622 Chr.17: 68271572-68271572 on Build GRCh38
rs782713539 Chr.17: 68271392-68271392 on Build GRCh38
rs782684229 Chr.17: 68271609-68271609 on Build GRCh38
rs139961659 Chr.17: 68271631-68271631 on Build GRCh38
rs782416591 Chr.17: 68271368-68271368 on Build GRCh38
rs782238996 Chr.17: 68271670-68271670 on Build GRCh38
rs542108620 Chr.17: 68271577-68271577 on Build GRCh38
rs782735791 Chr.17: 68271398-68271398 on Build GRCh38
rs782742904 Chr.17: 68271628-68271628 on Build GRCh38
rs782508730 Chr.17: 68271447-68271447 on Build GRCh38
rs781795998 Chr.17: 68271395-68271395 on Build GRCh38
rs569854171 Chr.17: 68271370-68271370 on Build GRCh38
rs782479737 Chr.17: 68271633-68271633 on Build GRCh38
rs782401318 Chr.17: 68271523-68271523 on Build GRCh38
rs782357159 Chr.17: 68271427-68271427 on Build GRCh38
rs781847076 Chr.17: 68271548-68271548 on Build GRCh38
rs782713311 Chr.17: 68271571-68271571 on Build GRCh38
rs200168065 Chr.17: 68271390-68271390 on Build GRCh38
rs7216446 Chr.17: 68271707-68271707 on Build GRCh38
rs781838059 Chr.17: 68271539-68271539 on Build GRCh38
rs781980063 Chr.17: 68271495-68271495 on Build GRCh38
rs782478993 Chr.17: 68271661-68271661 on Build GRCh38
rs141718604 Chr.17: 68271605-68271605 on Build GRCh38
rs782764638 Chr.17: 68271527-68271527 on Build GRCh38
rs782098825 Chr.17: 68271622-68271622 on Build GRCh38
rs782655449 Chr.17: 68271544-68271544 on Build GRCh38
rs201650215 Chr.17: 68271389-68271389 on Build GRCh38
rs781883104 Chr.17: 68271663-68271663 on Build GRCh38
rs367677855 Chr.17: 68271579-68271579 on Build GRCh38
rs782275679 Chr.17: 68271462-68271462 on Build GRCh38
rs782663213 Chr.17: 68271667-68271667 on Build GRCh38
rs372842877 Chr.17: 68271596-68271596 on Build GRCh38
rs3744307 Chr.17: 68271509-68271509 on Build GRCh38
rs782660310 Chr.17: 68271567-68271567 on Build GRCh38
rs782661111 Chr.17: 68271674-68271674 on Build GRCh38
rs781954164 Chr.17: 68271694-68271694 on Build GRCh38
rs190024933 Chr.17: 68271681-68271681 on Build GRCh38
rs782134895 Chr.17: 68271518-68271518 on Build GRCh38
rs781849963 Chr.17: 68271636-68271636 on Build GRCh38
rs782774786 Chr.17: 68271430-68271431 on Build GRCh38
rs782759567 Chr.17: 68271388-68271388 on Build GRCh38
rs781954926 Chr.17: 68271515-68271515 on Build GRCh38
rs781865429 Chr.17: 68271630-68271630 on Build GRCh38
rs373572305 Chr.17: 68271494-68271494 on Build GRCh38
rs368721903 Chr.17: 68271634-68271634 on Build GRCh38
rs144470811 Chr.17: 68271583-68271583 on Build GRCh38
rs138318896 Chr.17: 68271367-68271367 on Build GRCh38
rs782022906 Chr.17: 68271533-68271533 on Build GRCh38
rs782436259 Chr.17: 68271673-68271673 on Build GRCh38
rs782484654 Chr.17: 68271397-68271397 on Build GRCh38
rs781947096 Chr.17: 68271377-68271377 on Build GRCh38
rs782292749 Chr.17: 68271742-68271742 on Build GRCh38
rs370502404 Chr.17: 68271513-68271513 on Build GRCh38
rs61745585 Chr.17: 68271465-68271465 on Build GRCh38
rs772864281 Chr.17: 68271687-68271687 on Build GRCh38
rs145765322 Chr.17: 68271505-68271505 on Build GRCh38
rs142658044 Chr.17: 68271371-68271371 on Build GRCh38
rs782682614 Chr.17: 68271461-68271461 on Build GRCh38
rs782448073 Chr.17: 68271466-68271466 on Build GRCh38
rs4410141 Chr.17: 68271498-68271498 on Build GRCh38
rs62087130 Chr.17: 68271611-68271611 on Build GRCh38
rs782314366 Chr.17: 68271692-68271692 on Build GRCh38
rs782214885 Chr.17: 68271582-68271582 on Build GRCh38
rs782206782 Chr.17: 68271483-68271483 on Build GRCh38
rs61740584 Chr.17: 68271593-68271593 on Build GRCh38
rs782463305 Chr.17: 68271541-68271541 on Build GRCh38
rs192279074 Chr.17: 68271766-68271766 on Build GRCh38

View Full Product Details