Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 89575808-89576316 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TICRR
Entrez Gene ID
90381
Gene Name
TOPBP1 interacting checkpoint and replication regulator
Gene Aliases
C15orf42, SLD3, Treslin
UniGene
Hs.441708
Gene Chromosome Location
Chr.15: 89575587-89628023 on Build GRCh38
Transcripts
NM_001308025.1 NM_152259.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3706806 c.303C>A p.H101Q Chr.15: 89575889-89575889 on Build GRCh38
2015461 c.643G>A p.E215K Chr.15: 89576229-89576229 on Build GRCh38
3706808 c.572A>T p.K191M Chr.15: 89576158-89576158 on Build GRCh38
6505687 c.461T>A p.V154D Chr.15: 89576047-89576047 on Build GRCh38
3706807 c.370C>A p.P124T Chr.15: 89575956-89575956 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747380886 Chr.15: 89576270-89576270 on Build GRCh38
rs201861926 Chr.15: 89576148-89576148 on Build GRCh38
rs756871034 Chr.15: 89576026-89576026 on Build GRCh38
rs776628138 Chr.15: 89576193-89576193 on Build GRCh38
rs957980179 Chr.15: 89575865-89575865 on Build GRCh38
rs767691562 Chr.15: 89575899-89575899 on Build GRCh38
rs371496400 Chr.15: 89576111-89576111 on Build GRCh38
rs750164208 Chr.15: 89576140-89576140 on Build GRCh38
rs763446614 Chr.15: 89575872-89575872 on Build GRCh38
rs371327246 Chr.15: 89575854-89575854 on Build GRCh38
rs745930616 Chr.15: 89576278-89576278 on Build GRCh38
rs200087603 Chr.15: 89575830-89575830 on Build GRCh38
rs770335617 Chr.15: 89575949-89575949 on Build GRCh38
rs374963310 Chr.15: 89576045-89576045 on Build GRCh38
rs765031238 Chr.15: 89575964-89575964 on Build GRCh38
rs574767682 Chr.15: 89576003-89576003 on Build GRCh38
rs753401897 Chr.15: 89576156-89576156 on Build GRCh38
rs760335639 Chr.15: 89576228-89576228 on Build GRCh38
rs1009236317 Chr.15: 89576138-89576138 on Build GRCh38
rs780486490 Chr.15: 89576058-89576059 on Build GRCh38
rs759646216 Chr.15: 89576202-89576202 on Build GRCh38
rs763945849 Chr.15: 89576132-89576132 on Build GRCh38
rs150476518 Chr.15: 89576189-89576189 on Build GRCh38
rs753348868 Chr.15: 89576249-89576249 on Build GRCh38
rs760151150 Chr.15: 89575858-89575858 on Build GRCh38
rs749865830 Chr.15: 89576032-89576032 on Build GRCh38
rs750254245 Chr.15: 89576227-89576227 on Build GRCh38
rs767269348 Chr.15: 89576222-89576222 on Build GRCh38
rs748491726 Chr.15: 89576048-89576048 on Build GRCh38
rs773355437 Chr.15: 89576060-89576060 on Build GRCh38
rs766159194 Chr.15: 89576243-89576243 on Build GRCh38
rs774329342 Chr.15: 89576280-89576280 on Build GRCh38
rs959595732 Chr.15: 89575959-89575959 on Build GRCh38
rs766270036 Chr.15: 89576212-89576212 on Build GRCh38
rs774208070 Chr.15: 89576215-89576215 on Build GRCh38
rs772288001 Chr.15: 89576054-89576054 on Build GRCh38
rs983558235 Chr.15: 89575987-89575987 on Build GRCh38
rs747291023 Chr.15: 89576187-89576187 on Build GRCh38
rs766513294 Chr.15: 89576109-89576109 on Build GRCh38
rs1033961947 Chr.15: 89575954-89575954 on Build GRCh38
rs1038593726 Chr.15: 89576018-89576018 on Build GRCh38
rs777374498 Chr.15: 89575940-89575940 on Build GRCh38
rs765137741 Chr.15: 89576008-89576008 on Build GRCh38
rs779309777 Chr.15: 89576158-89576158 on Build GRCh38
rs960234698 Chr.15: 89575980-89575980 on Build GRCh38
rs901013150 Chr.15: 89575852-89575852 on Build GRCh38
rs558205500 Chr.15: 89575864-89575864 on Build GRCh38
rs967411239 Chr.15: 89575921-89575921 on Build GRCh38
rs747292046 Chr.15: 89576129-89576131 on Build GRCh38
rs751867337 Chr.15: 89575885-89575885 on Build GRCh38
rs764385206 Chr.15: 89575875-89575875 on Build GRCh38
rs199518473 Chr.15: 89576126-89576126 on Build GRCh38
rs768593431 Chr.15: 89576207-89576207 on Build GRCh38
rs750280303 Chr.15: 89576025-89576025 on Build GRCh38
rs939730432 Chr.15: 89576058-89576058 on Build GRCh38
rs760714954 Chr.15: 89576097-89576097 on Build GRCh38
rs749634970 Chr.15: 89575981-89575981 on Build GRCh38
rs758213007 Chr.15: 89576135-89576135 on Build GRCh38
rs556549165 Chr.15: 89575838-89575838 on Build GRCh38
rs78756713 Chr.15: 89576292-89576292 on Build GRCh38
rs773620037 Chr.15: 89576024-89576024 on Build GRCh38
rs767162484 Chr.15: 89576137-89576137 on Build GRCh38
rs866021379 Chr.15: 89575861-89575861 on Build GRCh38
rs200929837 Chr.15: 89576171-89576171 on Build GRCh38
rs200336344 Chr.15: 89576170-89576170 on Build GRCh38
rs916100501 Chr.15: 89576285-89576285 on Build GRCh38
rs773301628 Chr.15: 89576006-89576006 on Build GRCh38
rs917929201 Chr.15: 89575973-89575973 on Build GRCh38
rs769031093 Chr.15: 89576235-89576236 on Build GRCh38
rs772126308 Chr.15: 89575911-89575912 on Build GRCh38
rs1041247402 Chr.15: 89575845-89575845 on Build GRCh38
rs745337713 Chr.15: 89576028-89576028 on Build GRCh38
rs374324640 Chr.15: 89576120-89576120 on Build GRCh38
rs780743454 Chr.15: 89576027-89576027 on Build GRCh38
rs761594931 Chr.15: 89576289-89576289 on Build GRCh38
rs771103862 Chr.15: 89576274-89576274 on Build GRCh38
rs776184882 Chr.15: 89575974-89575974 on Build GRCh38
rs780698335 Chr.15: 89576031-89576031 on Build GRCh38
rs200014273 Chr.15: 89575889-89575889 on Build GRCh38
rs967409043 Chr.15: 89576195-89576195 on Build GRCh38
rs562381739 Chr.15: 89576012-89576012 on Build GRCh38
rs754495438 Chr.15: 89576253-89576253 on Build GRCh38
rs761659905 Chr.15: 89576217-89576217 on Build GRCh38
rs968286940 Chr.15: 89575941-89575941 on Build GRCh38
rs778853022 Chr.15: 89575923-89575923 on Build GRCh38
rs779444581 Chr.15: 89576258-89576258 on Build GRCh38
rs983843982 Chr.15: 89576262-89576262 on Build GRCh38
rs780697286 Chr.15: 89575957-89575957 on Build GRCh38
rs762035857 Chr.15: 89575997-89575997 on Build GRCh38
rs886840128 Chr.15: 89575832-89575832 on Build GRCh38
rs754876698 Chr.15: 89575922-89575922 on Build GRCh38
rs769207727 Chr.15: 89575985-89575985 on Build GRCh38
rs769297665 Chr.15: 89576043-89576043 on Build GRCh38
rs750481691 Chr.15: 89575913-89575913 on Build GRCh38
rs370178732 Chr.15: 89575988-89575988 on Build GRCh38
rs777943438 Chr.15: 89576265-89576265 on Build GRCh38
rs963814310 Chr.15: 89576279-89576279 on Build GRCh38
rs767636964 Chr.15: 89576000-89576000 on Build GRCh38
rs79284973 Chr.15: 89576264-89576264 on Build GRCh38
rs746745230 Chr.15: 89575943-89575943 on Build GRCh38
rs753211177 Chr.15: 89576261-89576261 on Build GRCh38
rs752672402 Chr.15: 89576011-89576011 on Build GRCh38
rs755500392 Chr.15: 89576157-89576157 on Build GRCh38
rs775935071 Chr.15: 89575867-89575867 on Build GRCh38
rs1028238719 Chr.15: 89575862-89575862 on Build GRCh38
rs751333018 Chr.15: 89576133-89576133 on Build GRCh38
rs779651052 Chr.15: 89576155-89576155 on Build GRCh38
rs571728915 Chr.15: 89576275-89576275 on Build GRCh38
rs748523531 Chr.15: 89576165-89576165 on Build GRCh38
rs752439482 Chr.15: 89575934-89575934 on Build GRCh38
rs770387851 Chr.15: 89575970-89575970 on Build GRCh38
rs932569999 Chr.15: 89576093-89576093 on Build GRCh38

View Full Product Details