Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 93186745-93187251 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM251
Entrez Gene ID
26175
Gene Name
transmembrane protein 251
Gene Aliases
C14orf109
UniGene
Hs.275352
Gene Chromosome Location
Chr.14: 93184951-93187086 on Build GRCh38
Transcripts
NM_001098621.1 NM_015676.1

SNP Detail

SNP ID Chromosome Location
rs1020891442 Chr.14: 93187026-93187026 on Build GRCh38
rs758183928 Chr.14: 93186914-93186914 on Build GRCh38
rs997899262 Chr.14: 93186955-93186955 on Build GRCh38
rs990570622 Chr.14: 93187095-93187095 on Build GRCh38
rs916173825 Chr.14: 93187157-93187157 on Build GRCh38
rs899731711 Chr.14: 93186967-93186967 on Build GRCh38
rs959188492 Chr.14: 93187082-93187082 on Build GRCh38
rs907287231 Chr.14: 93186886-93186886 on Build GRCh38
rs981725194 Chr.14: 93186770-93186770 on Build GRCh38
rs982924425 Chr.14: 93187030-93187030 on Build GRCh38
rs1022186935 Chr.14: 93186960-93186960 on Build GRCh38
rs541698597 Chr.14: 93187128-93187128 on Build GRCh38
rs948910579 Chr.14: 93187168-93187168 on Build GRCh38
rs369313359 Chr.14: 93186790-93186790 on Build GRCh38
rs554630763 Chr.14: 93187006-93187007 on Build GRCh38
rs200715503 Chr.14: 93186917-93186917 on Build GRCh38
rs928617743 Chr.14: 93187198-93187198 on Build GRCh38
rs972785784 Chr.14: 93187214-93187214 on Build GRCh38
rs970431675 Chr.14: 93187180-93187180 on Build GRCh38
rs916139706 Chr.14: 93186826-93186826 on Build GRCh38
rs1029706102 Chr.14: 93187024-93187024 on Build GRCh38
rs116029458 Chr.14: 93186819-93186819 on Build GRCh38
rs556201455 Chr.14: 93187097-93187097 on Build GRCh38
rs556605980 Chr.14: 93187036-93187036 on Build GRCh38
rs372910344 Chr.14: 93187126-93187126 on Build GRCh38
rs955191410 Chr.14: 93187028-93187028 on Build GRCh38
rs752334828 Chr.14: 93186830-93186830 on Build GRCh38
rs777618225 Chr.14: 93187017-93187017 on Build GRCh38
rs147298582 Chr.14: 93187007-93187007 on Build GRCh38
rs1038612071 Chr.14: 93186811-93186811 on Build GRCh38
rs74984385 Chr.14: 93187059-93187059 on Build GRCh38
rs962888139 Chr.14: 93187064-93187064 on Build GRCh38
rs892282558 Chr.14: 93186940-93186940 on Build GRCh38
rs948710831 Chr.14: 93186831-93186831 on Build GRCh38
rs764326014 Chr.14: 93186789-93186790 on Build GRCh38
rs908703089 Chr.14: 93186794-93186794 on Build GRCh38
rs192397446 Chr.14: 93187127-93187127 on Build GRCh38
rs771017064 Chr.14: 93187098-93187098 on Build GRCh38
rs533609211 Chr.14: 93187043-93187043 on Build GRCh38
rs199810758 Chr.14: 93186795-93186796 on Build GRCh38
rs1039057182 Chr.14: 93186916-93186916 on Build GRCh38
rs901861310 Chr.14: 93186920-93186920 on Build GRCh38
rs543135912 Chr.14: 93186963-93186963 on Build GRCh38
rs1049724968 Chr.14: 93186870-93186870 on Build GRCh38
rs887940571 Chr.14: 93186906-93186906 on Build GRCh38

View Full Product Details