Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 58019831-58020345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USB1
Entrez Gene ID
79650
Gene Name
U6 snRNA biogenesis phosphodiesterase 1
Gene Aliases
C16orf57, HVSL1, Mpn1, PN, hUsb1
UniGene
Hs.408702
Gene Chromosome Location
Chr.16: 57999600-58021623 on Build GRCh38
Transcripts
NM_001195302.1 NM_024598.3 XM_005256144.3 XM_011523328.2 XM_011523329.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
971943 c.734G>A p.R245H Chr.16: 58020181-58020181 on Build GRCh38
3712053 c.759C>A p.C253* Chr.16: 58020206-58020206 on Build GRCh38
5515601 c.733C>T p.R245C Chr.16: 58020180-58020180 on Build GRCh38
1198175 c.755G>A p.R252H Chr.16: 58020202-58020202 on Build GRCh38
5964867 c.777C>T p.F259F Chr.16: 58020224-58020224 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760767630 Chr.16: 58020242-58020242 on Build GRCh38
rs1011390674 Chr.16: 58020040-58020040 on Build GRCh38
rs980800943 Chr.16: 58020046-58020046 on Build GRCh38
rs906865294 Chr.16: 58019924-58019924 on Build GRCh38
rs914167381 Chr.16: 58020238-58020238 on Build GRCh38
rs1038243600 Chr.16: 58020134-58020134 on Build GRCh38
rs28504996 Chr.16: 58019917-58019917 on Build GRCh38
rs536703103 Chr.16: 58020082-58020082 on Build GRCh38
rs143382202 Chr.16: 58020231-58020231 on Build GRCh38
rs748907589 Chr.16: 58020055-58020055 on Build GRCh38
rs774669193 Chr.16: 58020146-58020146 on Build GRCh38
rs762033139 Chr.16: 58020147-58020147 on Build GRCh38
rs1008669281 Chr.16: 58019975-58019975 on Build GRCh38
rs979665524 Chr.16: 58020319-58020319 on Build GRCh38
rs768771510 Chr.16: 58020125-58020125 on Build GRCh38
rs1002648049 Chr.16: 58019944-58019944 on Build GRCh38
rs370086533 Chr.16: 58020133-58020133 on Build GRCh38
rs536981078 Chr.16: 58020018-58020018 on Build GRCh38
rs763780991 Chr.16: 58020182-58020182 on Build GRCh38
rs16959641 Chr.16: 58020195-58020195 on Build GRCh38
rs766841291 Chr.16: 58020172-58020172 on Build GRCh38
rs1047204554 Chr.16: 58019923-58019923 on Build GRCh38
rs999873471 Chr.16: 58019912-58019912 on Build GRCh38
rs868421195 Chr.16: 58020263-58020263 on Build GRCh38
rs771359873 Chr.16: 58020247-58020247 on Build GRCh38
rs769032208 Chr.16: 58020144-58020144 on Build GRCh38
rs774051099 Chr.16: 58020285-58020285 on Build GRCh38
rs146419360 Chr.16: 58020201-58020201 on Build GRCh38
rs1041826191 Chr.16: 58020083-58020083 on Build GRCh38
rs769693185 Chr.16: 58020126-58020126 on Build GRCh38
rs149041481 Chr.16: 58020218-58020218 on Build GRCh38
rs778127933 Chr.16: 58019875-58019875 on Build GRCh38
rs759932035 Chr.16: 58020259-58020259 on Build GRCh38
rs1018834431 Chr.16: 58020151-58020151 on Build GRCh38
rs190283472 Chr.16: 58020176-58020176 on Build GRCh38
rs761098113 Chr.16: 58020170-58020170 on Build GRCh38
rs772360295 Chr.16: 58020234-58020234 on Build GRCh38
rs914033441 Chr.16: 58020074-58020074 on Build GRCh38
rs897612646 Chr.16: 58020171-58020171 on Build GRCh38
rs967273752 Chr.16: 58020313-58020313 on Build GRCh38
rs375820775 Chr.16: 58020127-58020127 on Build GRCh38
rs140289507 Chr.16: 58020181-58020181 on Build GRCh38
rs185437684 Chr.16: 58020099-58020099 on Build GRCh38
rs935668829 Chr.16: 58020118-58020118 on Build GRCh38
rs756831672 Chr.16: 58020192-58020192 on Build GRCh38
rs772272501 Chr.16: 58020159-58020159 on Build GRCh38
rs576904006 Chr.16: 58020116-58020116 on Build GRCh38
rs889073580 Chr.16: 58019952-58019952 on Build GRCh38
rs765579501 Chr.16: 58020269-58020269 on Build GRCh38
rs779320272 Chr.16: 58020230-58020230 on Build GRCh38
rs368558020 Chr.16: 58020229-58020229 on Build GRCh38
rs777021661 Chr.16: 58020134-58020134 on Build GRCh38
rs779874096 Chr.16: 58020050-58020050 on Build GRCh38
rs961915602 Chr.16: 58020009-58020009 on Build GRCh38
rs781610947 Chr.16: 58020062-58020062 on Build GRCh38
rs780003077 Chr.16: 58020202-58020202 on Build GRCh38
rs773826400 Chr.16: 58020163-58020163 on Build GRCh38
rs751177454 Chr.16: 58020183-58020183 on Build GRCh38
rs777169639 Chr.16: 58020249-58020249 on Build GRCh38
rs182418203 Chr.16: 58020180-58020180 on Build GRCh38
rs570256320 Chr.16: 58020041-58020041 on Build GRCh38
rs867904652 Chr.16: 58020005-58020005 on Build GRCh38
rs773169264 Chr.16: 58020235-58020235 on Build GRCh38
rs12102539 Chr.16: 58020027-58020027 on Build GRCh38
rs1021911386 Chr.16: 58020224-58020224 on Build GRCh38
rs986056898 Chr.16: 58020254-58020254 on Build GRCh38

View Full Product Details