Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 868284-868814 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM41C
Entrez Gene ID
284593
Gene Name
family with sequence similarity 41 member C
Gene Aliases
-
UniGene
Hs.449006
Gene Chromosome Location
Chr.1: 868071-876802 on Build GRCh38
Transcripts
NR_027055.1

SNP Detail

SNP ID Chromosome Location
rs537050962 Chr.1: 868739-868739 on Build GRCh38
rs1000726994 Chr.1: 868537-868537 on Build GRCh38
rs781526692 Chr.1: 868629-868630 on Build GRCh38
rs796458342 Chr.1: 868322-868322 on Build GRCh38
rs559492584 Chr.1: 868373-868374 on Build GRCh38
rs867119210 Chr.1: 868416-868416 on Build GRCh38
rs563372565 Chr.1: 868553-868553 on Build GRCh38
rs991915454 Chr.1: 868614-868614 on Build GRCh38
rs573479201 Chr.1: 868772-868772 on Build GRCh38
rs1009325247 Chr.1: 868468-868468 on Build GRCh38
rs553317956 Chr.1: 868755-868755 on Build GRCh38
rs552119867 Chr.1: 868758-868758 on Build GRCh38
rs35452722 Chr.1: 868561-868562 on Build GRCh38
rs1050027965 Chr.1: 868374-868374 on Build GRCh38
rs959198587 Chr.1: 868599-868599 on Build GRCh38
rs565945225 Chr.1: 868660-868660 on Build GRCh38
rs184328933 Chr.1: 868741-868741 on Build GRCh38
rs372466154 Chr.1: 868635-868635 on Build GRCh38
rs1040143121 Chr.1: 868431-868431 on Build GRCh38
rs533047898 Chr.1: 868382-868382 on Build GRCh38
rs536431802 Chr.1: 868752-868752 on Build GRCh38
rs551746123 Chr.1: 868729-868729 on Build GRCh38
rs563972649 Chr.1: 868378-868378 on Build GRCh38
rs1037406244 Chr.1: 868310-868310 on Build GRCh38
rs901550009 Chr.1: 868465-868466 on Build GRCh38
rs1021008848 Chr.1: 868479-868479 on Build GRCh38
rs879017649 Chr.1: 868731-868731 on Build GRCh38
rs9725068 Chr.1: 868735-868735 on Build GRCh38
rs1041573711 Chr.1: 868778-868778 on Build GRCh38
rs573813995 Chr.1: 868745-868745 on Build GRCh38
rs549859341 Chr.1: 868520-868520 on Build GRCh38
rs944273274 Chr.1: 868764-868764 on Build GRCh38
rs985701114 Chr.1: 868744-868744 on Build GRCh38
rs376969480 Chr.1: 868693-868693 on Build GRCh38
rs750333900 Chr.1: 868710-868710 on Build GRCh38
rs117720955 Chr.1: 868633-868633 on Build GRCh38
rs1008434346 Chr.1: 868389-868389 on Build GRCh38
rs972860255 Chr.1: 868683-868683 on Build GRCh38
rs545418299 Chr.1: 868788-868788 on Build GRCh38
rs143127861 Chr.1: 868326-868326 on Build GRCh38
rs534659898 Chr.1: 868727-868727 on Build GRCh38

View Full Product Details