Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 978712-979202 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PERM1
Entrez Gene ID
84808
Gene Name
PPARGC1 and ESRR induced regulator, muscle 1
Gene Aliases
C1orf170
UniGene
Hs.271462
Gene Chromosome Location
Chr.1: 975198-982117 on Build GRCh38
Transcripts
NM_001291366.1 NM_001291367.1 XM_017002583.1 XM_017002584.1 XM_017002585.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4445463 c.1656G>A p.G552G Chr.1: 979032-979032 on Build GRCh38
4445462 c.1709C>T p.A570V Chr.1: 978979-978979 on Build GRCh38
5969582 c.1685C>T p.P562L Chr.1: 979003-979003 on Build GRCh38
4969117 c.1553G>T p.G518V Chr.1: 979135-979135 on Build GRCh38
2259584 c.1496C>T p.P499L Chr.1: 979192-979192 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761366853 Chr.1: 978825-978825 on Build GRCh38
rs988698064 Chr.1: 978733-978733 on Build GRCh38
rs777332674 Chr.1: 979113-979113 on Build GRCh38
rs945851953 Chr.1: 978774-978774 on Build GRCh38
rs772976435 Chr.1: 979077-979077 on Build GRCh38
rs2340595 Chr.1: 978812-978812 on Build GRCh38
rs768118255 Chr.1: 978797-978797 on Build GRCh38
rs750624982 Chr.1: 979139-979139 on Build GRCh38
rs775387917 Chr.1: 978868-978868 on Build GRCh38
rs755873632 Chr.1: 979014-979014 on Build GRCh38
rs978984739 Chr.1: 978940-978940 on Build GRCh38
rs371221955 Chr.1: 978804-978804 on Build GRCh38
rs904427765 Chr.1: 979087-979087 on Build GRCh38
rs1027788790 Chr.1: 979072-979072 on Build GRCh38
rs573314277 Chr.1: 979098-979098 on Build GRCh38
rs750848990 Chr.1: 978917-978917 on Build GRCh38
rs552592144 Chr.1: 978887-978887 on Build GRCh38
rs761157931 Chr.1: 979082-979082 on Build GRCh38
rs575660840 Chr.1: 979128-979128 on Build GRCh38
rs1007671288 Chr.1: 979164-979164 on Build GRCh38
rs571296882 Chr.1: 978964-978964 on Build GRCh38
rs765531563 Chr.1: 978908-978908 on Build GRCh38
rs779857282 Chr.1: 979147-979147 on Build GRCh38
rs540568745 Chr.1: 979182-979182 on Build GRCh38
rs544500813 Chr.1: 979132-979132 on Build GRCh38
rs937415720 Chr.1: 979003-979003 on Build GRCh38
rs770092392 Chr.1: 978921-978921 on Build GRCh38
rs764219592 Chr.1: 978938-978938 on Build GRCh38
rs771192500 Chr.1: 979138-979138 on Build GRCh38
rs372753735 Chr.1: 978996-978996 on Build GRCh38
rs769750320 Chr.1: 978846-978846 on Build GRCh38
rs752757961 Chr.1: 978960-978960 on Build GRCh38
rs773737688 Chr.1: 979038-979038 on Build GRCh38
rs904502571 Chr.1: 979170-979170 on Build GRCh38
rs771873640 Chr.1: 979055-979055 on Build GRCh38
rs759972334 Chr.1: 979122-979122 on Build GRCh38
rs1049026513 Chr.1: 979002-979002 on Build GRCh38
rs937132128 Chr.1: 978795-978795 on Build GRCh38
rs758582513 Chr.1: 978961-978961 on Build GRCh38
rs1037204423 Chr.1: 978851-978851 on Build GRCh38
rs950134049 Chr.1: 979137-979137 on Build GRCh38
rs745715987 Chr.1: 979013-979013 on Build GRCh38
rs565368985 Chr.1: 978855-978855 on Build GRCh38
rs759928584 Chr.1: 978905-978905 on Build GRCh38
rs779838732 Chr.1: 979179-979179 on Build GRCh38
rs574803551 Chr.1: 979146-979146 on Build GRCh38
rs961653576 Chr.1: 978968-978968 on Build GRCh38
rs544777665 Chr.1: 978883-978883 on Build GRCh38
rs567468943 Chr.1: 979016-979016 on Build GRCh38
rs991594494 Chr.1: 979130-979130 on Build GRCh38
rs995399609 Chr.1: 978924-978924 on Build GRCh38
rs757347252 Chr.1: 979032-979032 on Build GRCh38
rs918834820 Chr.1: 978981-978981 on Build GRCh38
rs771720044 Chr.1: 978840-978840 on Build GRCh38
rs201808865 Chr.1: 979163-979163 on Build GRCh38
rs748680927 Chr.1: 978848-978848 on Build GRCh38
rs945957682 Chr.1: 978994-978994 on Build GRCh38
rs72891183 Chr.1: 979141-979141 on Build GRCh38
rs536503119 Chr.1: 979034-979034 on Build GRCh38
rs1027749048 Chr.1: 978928-978929 on Build GRCh38
rs748758211 Chr.1: 979035-979037 on Build GRCh38
rs570864131 Chr.1: 978918-978918 on Build GRCh38
rs895785508 Chr.1: 978827-978827 on Build GRCh38
rs530392317 Chr.1: 978885-978885 on Build GRCh38
rs528069699 Chr.1: 978946-978946 on Build GRCh38
rs753186055 Chr.1: 978912-978912 on Build GRCh38
rs978651719 Chr.1: 978754-978754 on Build GRCh38
rs950072658 Chr.1: 979035-979035 on Build GRCh38
rs761386654 Chr.1: 978881-978881 on Build GRCh38
rs561439651 Chr.1: 978884-978884 on Build GRCh38
rs553412088 Chr.1: 979074-979074 on Build GRCh38
rs767129412 Chr.1: 978907-978907 on Build GRCh38
rs539031218 Chr.1: 979099-979099 on Build GRCh38
rs1014541432 Chr.1: 978864-978864 on Build GRCh38
rs942879786 Chr.1: 979028-979028 on Build GRCh38
rs889849135 Chr.1: 978882-978882 on Build GRCh38
rs1017960343 Chr.1: 979092-979092 on Build GRCh38
rs1007004165 Chr.1: 978886-978886 on Build GRCh38
rs767038967 Chr.1: 978925-978925 on Build GRCh38
rs1020206705 Chr.1: 978934-978934 on Build GRCh38
rs913104036 Chr.1: 978750-978750 on Build GRCh38
rs140514242 Chr.1: 978945-978945 on Build GRCh38
rs536881591 Chr.1: 978978-978978 on Build GRCh38
rs113247740 Chr.1: 979123-979123 on Build GRCh38
rs940382085 Chr.1: 978838-978838 on Build GRCh38
rs747652699 Chr.1: 979053-979053 on Build GRCh38
rs927033272 Chr.1: 978772-978772 on Build GRCh38
rs13302979 Chr.1: 978953-978953 on Build GRCh38
rs1020216623 Chr.1: 978742-978742 on Build GRCh38
rs1019722252 Chr.1: 978894-978894 on Build GRCh38
rs774113351 Chr.1: 978876-978876 on Build GRCh38
rs898083103 Chr.1: 979039-979039 on Build GRCh38
rs1014509838 Chr.1: 979116-979116 on Build GRCh38
rs763189910 Chr.1: 979117-979117 on Build GRCh38
rs762491595 Chr.1: 978869-978869 on Build GRCh38
rs1041839859 Chr.1: 979166-979166 on Build GRCh38
rs1055805507 Chr.1: 978820-978820 on Build GRCh38
rs968507387 Chr.1: 978746-978746 on Build GRCh38
rs1000608524 Chr.1: 979181-979181 on Build GRCh38
rs930930835 Chr.1: 979142-979142 on Build GRCh38
rs937152501 Chr.1: 979129-979129 on Build GRCh38
rs531040994 Chr.1: 978870-978870 on Build GRCh38

View Full Product Details