Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 979744-980247 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PERM1
Entrez Gene ID
84808
Gene Name
PPARGC1 and ESRR induced regulator, muscle 1
Gene Aliases
C1orf170
UniGene
Hs.271462
Gene Chromosome Location
Chr.1: 975198-982117 on Build GRCh38
Transcripts
NM_001291366.1 NM_001291367.1 XM_017002583.1 XM_017002584.1 XM_017002585.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5810275 c.647T>A p.V216E Chr.1: 980041-980041 on Build GRCh38
6455155 c.708T>C p.S236S Chr.1: 979980-979980 on Build GRCh38
3711406 c.880C>A p.H294N Chr.1: 979808-979808 on Build GRCh38
4596940 c.479G>T p.R160L Chr.1: 980209-980209 on Build GRCh38
2259592 c.635G>C p.R212T Chr.1: 980053-980053 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770912161 Chr.1: 979859-979859 on Build GRCh38
rs531767214 Chr.1: 980073-980073 on Build GRCh38
rs529787043 Chr.1: 979935-979935 on Build GRCh38
rs907342141 Chr.1: 980044-980044 on Build GRCh38
rs779262383 Chr.1: 979822-979822 on Build GRCh38
rs374701884 Chr.1: 980193-980194 on Build GRCh38
rs754368752 Chr.1: 979964-980008 on Build GRCh38
rs936805991 Chr.1: 979839-979839 on Build GRCh38
rs560349694 Chr.1: 979948-979948 on Build GRCh38
rs569036408 Chr.1: 980020-980020 on Build GRCh38
rs1047633190 Chr.1: 979906-979906 on Build GRCh38
rs780403874 Chr.1: 980060-980060 on Build GRCh38
rs907394384 Chr.1: 979865-979865 on Build GRCh38
rs770166811 Chr.1: 980053-980053 on Build GRCh38
rs748012761 Chr.1: 980064-980064 on Build GRCh38
rs1013498140 Chr.1: 979835-979835 on Build GRCh38
rs767542373 Chr.1: 979944-979944 on Build GRCh38
rs576645567 Chr.1: 980209-980209 on Build GRCh38
rs915678052 Chr.1: 979854-979854 on Build GRCh38
rs923682963 Chr.1: 980184-980184 on Build GRCh38
rs764828181 Chr.1: 979989-979989 on Build GRCh38
rs887634645 Chr.1: 979911-979911 on Build GRCh38
rs977795228 Chr.1: 979980-979980 on Build GRCh38
rs777052561 Chr.1: 979866-979866 on Build GRCh38
rs759191878 Chr.1: 979988-979988 on Build GRCh38
rs1016595570 Chr.1: 979930-979930 on Build GRCh38
rs13303355 Chr.1: 979847-979847 on Build GRCh38
rs747154679 Chr.1: 979857-979857 on Build GRCh38
rs532200365 Chr.1: 980000-980000 on Build GRCh38
rs553953341 Chr.1: 980148-980148 on Build GRCh38
rs997969634 Chr.1: 979931-979931 on Build GRCh38
rs923068922 Chr.1: 979970-979970 on Build GRCh38
rs936326528 Chr.1: 979992-979992 on Build GRCh38
rs200050434 Chr.1: 980115-980115 on Build GRCh38
rs947727148 Chr.1: 980215-980215 on Build GRCh38
rs758996080 Chr.1: 980114-980114 on Build GRCh38
rs758662102 Chr.1: 980007-980007 on Build GRCh38
rs556569834 Chr.1: 979871-979871 on Build GRCh38
rs550142087 Chr.1: 980218-980218 on Build GRCh38
rs780666767 Chr.1: 979945-979946 on Build GRCh38
rs1030338645 Chr.1: 979941-979941 on Build GRCh38
rs765857814 Chr.1: 979974-979974 on Build GRCh38
rs753283423 Chr.1: 979976-979976 on Build GRCh38
rs760296428 Chr.1: 979955-979955 on Build GRCh38
rs773255636 Chr.1: 980069-980069 on Build GRCh38
rs929275810 Chr.1: 979893-979893 on Build GRCh38
rs746101959 Chr.1: 979872-979872 on Build GRCh38
rs112622122 Chr.1: 980019-980019 on Build GRCh38
rs760538801 Chr.1: 980080-980080 on Build GRCh38
rs753125814 Chr.1: 979997-979997 on Build GRCh38
rs577120404 Chr.1: 979831-979831 on Build GRCh38
rs755389421 Chr.1: 979812-979812 on Build GRCh38
rs745907527 Chr.1: 980034-980034 on Build GRCh38
rs553918278 Chr.1: 979820-979820 on Build GRCh38
rs763434938 Chr.1: 980149-980149 on Build GRCh38
rs868530532 Chr.1: 980105-980105 on Build GRCh38
rs138027487 Chr.1: 980110-980110 on Build GRCh38
rs188954903 Chr.1: 980185-980185 on Build GRCh38
rs866876428 Chr.1: 980098-980098 on Build GRCh38
rs1055082558 Chr.1: 979805-979805 on Build GRCh38
rs771542003 Chr.1: 979952-979952 on Build GRCh38
rs533981218 Chr.1: 980112-980112 on Build GRCh38
rs181321538 Chr.1: 980214-980214 on Build GRCh38
rs560999553 Chr.1: 979918-979918 on Build GRCh38
rs781747747 Chr.1: 980190-980190 on Build GRCh38
rs143775054 Chr.1: 980212-980212 on Build GRCh38
rs200169464 Chr.1: 980213-980213 on Build GRCh38
rs1005994751 Chr.1: 979916-979916 on Build GRCh38
rs753482081 Chr.1: 979790-979790 on Build GRCh38
rs778412567 Chr.1: 980008-980008 on Build GRCh38
rs570750775 Chr.1: 980152-980152 on Build GRCh38
rs964684993 Chr.1: 979943-979943 on Build GRCh38
rs751885271 Chr.1: 980157-980157 on Build GRCh38
rs756234018 Chr.1: 980210-980210 on Build GRCh38
rs540430710 Chr.1: 979945-979945 on Build GRCh38
rs748444328 Chr.1: 979828-979828 on Build GRCh38
rs771756338 Chr.1: 980065-980065 on Build GRCh38
rs941926368 Chr.1: 980162-980162 on Build GRCh38
rs113243246 Chr.1: 979884-979884 on Build GRCh38
rs752289239 Chr.1: 980125-980125 on Build GRCh38
rs543839758 Chr.1: 979917-979917 on Build GRCh38
rs114358412 Chr.1: 980163-980163 on Build GRCh38
rs548142321 Chr.1: 980109-980109 on Build GRCh38

View Full Product Details