Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 173865496-173866041 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAS5
Entrez Gene ID
60674
Gene Name
growth arrest specific 5 (non-protein coding)
Gene Aliases
NCRNA00030, SNHG2
UniGene
Hs.531856
Gene Chromosome Location
Chr.1: 173863901-173867987 on Build GRCh38
Transcripts
NR_002578.2 NR_002750.2 NR_003944.1

Gene Symbol
SNORD44
Entrez Gene ID
26806
Gene Name
small nucleolar RNA, C/D box 44
Gene Aliases
RNU44, U44
UniGene
Gene Chromosome Location
Chr.1: 173865968-173866028 on Build GRCh38
Transcripts
NR_002578.2 NR_002750.2 NR_003944.1

Gene Symbol
SNORD78
Entrez Gene ID
692198
Gene Name
small nucleolar RNA, C/D box 78
Gene Aliases
U78
UniGene
Gene Chromosome Location
Chr.1: 173865633-173865686 on Build GRCh38
Transcripts
NR_002578.2 NR_002750.2 NR_003944.1

SNP Detail

SNP ID Chromosome Location
rs755862759 Chr.1: 173866017-173866017 on Build GRCh38
rs778136322 Chr.1: 173865694-173865694 on Build GRCh38
rs545738307 Chr.1: 173865782-173865782 on Build GRCh38
rs1004116595 Chr.1: 173865707-173865707 on Build GRCh38
rs757856797 Chr.1: 173865653-173865653 on Build GRCh38
rs370941080 Chr.1: 173865578-173865578 on Build GRCh38
rs746509971 Chr.1: 173865657-173865657 on Build GRCh38
rs903059976 Chr.1: 173865910-173865910 on Build GRCh38
rs772103330 Chr.1: 173865976-173865976 on Build GRCh38
rs989847075 Chr.1: 173865887-173865887 on Build GRCh38
rs748252116 Chr.1: 173865966-173865966 on Build GRCh38
rs768621674 Chr.1: 173865943-173865943 on Build GRCh38
rs778473861 Chr.1: 173865718-173865721 on Build GRCh38
rs140290587 Chr.1: 173865929-173865929 on Build GRCh38
rs754135848 Chr.1: 173865723-173865723 on Build GRCh38
rs975519098 Chr.1: 173865938-173865938 on Build GRCh38
rs756777696 Chr.1: 173865917-173865917 on Build GRCh38
rs943390832 Chr.1: 173865754-173865754 on Build GRCh38
rs755785244 Chr.1: 173865617-173865626 on Build GRCh38
rs893069323 Chr.1: 173865835-173865835 on Build GRCh38
rs748940309 Chr.1: 173865645-173865645 on Build GRCh38
rs771981446 Chr.1: 173865940-173865940 on Build GRCh38
rs947172199 Chr.1: 173865856-173865856 on Build GRCh38
rs777364347 Chr.1: 173865635-173865635 on Build GRCh38
rs775442364 Chr.1: 173865951-173865954 on Build GRCh38
rs374299531 Chr.1: 173865690-173865690 on Build GRCh38
rs770397079 Chr.1: 173865673-173865675 on Build GRCh38
rs12501 Chr.1: 173865862-173865862 on Build GRCh38
rs756481773 Chr.1: 173865633-173865633 on Build GRCh38
rs1041556997 Chr.1: 173865878-173865878 on Build GRCh38
rs577553605 Chr.1: 173865902-173865902 on Build GRCh38
rs756449931 Chr.1: 173865676-173865678 on Build GRCh38
rs971101548 Chr.1: 173865803-173865803 on Build GRCh38
rs745554616 Chr.1: 173865744-173865744 on Build GRCh38
rs14262 Chr.1: 173865545-173865545 on Build GRCh38
rs987315187 Chr.1: 173865955-173865955 on Build GRCh38
rs528960644 Chr.1: 173866012-173866012 on Build GRCh38
rs751862476 Chr.1: 173865601-173865609 on Build GRCh38
rs565468741 Chr.1: 173865870-173865870 on Build GRCh38
rs753212754 Chr.1: 173866008-173866008 on Build GRCh38
rs765442504 Chr.1: 173865720-173865721 on Build GRCh38
rs750829389 Chr.1: 173865617-173865617 on Build GRCh38
rs145305710 Chr.1: 173865940-173865940 on Build GRCh38
rs745669877 Chr.1: 173865686-173865686 on Build GRCh38
rs762451946 Chr.1: 173865712-173865712 on Build GRCh38
rs751160537 Chr.1: 173865735-173865735 on Build GRCh38
rs780968093 Chr.1: 173865681-173865681 on Build GRCh38
rs762892191 Chr.1: 173865692-173865692 on Build GRCh38
rs764168883 Chr.1: 173865590-173865590 on Build GRCh38
rs745457095 Chr.1: 173865923-173865923 on Build GRCh38
rs988837304 Chr.1: 173865581-173865581 on Build GRCh38
rs993734145 Chr.1: 173865988-173865988 on Build GRCh38
rs762082420 Chr.1: 173865609-173865609 on Build GRCh38
rs146717094 Chr.1: 173865756-173865756 on Build GRCh38
rs185805390 Chr.1: 173865550-173865550 on Build GRCh38
rs780451525 Chr.1: 173865635-173865635 on Build GRCh38
rs762973214 Chr.1: 173865587-173865587 on Build GRCh38
rs774253252 Chr.1: 173865945-173865945 on Build GRCh38
rs777091877 Chr.1: 173865997-173865997 on Build GRCh38
rs755571777 Chr.1: 173865726-173865726 on Build GRCh38
rs939985678 Chr.1: 173865979-173865979 on Build GRCh38
rs770743575 Chr.1: 173865671-173865671 on Build GRCh38
rs753164616 Chr.1: 173865572-173865572 on Build GRCh38
rs768135323 Chr.1: 173865743-173865743 on Build GRCh38
rs375212679 Chr.1: 173865536-173865536 on Build GRCh38
rs1002617054 Chr.1: 173865916-173865916 on Build GRCh38
rs775449543 Chr.1: 173865582-173865582 on Build GRCh38
rs559123072 Chr.1: 173865972-173865975 on Build GRCh38
rs766376871 Chr.1: 173865876-173865876 on Build GRCh38
rs536835068 Chr.1: 173865680-173865680 on Build GRCh38
rs766702633 Chr.1: 173865722-173865722 on Build GRCh38
rs780399356 Chr.1: 173865597-173865598 on Build GRCh38
rs955920578 Chr.1: 173865853-173865853 on Build GRCh38
rs775293030 Chr.1: 173865691-173865691 on Build GRCh38
rs759794258 Chr.1: 173866000-173866000 on Build GRCh38
rs201246074 Chr.1: 173866013-173866013 on Build GRCh38
rs1035354710 Chr.1: 173865529-173865529 on Build GRCh38
rs1038719060 Chr.1: 173865708-173865708 on Build GRCh38
rs924278781 Chr.1: 173865563-173865563 on Build GRCh38
rs1031332681 Chr.1: 173865850-173865850 on Build GRCh38
rs766501603 Chr.1: 173865992-173865992 on Build GRCh38
rs775264686 Chr.1: 173866016-173866018 on Build GRCh38
rs767723393 Chr.1: 173865612-173865612 on Build GRCh38
rs865807941 Chr.1: 173865771-173865771 on Build GRCh38
rs765865942 Chr.1: 173866007-173866007 on Build GRCh38
rs373937676 Chr.1: 173865664-173865664 on Build GRCh38
rs761851569 Chr.1: 173865698-173865698 on Build GRCh38
rs758937173 Chr.1: 173865583-173865585 on Build GRCh38
rs773314679 Chr.1: 173865983-173865983 on Build GRCh38
rs754333723 Chr.1: 173865646-173865646 on Build GRCh38
rs769491214 Chr.1: 173865928-173865928 on Build GRCh38
rs201497600 Chr.1: 173865703-173865703 on Build GRCh38
rs767778804 Chr.1: 173865699-173865699 on Build GRCh38
rs762004136 Chr.1: 173865854-173865854 on Build GRCh38
rs574090474 Chr.1: 173865591-173865591 on Build GRCh38
rs1003069702 Chr.1: 173865815-173865815 on Build GRCh38
rs781023035 Chr.1: 173865920-173865920 on Build GRCh38
rs868375277 Chr.1: 173865757-173865757 on Build GRCh38
rs761011912 Chr.1: 173865985-173865985 on Build GRCh38
rs867922479 Chr.1: 173865843-173865843 on Build GRCh38
rs374406074 Chr.1: 173865732-173865732 on Build GRCh38
rs750596949 Chr.1: 173865701-173865701 on Build GRCh38
rs35207866 Chr.1: 173865712-173865713 on Build GRCh38

View Full Product Details