Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 173865863-173866360 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAS5
Entrez Gene ID
60674
Gene Name
growth arrest specific 5 (non-protein coding)
Gene Aliases
NCRNA00030, SNHG2
UniGene
Hs.531856
Gene Chromosome Location
Chr.1: 173863901-173867987 on Build GRCh38
Transcripts
NR_002578.2 NR_132783.1 NR_002750.2 NR_003943.1

Gene Symbol
SNORA103
Entrez Gene ID
106635533
Gene Name
small nucleolar RNA, H/ACA box 103
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.1: 173866205-173866290 on Build GRCh38
Transcripts
NR_002578.2 NR_132783.1 NR_002750.2 NR_003943.1

Gene Symbol
SNORD44
Entrez Gene ID
26806
Gene Name
small nucleolar RNA, C/D box 44
Gene Aliases
RNU44, U44
UniGene
Gene Chromosome Location
Chr.1: 173865968-173866028 on Build GRCh38
Transcripts
NR_002578.2 NR_132783.1 NR_002750.2 NR_003943.1

Gene Symbol
SNORD77
Entrez Gene ID
692197
Gene Name
small nucleolar RNA, C/D box 77
Gene Aliases
SNORD77A, U77
UniGene
Gene Chromosome Location
Chr.1: 173866311-173866371 on Build GRCh38
Transcripts
NR_002578.2 NR_132783.1 NR_002750.2 NR_003943.1

SNP Detail

SNP ID Chromosome Location
rs779543269 Chr.1: 173866027-173866027 on Build GRCh38
rs755862759 Chr.1: 173866017-173866017 on Build GRCh38
rs903059976 Chr.1: 173865910-173865910 on Build GRCh38
rs772103330 Chr.1: 173865976-173865976 on Build GRCh38
rs948615821 Chr.1: 173866211-173866211 on Build GRCh38
rs989847075 Chr.1: 173865887-173865887 on Build GRCh38
rs748252116 Chr.1: 173865966-173865966 on Build GRCh38
rs572699556 Chr.1: 173866305-173866305 on Build GRCh38
rs371808067 Chr.1: 173866045-173866045 on Build GRCh38
rs768621674 Chr.1: 173865943-173865943 on Build GRCh38
rs140290587 Chr.1: 173865929-173865929 on Build GRCh38
rs1011315612 Chr.1: 173866248-173866248 on Build GRCh38
rs975519098 Chr.1: 173865938-173865938 on Build GRCh38
rs756932465 Chr.1: 173866321-173866322 on Build GRCh38
rs756777696 Chr.1: 173865917-173865917 on Build GRCh38
rs753395193 Chr.1: 173866259-173866259 on Build GRCh38
rs891505186 Chr.1: 173866237-173866237 on Build GRCh38
rs747929634 Chr.1: 173866264-173866264 on Build GRCh38
rs778293523 Chr.1: 173866325-173866326 on Build GRCh38
rs907022036 Chr.1: 173866220-173866220 on Build GRCh38
rs369212964 Chr.1: 173866331-173866331 on Build GRCh38
rs762078905 Chr.1: 173866049-173866052 on Build GRCh38
rs771981446 Chr.1: 173865940-173865940 on Build GRCh38
rs775442364 Chr.1: 173865951-173865954 on Build GRCh38
rs763382313 Chr.1: 173866077-173866080 on Build GRCh38
rs531471279 Chr.1: 173866074-173866074 on Build GRCh38
rs563426421 Chr.1: 173866323-173866323 on Build GRCh38
rs192922148 Chr.1: 173866333-173866333 on Build GRCh38
rs772156464 Chr.1: 173866056-173866056 on Build GRCh38
rs781479641 Chr.1: 173866321-173866321 on Build GRCh38
rs754776961 Chr.1: 173866260-173866260 on Build GRCh38
rs577553605 Chr.1: 173865902-173865902 on Build GRCh38
rs530842847 Chr.1: 173866325-173866325 on Build GRCh38
rs975678876 Chr.1: 173866105-173866105 on Build GRCh38
rs773403339 Chr.1: 173866057-173866057 on Build GRCh38
rs766821976 Chr.1: 173866266-173866268 on Build GRCh38
rs964809795 Chr.1: 173866226-173866226 on Build GRCh38
rs764504768 Chr.1: 173866256-173866256 on Build GRCh38
rs567704467 Chr.1: 173866153-173866153 on Build GRCh38
rs747132163 Chr.1: 173866315-173866315 on Build GRCh38
rs987315187 Chr.1: 173865955-173865955 on Build GRCh38
rs187820495 Chr.1: 173866183-173866183 on Build GRCh38
rs528960644 Chr.1: 173866012-173866012 on Build GRCh38
rs753212754 Chr.1: 173866008-173866008 on Build GRCh38
rs376962886 Chr.1: 173866257-173866257 on Build GRCh38
rs369159932 Chr.1: 173866250-173866250 on Build GRCh38
rs777038575 Chr.1: 173866042-173866045 on Build GRCh38
rs145305710 Chr.1: 173865940-173865940 on Build GRCh38
rs1002607397 Chr.1: 173866052-173866052 on Build GRCh38
rs758435682 Chr.1: 173866332-173866336 on Build GRCh38
rs997043176 Chr.1: 173866311-173866311 on Build GRCh38
rs908595307 Chr.1: 173866176-173866176 on Build GRCh38
rs745457095 Chr.1: 173865923-173865923 on Build GRCh38
rs778856850 Chr.1: 173866046-173866046 on Build GRCh38
rs114972656 Chr.1: 173866209-173866209 on Build GRCh38
rs993734145 Chr.1: 173865988-173865988 on Build GRCh38
rs745329212 Chr.1: 173866331-173866333 on Build GRCh38
rs1028431332 Chr.1: 173866126-173866126 on Build GRCh38
rs898875112 Chr.1: 173866171-173866171 on Build GRCh38
rs915240464 Chr.1: 173866267-173866267 on Build GRCh38
rs771231173 Chr.1: 173866062-173866062 on Build GRCh38
rs774253252 Chr.1: 173865945-173865945 on Build GRCh38
rs16846493 Chr.1: 173866088-173866088 on Build GRCh38
rs551628123 Chr.1: 173866078-173866078 on Build GRCh38
rs939985678 Chr.1: 173865979-173865979 on Build GRCh38
rs777091877 Chr.1: 173865997-173865997 on Build GRCh38
rs367653326 Chr.1: 173866164-173866165 on Build GRCh38
rs986998749 Chr.1: 173866155-173866155 on Build GRCh38
rs1052962173 Chr.1: 173866229-173866229 on Build GRCh38
rs544949882 Chr.1: 173866094-173866094 on Build GRCh38
rs758213780 Chr.1: 173866091-173866091 on Build GRCh38
rs778626036 Chr.1: 173866262-173866262 on Build GRCh38
rs1002617054 Chr.1: 173865916-173865916 on Build GRCh38
rs563547318 Chr.1: 173866309-173866312 on Build GRCh38
rs559123072 Chr.1: 173865972-173865975 on Build GRCh38
rs755376938 Chr.1: 173866306-173866308 on Build GRCh38
rs777546426 Chr.1: 173866309-173866309 on Build GRCh38
rs765297755 Chr.1: 173866058-173866060 on Build GRCh38
rs749158368 Chr.1: 173866034-173866034 on Build GRCh38
rs915254241 Chr.1: 173866087-173866087 on Build GRCh38
rs746512954 Chr.1: 173866063-173866063 on Build GRCh38
rs759794258 Chr.1: 173866000-173866000 on Build GRCh38
rs761406485 Chr.1: 173866163-173866164 on Build GRCh38
rs1009037298 Chr.1: 173866222-173866222 on Build GRCh38
rs763613766 Chr.1: 173866252-173866252 on Build GRCh38
rs751952352 Chr.1: 173866296-173866297 on Build GRCh38
rs201246074 Chr.1: 173866013-173866013 on Build GRCh38
rs191700140 Chr.1: 173866127-173866127 on Build GRCh38
rs183737539 Chr.1: 173866128-173866128 on Build GRCh38
rs1041692808 Chr.1: 173866102-173866102 on Build GRCh38
rs201901194 Chr.1: 173866318-173866318 on Build GRCh38
rs766501603 Chr.1: 173865992-173865992 on Build GRCh38
rs775264686 Chr.1: 173866016-173866018 on Build GRCh38
rs755760054 Chr.1: 173866104-173866104 on Build GRCh38
rs542054439 Chr.1: 173866322-173866322 on Build GRCh38
rs765865942 Chr.1: 173866007-173866007 on Build GRCh38
rs766080595 Chr.1: 173866258-173866258 on Build GRCh38
rs777590875 Chr.1: 173866125-173866125 on Build GRCh38
rs773314679 Chr.1: 173865983-173865983 on Build GRCh38
rs974034193 Chr.1: 173866205-173866205 on Build GRCh38
rs758368948 Chr.1: 173866301-173866301 on Build GRCh38
rs769491214 Chr.1: 173865928-173865928 on Build GRCh38
rs539271020 Chr.1: 173866225-173866225 on Build GRCh38
rs2067079 Chr.1: 173866073-173866073 on Build GRCh38
rs770873429 Chr.1: 173866317-173866317 on Build GRCh38
rs781023035 Chr.1: 173865920-173865920 on Build GRCh38
rs761011912 Chr.1: 173865985-173865985 on Build GRCh38
rs750448960 Chr.1: 173866066-173866068 on Build GRCh38
rs200757519 Chr.1: 173866050-173866050 on Build GRCh38
rs137905547 Chr.1: 173866215-173866215 on Build GRCh38
rs747026122 Chr.1: 173866059-173866059 on Build GRCh38
rs924408176 Chr.1: 173866118-173866118 on Build GRCh38

View Full Product Details