Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 173866739-173867236 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAS5
Entrez Gene ID
60674
Gene Name
growth arrest specific 5 (non-protein coding)
Gene Aliases
NCRNA00030, SNHG2
UniGene
Hs.531856
Gene Chromosome Location
Chr.1: 173863901-173867987 on Build GRCh38
Transcripts
NR_002578.2 NR_003941.1

Gene Symbol
SNORD75
Entrez Gene ID
692195
Gene Name
small nucleolar RNA, C/D box 75
Gene Aliases
U75
UniGene
Gene Chromosome Location
Chr.1: 173866879-173866938 on Build GRCh38
Transcripts
NR_002578.2 NR_003941.1

SNP Detail

SNP ID Chromosome Location
rs1044037104 Chr.1: 173867087-173867088 on Build GRCh38
rs773178650 Chr.1: 173866936-173866936 on Build GRCh38
rs1010200901 Chr.1: 173867102-173867102 on Build GRCh38
rs190732670 Chr.1: 173866833-173866833 on Build GRCh38
rs895908480 Chr.1: 173867096-173867096 on Build GRCh38
rs764897254 Chr.1: 173866923-173866924 on Build GRCh38
rs527555345 Chr.1: 173866779-173866779 on Build GRCh38
rs924071662 Chr.1: 173867088-173867091 on Build GRCh38
rs897378416 Chr.1: 173867043-173867043 on Build GRCh38
rs749376235 Chr.1: 173867087-173867087 on Build GRCh38
rs535246393 Chr.1: 173867116-173867116 on Build GRCh38
rs566162011 Chr.1: 173867083-173867083 on Build GRCh38
rs17359906 Chr.1: 173867056-173867056 on Build GRCh38
rs1028763380 Chr.1: 173867175-173867175 on Build GRCh38
rs13476 Chr.1: 173866781-173866781 on Build GRCh38
rs530900154 Chr.1: 173867084-173867084 on Build GRCh38
rs192830163 Chr.1: 173866856-173866856 on Build GRCh38
rs987261794 Chr.1: 173866989-173866989 on Build GRCh38
rs761517103 Chr.1: 173866844-173866844 on Build GRCh38
rs745778323 Chr.1: 173867013-173867013 on Build GRCh38
rs747785188 Chr.1: 173866933-173866933 on Build GRCh38
rs1036083529 Chr.1: 173866825-173866825 on Build GRCh38
rs950585563 Chr.1: 173867044-173867044 on Build GRCh38
rs756876358 Chr.1: 173866885-173866889 on Build GRCh38
rs779694118 Chr.1: 173866984-173866986 on Build GRCh38
rs752927344 Chr.1: 173866889-173866889 on Build GRCh38
rs185310879 Chr.1: 173867064-173867064 on Build GRCh38
rs376398906 Chr.1: 173866971-173866971 on Build GRCh38
rs1027165049 Chr.1: 173866770-173866770 on Build GRCh38
rs773996807 Chr.1: 173866843-173866843 on Build GRCh38
rs1035751631 Chr.1: 173867095-173867095 on Build GRCh38
rs764392117 Chr.1: 173867183-173867183 on Build GRCh38
rs987724224 Chr.1: 173867204-173867204 on Build GRCh38
rs759569729 Chr.1: 173866949-173866949 on Build GRCh38
rs374895409 Chr.1: 173866905-173866905 on Build GRCh38
rs751410044 Chr.1: 173866941-173866941 on Build GRCh38
rs780995153 Chr.1: 173866983-173866983 on Build GRCh38
rs868483738 Chr.1: 173866830-173866830 on Build GRCh38
rs920455997 Chr.1: 173866817-173866817 on Build GRCh38
rs564940972 Chr.1: 173866778-173866778 on Build GRCh38
rs1020248783 Chr.1: 173867111-173867111 on Build GRCh38
rs367903781 Chr.1: 173866935-173866935 on Build GRCh38
rs768144068 Chr.1: 173866928-173866928 on Build GRCh38
rs542303124 Chr.1: 173866991-173866991 on Build GRCh38
rs758026343 Chr.1: 173866952-173866953 on Build GRCh38
rs371771507 Chr.1: 173866962-173866962 on Build GRCh38
rs771010903 Chr.1: 173866937-173866937 on Build GRCh38
rs12090169 Chr.1: 173867180-173867180 on Build GRCh38
rs898533529 Chr.1: 173867162-173867162 on Build GRCh38
rs763203322 Chr.1: 173866973-173866973 on Build GRCh38
rs775047821 Chr.1: 173866834-173866834 on Build GRCh38
rs144869587 Chr.1: 173867109-173867110 on Build GRCh38
rs911651098 Chr.1: 173867059-173867060 on Build GRCh38
rs756167832 Chr.1: 173867021-173867021 on Build GRCh38
rs973655011 Chr.1: 173866816-173866816 on Build GRCh38
rs530108354 Chr.1: 173866868-173866868 on Build GRCh38
rs753928397 Chr.1: 173866869-173866869 on Build GRCh38
rs757829227 Chr.1: 173866910-173866910 on Build GRCh38
rs992724254 Chr.1: 173866771-173866771 on Build GRCh38
rs767378584 Chr.1: 173866846-173866846 on Build GRCh38
rs1036455521 Chr.1: 173867020-173867020 on Build GRCh38
rs367688201 Chr.1: 173866837-173866837 on Build GRCh38
rs577380006 Chr.1: 173867069-173867069 on Build GRCh38
rs370135403 Chr.1: 173866900-173866900 on Build GRCh38
rs545139982 Chr.1: 173866772-173866772 on Build GRCh38
rs112727149 Chr.1: 173866800-173866800 on Build GRCh38
rs546941324 Chr.1: 173866802-173866805 on Build GRCh38
rs12086583 Chr.1: 173866923-173866923 on Build GRCh38
rs552701795 Chr.1: 173867074-173867074 on Build GRCh38
rs749936914 Chr.1: 173866947-173866948 on Build GRCh38
rs1045452532 Chr.1: 173867129-173867129 on Build GRCh38
rs773301995 Chr.1: 173866851-173866851 on Build GRCh38
rs372060980 Chr.1: 173866917-173866917 on Build GRCh38
rs748761612 Chr.1: 173866924-173866924 on Build GRCh38
rs532660612 Chr.1: 173866779-173866779 on Build GRCh38
rs767577315 Chr.1: 173866820-173866820 on Build GRCh38
rs994703957 Chr.1: 173867165-173867165 on Build GRCh38
rs760740571 Chr.1: 173866853-173866853 on Build GRCh38
rs978686555 Chr.1: 173867138-173867138 on Build GRCh38
rs758787436 Chr.1: 173866891-173866891 on Build GRCh38
rs896576172 Chr.1: 173867119-173867119 on Build GRCh38
rs1022852860 Chr.1: 173866975-173866975 on Build GRCh38
rs765385627 Chr.1: 173866876-173866876 on Build GRCh38
rs929037959 Chr.1: 173866828-173866828 on Build GRCh38
rs776568017 Chr.1: 173866948-173866948 on Build GRCh38
rs747578344 Chr.1: 173866825-173866828 on Build GRCh38
rs781715172 Chr.1: 173866922-173866922 on Build GRCh38
rs968607578 Chr.1: 173866953-173866953 on Build GRCh38
rs931728456 Chr.1: 173866982-173866982 on Build GRCh38
rs968596482 Chr.1: 173867118-173867118 on Build GRCh38
rs575778554 Chr.1: 173866768-173866768 on Build GRCh38
rs768250760 Chr.1: 173866842-173866842 on Build GRCh38
rs1012359313 Chr.1: 173867121-173867123 on Build GRCh38
rs1056538589 Chr.1: 173867117-173867117 on Build GRCh38
rs1056569257 Chr.1: 173866965-173866965 on Build GRCh38
rs758084813 Chr.1: 173866808-173866808 on Build GRCh38
rs777396072 Chr.1: 173866890-173866890 on Build GRCh38
rs1027440507 Chr.1: 173867022-173867022 on Build GRCh38

View Full Product Details