Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 178545436-178545938 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf220
Entrez Gene ID
400798
Gene Name
chromosome 1 open reading frame 220
Gene Aliases
-
UniGene
Hs.668085
Gene Chromosome Location
Chr.1: 178542796-178548889 on Build GRCh38
Transcripts
NR_033186.1

SNP Detail

SNP ID Chromosome Location
rs555372881 Chr.1: 178545569-178545569 on Build GRCh38
rs12568310 Chr.1: 178545487-178545487 on Build GRCh38
rs372398137 Chr.1: 178545640-178545640 on Build GRCh38
rs111247458 Chr.1: 178545832-178545832 on Build GRCh38
rs1048797491 Chr.1: 178545595-178545595 on Build GRCh38
rs760077452 Chr.1: 178545857-178545857 on Build GRCh38
rs1012575424 Chr.1: 178545761-178545761 on Build GRCh38
rs772218146 Chr.1: 178545830-178545830 on Build GRCh38
rs767512880 Chr.1: 178545524-178545524 on Build GRCh38
rs751572628 Chr.1: 178545663-178545663 on Build GRCh38
rs982258328 Chr.1: 178545521-178545521 on Build GRCh38
rs749359940 Chr.1: 178545904-178545904 on Build GRCh38
rs774038209 Chr.1: 178545511-178545511 on Build GRCh38
rs768132402 Chr.1: 178545489-178545489 on Build GRCh38
rs201774707 Chr.1: 178545617-178545617 on Build GRCh38
rs925355764 Chr.1: 178545486-178545486 on Build GRCh38
rs115905174 Chr.1: 178545691-178545691 on Build GRCh38
rs764219640 Chr.1: 178545575-178545575 on Build GRCh38
rs745373791 Chr.1: 178545783-178545783 on Build GRCh38
rs775358506 Chr.1: 178545708-178545708 on Build GRCh38
rs781370317 Chr.1: 178545479-178545479 on Build GRCh38
rs754983778 Chr.1: 178545912-178545912 on Build GRCh38
rs781728699 Chr.1: 178545587-178545587 on Build GRCh38
rs768537370 Chr.1: 178545806-178545806 on Build GRCh38
rs200961828 Chr.1: 178545606-178545606 on Build GRCh38
rs144816072 Chr.1: 178545881-178545881 on Build GRCh38
rs145009184 Chr.1: 178545673-178545673 on Build GRCh38
rs747979398 Chr.1: 178545833-178545834 on Build GRCh38
rs774332277 Chr.1: 178545820-178545820 on Build GRCh38
rs913170272 Chr.1: 178545513-178545513 on Build GRCh38
rs756021448 Chr.1: 178545787-178545787 on Build GRCh38
rs368806529 Chr.1: 178545599-178545599 on Build GRCh38
rs878883364 Chr.1: 178545908-178545908 on Build GRCh38
rs751630737 Chr.1: 178545656-178545656 on Build GRCh38
rs370818433 Chr.1: 178545916-178545916 on Build GRCh38
rs778065666 Chr.1: 178545464-178545464 on Build GRCh38
rs376810333 Chr.1: 178545660-178545660 on Build GRCh38
rs937032283 Chr.1: 178545866-178545866 on Build GRCh38
rs779424584 Chr.1: 178545652-178545653 on Build GRCh38
rs755574214 Chr.1: 178545887-178545887 on Build GRCh38
rs775198430 Chr.1: 178545642-178545642 on Build GRCh38
rs757860584 Chr.1: 178545478-178545478 on Build GRCh38
rs573927953 Chr.1: 178545705-178545705 on Build GRCh38
rs761979235 Chr.1: 178545732-178545732 on Build GRCh38
rs765563594 Chr.1: 178545870-178545870 on Build GRCh38
rs776225453 Chr.1: 178545545-178545545 on Build GRCh38
rs756909151 Chr.1: 178545763-178545763 on Build GRCh38
rs766507059 Chr.1: 178545849-178545849 on Build GRCh38
rs375770731 Chr.1: 178545535-178545535 on Build GRCh38
rs746529107 Chr.1: 178545626-178545626 on Build GRCh38
rs753083308 Chr.1: 178545871-178545871 on Build GRCh38
rs368877063 Chr.1: 178545639-178545639 on Build GRCh38
rs373571204 Chr.1: 178545618-178545618 on Build GRCh38
rs146427979 Chr.1: 178545602-178545602 on Build GRCh38
rs748349699 Chr.1: 178545826-178545826 on Build GRCh38
rs756625377 Chr.1: 178545665-178545665 on Build GRCh38
rs148275235 Chr.1: 178545692-178545692 on Build GRCh38
rs145498551 Chr.1: 178545707-178545707 on Build GRCh38
rs758734887 Chr.1: 178545572-178545572 on Build GRCh38
rs763226938 Chr.1: 178545649-178545649 on Build GRCh38
rs370181490 Chr.1: 178545889-178545889 on Build GRCh38
rs528072816 Chr.1: 178545517-178545517 on Build GRCh38
rs765001411 Chr.1: 178545567-178545567 on Build GRCh38
rs766337239 Chr.1: 178545538-178545538 on Build GRCh38
rs372738531 Chr.1: 178545474-178545474 on Build GRCh38
rs776618918 Chr.1: 178545638-178545638 on Build GRCh38
rs187076472 Chr.1: 178545520-178545520 on Build GRCh38
rs372936185 Chr.1: 178545773-178545773 on Build GRCh38
rs774708020 Chr.1: 178545731-178545731 on Build GRCh38
rs761969563 Chr.1: 178545657-178545657 on Build GRCh38
rs775370964 Chr.1: 178545462-178545462 on Build GRCh38
rs751720735 Chr.1: 178545581-178545581 on Build GRCh38
rs756866976 Chr.1: 178545876-178545876 on Build GRCh38
rs757770535 Chr.1: 178545586-178545586 on Build GRCh38
rs555260752 Chr.1: 178545744-178545744 on Build GRCh38
rs766804315 Chr.1: 178545743-178545743 on Build GRCh38
rs184208179 Chr.1: 178545715-178545715 on Build GRCh38
rs369861869 Chr.1: 178545661-178545661 on Build GRCh38
rs745796850 Chr.1: 178545699-178545699 on Build GRCh38
rs755325998 Chr.1: 178545754-178545754 on Build GRCh38
rs370618414 Chr.1: 178545740-178545740 on Build GRCh38
rs749304258 Chr.1: 178545801-178545801 on Build GRCh38
rs760685950 Chr.1: 178545833-178545833 on Build GRCh38
rs368783564 Chr.1: 178545760-178545760 on Build GRCh38
rs770719636 Chr.1: 178545634-178545634 on Build GRCh38
rs913096813 Chr.1: 178545809-178545809 on Build GRCh38
rs199947820 Chr.1: 178545752-178545752 on Build GRCh38
rs780124010 Chr.1: 178545799-178545799 on Build GRCh38
rs768629206 Chr.1: 178545729-178545729 on Build GRCh38
rs767089292 Chr.1: 178545877-178545877 on Build GRCh38
rs757929109 Chr.1: 178545526-178545529 on Build GRCh38
rs767608038 Chr.1: 178545734-178545734 on Build GRCh38
rs777119290 Chr.1: 178545856-178545856 on Build GRCh38
rs141267365 Chr.1: 178545562-178545562 on Build GRCh38
rs191954394 Chr.1: 178545616-178545616 on Build GRCh38
rs777420028 Chr.1: 178545688-178545688 on Build GRCh38
rs754849113 Chr.1: 178545700-178545701 on Build GRCh38
rs780705338 Chr.1: 178545774-178545774 on Build GRCh38
rs764415708 Chr.1: 178545652-178545652 on Build GRCh38
rs539095077 Chr.1: 178545674-178545674 on Build GRCh38
rs928161670 Chr.1: 178545852-178545852 on Build GRCh38
rs781236345 Chr.1: 178545760-178545760 on Build GRCh38
rs998788168 Chr.1: 178545728-178545728 on Build GRCh38
rs759389718 Chr.1: 178545560-178545560 on Build GRCh38
rs1047727663 Chr.1: 178545636-178545636 on Build GRCh38

View Full Product Details