Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 178545806-178546303 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf220
Entrez Gene ID
400798
Gene Name
chromosome 1 open reading frame 220
Gene Aliases
-
UniGene
Hs.668085
Gene Chromosome Location
Chr.1: 178542796-178548889 on Build GRCh38
Transcripts
NR_033186.1

SNP Detail

SNP ID Chromosome Location
rs375371216 Chr.1: 178545919-178545919 on Build GRCh38
rs570553556 Chr.1: 178546162-178546162 on Build GRCh38
rs111247458 Chr.1: 178545832-178545832 on Build GRCh38
rs1009687305 Chr.1: 178546079-178546079 on Build GRCh38
rs753083308 Chr.1: 178545871-178545871 on Build GRCh38
rs530775766 Chr.1: 178546147-178546147 on Build GRCh38
rs4650994 Chr.1: 178546177-178546177 on Build GRCh38
rs760077452 Chr.1: 178545857-178545857 on Build GRCh38
rs1051008966 Chr.1: 178546274-178546274 on Build GRCh38
rs772218146 Chr.1: 178545830-178545830 on Build GRCh38
rs368341542 Chr.1: 178545936-178545936 on Build GRCh38
rs1022649464 Chr.1: 178546075-178546075 on Build GRCh38
rs749359940 Chr.1: 178545904-178545904 on Build GRCh38
rs370181490 Chr.1: 178545889-178545889 on Build GRCh38
rs908912207 Chr.1: 178546222-178546222 on Build GRCh38
rs558845118 Chr.1: 178545921-178545921 on Build GRCh38
rs754983778 Chr.1: 178545912-178545912 on Build GRCh38
rs934087321 Chr.1: 178545967-178545967 on Build GRCh38
rs532560944 Chr.1: 178545988-178545988 on Build GRCh38
rs144816072 Chr.1: 178545881-178545881 on Build GRCh38
rs4650993 Chr.1: 178546052-178546052 on Build GRCh38
rs956678653 Chr.1: 178546061-178546061 on Build GRCh38
rs756866976 Chr.1: 178545876-178545876 on Build GRCh38
rs940354566 Chr.1: 178546231-178546231 on Build GRCh38
rs187452139 Chr.1: 178546055-178546055 on Build GRCh38
rs769657779 Chr.1: 178545921-178545921 on Build GRCh38
rs747979398 Chr.1: 178545833-178545834 on Build GRCh38
rs1012817688 Chr.1: 178546066-178546066 on Build GRCh38
rs878883364 Chr.1: 178545908-178545908 on Build GRCh38
rs548725742 Chr.1: 178545947-178545947 on Build GRCh38
rs569892199 Chr.1: 178546213-178546213 on Build GRCh38
rs922975132 Chr.1: 178546258-178546258 on Build GRCh38
rs370818433 Chr.1: 178545916-178545916 on Build GRCh38
rs760685950 Chr.1: 178545833-178545833 on Build GRCh38
rs568383512 Chr.1: 178546122-178546122 on Build GRCh38
rs962948451 Chr.1: 178546200-178546200 on Build GRCh38
rs767089292 Chr.1: 178545877-178545877 on Build GRCh38
rs937032283 Chr.1: 178545866-178545866 on Build GRCh38
rs933516693 Chr.1: 178546273-178546273 on Build GRCh38
rs1029249805 Chr.1: 178546011-178546011 on Build GRCh38
rs777119290 Chr.1: 178545856-178545856 on Build GRCh38
rs142048939 Chr.1: 178546023-178546023 on Build GRCh38
rs755574214 Chr.1: 178545887-178545887 on Build GRCh38
rs767272946 Chr.1: 178546158-178546158 on Build GRCh38
rs998090609 Chr.1: 178546001-178546001 on Build GRCh38
rs1036110622 Chr.1: 178546250-178546250 on Build GRCh38
rs4650992 Chr.1: 178546000-178546000 on Build GRCh38
rs928161670 Chr.1: 178545852-178545852 on Build GRCh38
rs765563594 Chr.1: 178545870-178545870 on Build GRCh38
rs556044787 Chr.1: 178546199-178546199 on Build GRCh38
rs757255431 Chr.1: 178546133-178546133 on Build GRCh38
rs766507059 Chr.1: 178545849-178545849 on Build GRCh38

View Full Product Details