Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 178548258-178548767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf220
Entrez Gene ID
400798
Gene Name
chromosome 1 open reading frame 220
Gene Aliases
-
UniGene
Hs.668085
Gene Chromosome Location
Chr.1: 178542796-178548889 on Build GRCh38
Transcripts
NR_033186.1

SNP Detail

SNP ID Chromosome Location
rs959126634 Chr.1: 178548743-178548743 on Build GRCh38
rs967803964 Chr.1: 178548711-178548711 on Build GRCh38
rs112634732 Chr.1: 178548373-178548373 on Build GRCh38
rs867007310 Chr.1: 178548544-178548544 on Build GRCh38
rs1008817733 Chr.1: 178548570-178548570 on Build GRCh38
rs531389996 Chr.1: 178548627-178548627 on Build GRCh38
rs577597918 Chr.1: 178548378-178548378 on Build GRCh38
rs537980378 Chr.1: 178548342-178548342 on Build GRCh38
rs964027524 Chr.1: 178548648-178548648 on Build GRCh38
rs888912887 Chr.1: 178548539-178548539 on Build GRCh38
rs946528457 Chr.1: 178548399-178548399 on Build GRCh38
rs968899070 Chr.1: 178548322-178548322 on Build GRCh38
rs1011707160 Chr.1: 178548649-178548649 on Build GRCh38
rs1021946121 Chr.1: 178548692-178548692 on Build GRCh38
rs565912101 Chr.1: 178548445-178548445 on Build GRCh38
rs755461434 Chr.1: 178548613-178548613 on Build GRCh38
rs898708385 Chr.1: 178548498-178548498 on Build GRCh38
rs1057495413 Chr.1: 178548379-178548379 on Build GRCh38
rs553799719 Chr.1: 178548442-178548442 on Build GRCh38
rs1026315283 Chr.1: 178548301-178548301 on Build GRCh38
rs1056509474 Chr.1: 178548446-178548446 on Build GRCh38
rs752174781 Chr.1: 178548662-178548662 on Build GRCh38
rs544344219 Chr.1: 178548656-178548656 on Build GRCh38
rs764832390 Chr.1: 178548642-178548642 on Build GRCh38
rs1038075902 Chr.1: 178548489-178548489 on Build GRCh38
rs915107333 Chr.1: 178548396-178548396 on Build GRCh38
rs542414381 Chr.1: 178548522-178548522 on Build GRCh38
rs978035756 Chr.1: 178548405-178548405 on Build GRCh38
rs1018373313 Chr.1: 178548338-178548338 on Build GRCh38
rs891346315 Chr.1: 178548618-178548618 on Build GRCh38
rs987174584 Chr.1: 178548323-178548323 on Build GRCh38
rs994706084 Chr.1: 178548573-178548573 on Build GRCh38
rs990215768 Chr.1: 178548389-178548389 on Build GRCh38
rs994415449 Chr.1: 178548283-178548283 on Build GRCh38
rs1021678687 Chr.1: 178548732-178548732 on Build GRCh38
rs759828980 Chr.1: 178548379-178548379 on Build GRCh38
rs955406212 Chr.1: 178548303-178548303 on Build GRCh38
rs117316015 Chr.1: 178548622-178548622 on Build GRCh38
rs4233175 Chr.1: 178548456-178548456 on Build GRCh38
rs967880986 Chr.1: 178548744-178548744 on Build GRCh38
rs927488643 Chr.1: 178548742-178548742 on Build GRCh38
rs878909109 Chr.1: 178548356-178548356 on Build GRCh38
rs372614165 Chr.1: 178548421-178548421 on Build GRCh38
rs1019276566 Chr.1: 178548599-178548599 on Build GRCh38
rs1036432404 Chr.1: 178548420-178548420 on Build GRCh38
rs1018715280 Chr.1: 178548634-178548634 on Build GRCh38
rs916226851 Chr.1: 178548455-178548455 on Build GRCh38
rs577312318 Chr.1: 178548677-178548677 on Build GRCh38
rs1006923436 Chr.1: 178548358-178548358 on Build GRCh38
rs940680946 Chr.1: 178548413-178548413 on Build GRCh38

View Full Product Details