Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 95784773-95785276 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTPD1
Entrez Gene ID
953
Gene Name
ectonucleoside triphosphate diphosphohydrolase 1
Gene Aliases
ATPDase, CD39, NTPDase-1, SPG64
UniGene
Hs.576612
Gene Chromosome Location
Chr.10: 95710901-95877266 on Build GRCh38
Transcripts
NM_001098175.1 NM_001164178.1 NM_001164179.1 NM_001164181.1 NM_001164182.1 NM_001164183.1 NM_001312654.1 NM_001320916.1 NM_001776.5 XM_011540370.2 XM_011540371.2 XM_011540372.2 XM_011540373.2 XM_011540374.2 XM_011540376.2 XM_011540377.2 XM_017016958.1 XM_017016959.1 XM_017016960.1 XM_017016961.1 XM_017016962.1 XM_017016963.1 XM_017016964.1 NR_038444.1

Gene Symbol
ENTPD1-AS1
Entrez Gene ID
728558
Gene Name
ENTPD1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.538374
Gene Chromosome Location
Chr.10: 95753206-96090235 on Build GRCh38
Transcripts
NM_001098175.1 NM_001164178.1 NM_001164179.1 NM_001164181.1 NM_001164182.1 NM_001164183.1 NM_001312654.1 NM_001320916.1 NM_001776.5 XM_011540370.2 XM_011540371.2 XM_011540372.2 XM_011540373.2 XM_011540374.2 XM_011540376.2 XM_011540377.2 XM_017016958.1 XM_017016959.1 XM_017016960.1 XM_017016961.1 XM_017016962.1 XM_017016963.1 XM_017016964.1 NR_038444.1

SNP Detail

SNP ID Chromosome Location
rs888735222 Chr.10: 95784803-95784803 on Build GRCh38
rs935884916 Chr.10: 95785142-95785142 on Build GRCh38
rs751271330 Chr.10: 95785029-95785029 on Build GRCh38
rs531286972 Chr.10: 95784833-95784833 on Build GRCh38
rs372105880 Chr.10: 95785168-95785168 on Build GRCh38
rs571381340 Chr.10: 95784873-95784873 on Build GRCh38
rs1050122858 Chr.10: 95785056-95785056 on Build GRCh38
rs918363055 Chr.10: 95785155-95785155 on Build GRCh38
rs867913980 Chr.10: 95784987-95784987 on Build GRCh38
rs536201260 Chr.10: 95785074-95785074 on Build GRCh38
rs956794687 Chr.10: 95784948-95784948 on Build GRCh38
rs940141300 Chr.10: 95784935-95784935 on Build GRCh38
rs187913293 Chr.10: 95784876-95784876 on Build GRCh38
rs888117361 Chr.10: 95785194-95785194 on Build GRCh38
rs1023850966 Chr.10: 95784835-95784835 on Build GRCh38
rs891628053 Chr.10: 95785213-95785213 on Build GRCh38
rs929722044 Chr.10: 95785160-95785160 on Build GRCh38
rs922598416 Chr.10: 95784957-95784957 on Build GRCh38
rs915236953 Chr.10: 95785009-95785009 on Build GRCh38
rs74679205 Chr.10: 95785072-95785072 on Build GRCh38
rs371262293 Chr.10: 95784834-95784834 on Build GRCh38
rs1033906314 Chr.10: 95784854-95784854 on Build GRCh38
rs146821982 Chr.10: 95785050-95785051 on Build GRCh38
rs1054319577 Chr.10: 95785163-95785163 on Build GRCh38
rs150915010 Chr.10: 95785235-95785235 on Build GRCh38
rs555658405 Chr.10: 95785201-95785201 on Build GRCh38
rs367999108 Chr.10: 95784814-95784814 on Build GRCh38
rs576233536 Chr.10: 95785198-95785198 on Build GRCh38
rs556778799 Chr.10: 95784951-95784951 on Build GRCh38
rs756899260 Chr.10: 95785217-95785217 on Build GRCh38
rs1040858083 Chr.10: 95785141-95785142 on Build GRCh38
rs1031521964 Chr.10: 95784938-95784938 on Build GRCh38
rs959548424 Chr.10: 95784869-95784869 on Build GRCh38
rs35360884 Chr.10: 95784796-95784796 on Build GRCh38
rs758169708 Chr.10: 95784846-95784846 on Build GRCh38
rs1011833844 Chr.10: 95785210-95785210 on Build GRCh38
rs780689389 Chr.10: 95785225-95785225 on Build GRCh38
rs567161490 Chr.10: 95785073-95785073 on Build GRCh38
rs370831302 Chr.10: 95784800-95784800 on Build GRCh38
rs943879736 Chr.10: 95785133-95785133 on Build GRCh38
rs972841102 Chr.10: 95784949-95784949 on Build GRCh38
rs911104598 Chr.10: 95785132-95785132 on Build GRCh38
rs541991946 Chr.10: 95785229-95785229 on Build GRCh38
rs984276749 Chr.10: 95784874-95784874 on Build GRCh38
rs1031155045 Chr.10: 95785236-95785236 on Build GRCh38

View Full Product Details