Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 117485356-117485900 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMX2OS
Entrez Gene ID
196047
Gene Name
EMX2 opposite strand/antisense RNA
Gene Aliases
EMX2-AS1, NCRNA00045
UniGene
Hs.312592
Gene Chromosome Location
Chr.10: 117484293-117545068 on Build GRCh38
Transcripts
NR_002791.2

SNP Detail

SNP ID Chromosome Location
rs180902557 Chr.10: 117485512-117485512 on Build GRCh38
rs866337905 Chr.10: 117485635-117485635 on Build GRCh38
rs575962735 Chr.10: 117485377-117485377 on Build GRCh38
rs897533200 Chr.10: 117485554-117485554 on Build GRCh38
rs117594999 Chr.10: 117485560-117485560 on Build GRCh38
rs186137792 Chr.10: 117485548-117485548 on Build GRCh38
rs1001106280 Chr.10: 117485694-117485694 on Build GRCh38
rs917711610 Chr.10: 117485468-117485468 on Build GRCh38
rs964054336 Chr.10: 117485434-117485434 on Build GRCh38
rs1034616335 Chr.10: 117485556-117485556 on Build GRCh38
rs891289258 Chr.10: 117485405-117485405 on Build GRCh38
rs371253073 Chr.10: 117485583-117485583 on Build GRCh38
rs751734189 Chr.10: 117485500-117485500 on Build GRCh38
rs34874330 Chr.10: 117485674-117485674 on Build GRCh38
rs973317119 Chr.10: 117485861-117485861 on Build GRCh38
rs192242128 Chr.10: 117485647-117485647 on Build GRCh38
rs980996911 Chr.10: 117485765-117485765 on Build GRCh38
rs936267701 Chr.10: 117485678-117485678 on Build GRCh38
rs374145954 Chr.10: 117485442-117485442 on Build GRCh38
rs12761166 Chr.10: 117485756-117485756 on Build GRCh38
rs979383114 Chr.10: 117485437-117485437 on Build GRCh38
rs386748058 Chr.10: 117485561-117485562 on Build GRCh38
rs543720404 Chr.10: 117485524-117485524 on Build GRCh38
rs767619935 Chr.10: 117485538-117485538 on Build GRCh38
rs748161637 Chr.10: 117485421-117485422 on Build GRCh38
rs369483354 Chr.10: 117485856-117485856 on Build GRCh38
rs1018468349 Chr.10: 117485660-117485660 on Build GRCh38
rs1000362671 Chr.10: 117485424-117485424 on Build GRCh38
rs941983279 Chr.10: 117485531-117485531 on Build GRCh38
rs762790879 Chr.10: 117485444-117485444 on Build GRCh38
rs2768325 Chr.10: 117485874-117485874 on Build GRCh38
rs956708736 Chr.10: 117485702-117485702 on Build GRCh38
rs558412980 Chr.10: 117485875-117485875 on Build GRCh38
rs576453174 Chr.10: 117485491-117485491 on Build GRCh38
rs570686591 Chr.10: 117485613-117485613 on Build GRCh38
rs992545638 Chr.10: 117485576-117485576 on Build GRCh38
rs114329417 Chr.10: 117485403-117485403 on Build GRCh38
rs770915994 Chr.10: 117485509-117485509 on Build GRCh38
rs1009407061 Chr.10: 117485448-117485448 on Build GRCh38
rs555758741 Chr.10: 117485791-117485791 on Build GRCh38
rs184033330 Chr.10: 117485788-117485788 on Build GRCh38
rs531423684 Chr.10: 117485622-117485622 on Build GRCh38
rs1025493545 Chr.10: 117485749-117485749 on Build GRCh38
rs988225674 Chr.10: 117485453-117485453 on Build GRCh38
rs945716252 Chr.10: 117485589-117485589 on Build GRCh38
rs371563943 Chr.10: 117485820-117485820 on Build GRCh38
rs79245240 Chr.10: 117485853-117485853 on Build GRCh38
rs897946776 Chr.10: 117485782-117485782 on Build GRCh38
rs1008443229 Chr.10: 117485418-117485418 on Build GRCh38
rs550876648 Chr.10: 117485582-117485582 on Build GRCh38
rs544593099 Chr.10: 117485834-117485834 on Build GRCh38
rs1018952899 Chr.10: 117485423-117485423 on Build GRCh38
rs1028093679 Chr.10: 117485789-117485789 on Build GRCh38
rs371233123 Chr.10: 117485382-117485382 on Build GRCh38
rs944731634 Chr.10: 117485744-117485744 on Build GRCh38
rs1032303377 Chr.10: 117485699-117485699 on Build GRCh38
rs1051516018 Chr.10: 117485572-117485572 on Build GRCh38
rs372827826 Chr.10: 117485463-117485463 on Build GRCh38
rs117169231 Chr.10: 117485383-117485383 on Build GRCh38
rs1009573606 Chr.10: 117485725-117485725 on Build GRCh38
rs749675445 Chr.10: 117485671-117485671 on Build GRCh38
rs112261993 Chr.10: 117485689-117485689 on Build GRCh38
rs369167391 Chr.10: 117485825-117485826 on Build GRCh38
rs540502793 Chr.10: 117485449-117485449 on Build GRCh38
rs889289386 Chr.10: 117485698-117485698 on Build GRCh38
rs780972658 Chr.10: 117485420-117485420 on Build GRCh38
rs764239121 Chr.10: 117485490-117485490 on Build GRCh38

View Full Product Details