Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 1683533-1684037 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM99B
Entrez Gene ID
100132464
Gene Name
family with sequence similarity 99 member B (non-protein coding)
Gene Aliases
-
UniGene
Hs.117797
Gene Chromosome Location
Chr.11: 1683270-1685629 on Build GRCh38
Transcripts
NR_026642.1

SNP Detail

SNP ID Chromosome Location
rs1009649872 Chr.11: 1683781-1683781 on Build GRCh38
rs957027928 Chr.11: 1683772-1683772 on Build GRCh38
rs995997521 Chr.11: 1683697-1683697 on Build GRCh38
rs891920233 Chr.11: 1683634-1683634 on Build GRCh38
rs1042921320 Chr.11: 1683636-1683636 on Build GRCh38
rs955216078 Chr.11: 1683818-1683818 on Build GRCh38
rs900322192 Chr.11: 1683663-1683663 on Build GRCh38
rs1011667900 Chr.11: 1683635-1683635 on Build GRCh38
rs1053074803 Chr.11: 1683626-1683626 on Build GRCh38
rs920085615 Chr.11: 1683570-1683570 on Build GRCh38
rs564878971 Chr.11: 1683850-1683850 on Build GRCh38
rs896650269 Chr.11: 1683586-1683586 on Build GRCh38
rs919600914 Chr.11: 1683861-1683861 on Build GRCh38
rs929750767 Chr.11: 1683871-1683871 on Build GRCh38
rs552750983 Chr.11: 1683862-1683862 on Build GRCh38
rs374855580 Chr.11: 1683561-1683561 on Build GRCh38
rs986557486 Chr.11: 1683821-1683821 on Build GRCh38
rs1032330197 Chr.11: 1683770-1683770 on Build GRCh38
rs911150705 Chr.11: 1683841-1683841 on Build GRCh38
rs928881030 Chr.11: 1683604-1683604 on Build GRCh38
rs530980220 Chr.11: 1683834-1683834 on Build GRCh38
rs1047301503 Chr.11: 1683569-1683569 on Build GRCh38
rs974302171 Chr.11: 1683563-1683563 on Build GRCh38
rs574924721 Chr.11: 1683599-1683599 on Build GRCh38
rs564032799 Chr.11: 1683713-1683713 on Build GRCh38
rs142951263 Chr.11: 1683607-1683607 on Build GRCh38
rs559778810 Chr.11: 1683591-1683591 on Build GRCh38
rs1045346348 Chr.11: 1683605-1683605 on Build GRCh38
rs541387682 Chr.11: 1683588-1683588 on Build GRCh38
rs1020108030 Chr.11: 1683794-1683794 on Build GRCh38
rs949021529 Chr.11: 1683581-1683581 on Build GRCh38
rs1001284559 Chr.11: 1683727-1683727 on Build GRCh38
rs1046856968 Chr.11: 1683923-1683923 on Build GRCh38
rs963982167 Chr.11: 1683843-1683843 on Build GRCh38
rs929724228 Chr.11: 1683564-1683564 on Build GRCh38

View Full Product Details