Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 31816468-31816979 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DKFZp686K1684
Entrez Gene ID
440034
Gene Name
uncharacterized LOC440034
Gene Aliases
-
UniGene
Hs.523576
Gene Chromosome Location
Chr.11: 31816566-31887041 on Build GRCh38
Transcripts
NR_033971.1 NM_001127612.1 NM_001258462.1

Gene Symbol
PAX6
Entrez Gene ID
5080
Gene Name
paired box 6
Gene Aliases
AN, AN2, D11S812E, FVH1, MGDA, WAGR
UniGene
Hs.270303
Gene Chromosome Location
Chr.11: 31784792-31817961 on Build GRCh38
Transcripts
NR_033971.1 NM_001127612.1 NM_001258462.1

SNP Detail

SNP ID Chromosome Location
rs766437520 Chr.11: 31816653-31816653 on Build GRCh38
rs984115644 Chr.11: 31816646-31816646 on Build GRCh38
rs5790869 Chr.11: 31816704-31816705 on Build GRCh38
rs1033409306 Chr.11: 31816866-31816866 on Build GRCh38
rs750659447 Chr.11: 31816644-31816644 on Build GRCh38
rs1011235264 Chr.11: 31816757-31816757 on Build GRCh38
rs561070862 Chr.11: 31816546-31816546 on Build GRCh38
rs952771377 Chr.11: 31816821-31816821 on Build GRCh38
rs888618273 Chr.11: 31816574-31816574 on Build GRCh38
rs976644042 Chr.11: 31816902-31816902 on Build GRCh38
rs751635981 Chr.11: 31816671-31816671 on Build GRCh38
rs1806184 Chr.11: 31816613-31816613 on Build GRCh38
rs947102121 Chr.11: 31816903-31816903 on Build GRCh38
rs531529786 Chr.11: 31816568-31816568 on Build GRCh38
rs967598424 Chr.11: 31816607-31816607 on Build GRCh38
rs972370801 Chr.11: 31816856-31816856 on Build GRCh38
rs911034479 Chr.11: 31816666-31816666 on Build GRCh38
rs577838390 Chr.11: 31816793-31816793 on Build GRCh38
rs761069603 Chr.11: 31816941-31816941 on Build GRCh38
rs900842429 Chr.11: 31816499-31816499 on Build GRCh38
rs989353388 Chr.11: 31816652-31816652 on Build GRCh38
rs929609011 Chr.11: 31816773-31816773 on Build GRCh38
rs964029798 Chr.11: 31816673-31816674 on Build GRCh38
rs998832805 Chr.11: 31816862-31816862 on Build GRCh38
rs1036602484 Chr.11: 31816511-31816511 on Build GRCh38
rs960507828 Chr.11: 31816816-31816816 on Build GRCh38
rs923071665 Chr.11: 31816706-31816706 on Build GRCh38
rs542699892 Chr.11: 31816879-31816879 on Build GRCh38
rs770835374 Chr.11: 31816635-31816635 on Build GRCh38
rs748484811 Chr.11: 31816791-31816791 on Build GRCh38
rs1036208745 Chr.11: 31816697-31816697 on Build GRCh38
rs1018148759 Chr.11: 31816895-31816895 on Build GRCh38
rs538701586 Chr.11: 31816594-31816594 on Build GRCh38
rs556015444 Chr.11: 31816649-31816649 on Build GRCh38
rs903452500 Chr.11: 31816790-31816790 on Build GRCh38
rs561336836 Chr.11: 31816885-31816885 on Build GRCh38
rs187389456 Chr.11: 31816678-31816678 on Build GRCh38
rs1014501038 Chr.11: 31816621-31816621 on Build GRCh38
rs749117053 Chr.11: 31816554-31816554 on Build GRCh38
rs908510610 Chr.11: 31816842-31816842 on Build GRCh38
rs752634681 Chr.11: 31816506-31816506 on Build GRCh38
rs922492488 Chr.11: 31816909-31816909 on Build GRCh38
rs560435847 Chr.11: 31816623-31816623 on Build GRCh38
rs74750543 Chr.11: 31816591-31816591 on Build GRCh38
rs531593169 Chr.11: 31816919-31816919 on Build GRCh38
rs778711468 Chr.11: 31816571-31816571 on Build GRCh38
rs547142440 Chr.11: 31816724-31816724 on Build GRCh38
rs933105941 Chr.11: 31816711-31816711 on Build GRCh38
rs1042716113 Chr.11: 31816930-31816930 on Build GRCh38
rs868381090 Chr.11: 31816935-31816935 on Build GRCh38
rs574206197 Chr.11: 31816657-31816657 on Build GRCh38
rs770808568 Chr.11: 31816570-31816570 on Build GRCh38
rs495902 Chr.11: 31816633-31816633 on Build GRCh38
rs1042662334 Chr.11: 31816769-31816769 on Build GRCh38
rs1050627215 Chr.11: 31816732-31816732 on Build GRCh38
rs571697344 Chr.11: 31816818-31816818 on Build GRCh38
rs1046758086 Chr.11: 31816549-31816549 on Build GRCh38
rs556305517 Chr.11: 31816736-31816736 on Build GRCh38
rs1046809673 Chr.11: 31816782-31816782 on Build GRCh38
rs888854884 Chr.11: 31816740-31816740 on Build GRCh38
rs1021865010 Chr.11: 31816605-31816605 on Build GRCh38
rs922276680 Chr.11: 31816750-31816750 on Build GRCh38
rs773832420 Chr.11: 31816832-31816832 on Build GRCh38
rs910304272 Chr.11: 31816901-31816901 on Build GRCh38
rs1050267319 Chr.11: 31816887-31816887 on Build GRCh38
rs903198385 Chr.11: 31816861-31816861 on Build GRCh38
rs61217094 Chr.11: 31816817-31816817 on Build GRCh38
rs777759794 Chr.11: 31816548-31816548 on Build GRCh38
rs961970401 Chr.11: 31816853-31816853 on Build GRCh38
rs4440995 Chr.11: 31816580-31816580 on Build GRCh38
rs952719237 Chr.11: 31816645-31816645 on Build GRCh38
rs398015701 Chr.11: 31816705-31816706 on Build GRCh38
rs901597381 Chr.11: 31816512-31816512 on Build GRCh38
rs572397522 Chr.11: 31816876-31816876 on Build GRCh38
rs769188360 Chr.11: 31816619-31816619 on Build GRCh38

View Full Product Details