Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 31886411-31886956 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DKFZp686K1684
Entrez Gene ID
440034
Gene Name
uncharacterized LOC440034
Gene Aliases
-
UniGene
Hs.523576
Gene Chromosome Location
Chr.11: 31816566-31887041 on Build GRCh38
Transcripts
NR_033971.1

SNP Detail

SNP ID Chromosome Location
rs905103426 Chr.11: 31886464-31886464 on Build GRCh38
rs967123962 Chr.11: 31886494-31886494 on Build GRCh38
rs903008363 Chr.11: 31886909-31886909 on Build GRCh38
rs538173779 Chr.11: 31886522-31886522 on Build GRCh38
rs1016247194 Chr.11: 31886706-31886706 on Build GRCh38
rs940108329 Chr.11: 31886551-31886551 on Build GRCh38
rs974472764 Chr.11: 31886718-31886718 on Build GRCh38
rs1056758180 Chr.11: 31886636-31886636 on Build GRCh38
rs996516825 Chr.11: 31886692-31886692 on Build GRCh38
rs555852814 Chr.11: 31886886-31886886 on Build GRCh38
rs750249193 Chr.11: 31886584-31886584 on Build GRCh38
rs992115329 Chr.11: 31886789-31886789 on Build GRCh38
rs931476407 Chr.11: 31886530-31886530 on Build GRCh38
rs1043519909 Chr.11: 31886679-31886679 on Build GRCh38
rs1028563881 Chr.11: 31886546-31886546 on Build GRCh38
rs746792527 Chr.11: 31886901-31886901 on Build GRCh38
rs35394188 Chr.11: 31886922-31886923 on Build GRCh38
rs560981784 Chr.11: 31886703-31886703 on Build GRCh38
rs1021809986 Chr.11: 31886485-31886485 on Build GRCh38
rs960317104 Chr.11: 31886518-31886518 on Build GRCh38
rs541068250 Chr.11: 31886471-31886471 on Build GRCh38
rs932420656 Chr.11: 31886900-31886900 on Build GRCh38
rs1028449193 Chr.11: 31886699-31886706 on Build GRCh38
rs531194553 Chr.11: 31886705-31886705 on Build GRCh38
rs1049719136 Chr.11: 31886885-31886885 on Build GRCh38
rs1050306227 Chr.11: 31886590-31886590 on Build GRCh38
rs563138542 Chr.11: 31886894-31886894 on Build GRCh38
rs980609458 Chr.11: 31886612-31886612 on Build GRCh38
rs553460443 Chr.11: 31886601-31886601 on Build GRCh38
rs529432943 Chr.11: 31886588-31886588 on Build GRCh38
rs970134142 Chr.11: 31886739-31886739 on Build GRCh38
rs960517154 Chr.11: 31886786-31886786 on Build GRCh38
rs1026952890 Chr.11: 31886737-31886737 on Build GRCh38
rs1022452361 Chr.11: 31886528-31886528 on Build GRCh38
rs763207838 Chr.11: 31886457-31886457 on Build GRCh38
rs892162423 Chr.11: 31886640-31886640 on Build GRCh38
rs901171187 Chr.11: 31886572-31886572 on Build GRCh38
rs2440238 Chr.11: 31886440-31886440 on Build GRCh38
rs147225074 Chr.11: 31886442-31886442 on Build GRCh38
rs1021444543 Chr.11: 31886595-31886595 on Build GRCh38
rs567542114 Chr.11: 31886514-31886514 on Build GRCh38
rs1035389430 Chr.11: 31886647-31886647 on Build GRCh38
rs940151042 Chr.11: 31886776-31886776 on Build GRCh38
rs572062613 Chr.11: 31886618-31886618 on Build GRCh38
rs1011836359 Chr.11: 31886594-31886594 on Build GRCh38
rs1037320972 Chr.11: 31886923-31886923 on Build GRCh38
rs577438905 Chr.11: 31886548-31886548 on Build GRCh38
rs932671674 Chr.11: 31886824-31886824 on Build GRCh38
rs891322733 Chr.11: 31886916-31886916 on Build GRCh38
rs959702299 Chr.11: 31886653-31886653 on Build GRCh38
rs991115324 Chr.11: 31886678-31886678 on Build GRCh38
rs555777923 Chr.11: 31886489-31886489 on Build GRCh38
rs913808504 Chr.11: 31886822-31886822 on Build GRCh38
rs976810971 Chr.11: 31886782-31886782 on Build GRCh38
rs370884127 Chr.11: 31886804-31886804 on Build GRCh38
rs140587985 Chr.11: 31886535-31886535 on Build GRCh38
rs188837926 Chr.11: 31886920-31886920 on Build GRCh38
rs888952327 Chr.11: 31886451-31886451 on Build GRCh38
rs916132208 Chr.11: 31886526-31886526 on Build GRCh38
rs945093953 Chr.11: 31886834-31886834 on Build GRCh38
rs753773434 Chr.11: 31886684-31886684 on Build GRCh38
rs544770953 Chr.11: 31886583-31886583 on Build GRCh38
rs764395366 Chr.11: 31886495-31886495 on Build GRCh38
rs542357841 Chr.11: 31886701-31886701 on Build GRCh38
rs547550411 Chr.11: 31886592-31886592 on Build GRCh38
rs61879845 Chr.11: 31886740-31886740 on Build GRCh38
rs939269498 Chr.11: 31886630-31886630 on Build GRCh38
rs1043789754 Chr.11: 31886836-31886836 on Build GRCh38
rs974213945 Chr.11: 31886591-31886591 on Build GRCh38
rs984726105 Chr.11: 31886712-31886712 on Build GRCh38
rs926406544 Chr.11: 31886614-31886614 on Build GRCh38
rs1056904916 Chr.11: 31886922-31886922 on Build GRCh38
rs1015613191 Chr.11: 31886576-31886576 on Build GRCh38
rs996965995 Chr.11: 31886539-31886539 on Build GRCh38
rs998602226 Chr.11: 31886641-31886641 on Build GRCh38
rs926458115 Chr.11: 31886742-31886742 on Build GRCh38
rs902955003 Chr.11: 31886611-31886611 on Build GRCh38
rs1041050627 Chr.11: 31886567-31886567 on Build GRCh38
rs922647512 Chr.11: 31886821-31886821 on Build GRCh38

View Full Product Details