Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 45772215-45772716 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DKFZp779M0652
Entrez Gene ID
374387
Gene Name
uncharacterized DKFZp779M0652
Gene Aliases
-
UniGene
Hs.632142
Gene Chromosome Location
Chr.11: 45771432-45772358 on Build GRCh38
Transcripts
NR_027134.1

SNP Detail

SNP ID Chromosome Location
rs944605102 Chr.11: 45772470-45772470 on Build GRCh38
rs996358920 Chr.11: 45772607-45772607 on Build GRCh38
rs903632036 Chr.11: 45772262-45772262 on Build GRCh38
rs562458686 Chr.11: 45772245-45772245 on Build GRCh38
rs1004556128 Chr.11: 45772672-45772672 on Build GRCh38
rs1048857294 Chr.11: 45772608-45772608 on Build GRCh38
rs1052952196 Chr.11: 45772276-45772276 on Build GRCh38
rs925982757 Chr.11: 45772537-45772537 on Build GRCh38
rs962238467 Chr.11: 45772392-45772392 on Build GRCh38
rs903585497 Chr.11: 45772601-45772601 on Build GRCh38
rs529942375 Chr.11: 45772409-45772409 on Build GRCh38
rs958930403 Chr.11: 45772297-45772297 on Build GRCh38
rs140844109 Chr.11: 45772584-45772584 on Build GRCh38
rs558602446 Chr.11: 45772286-45772286 on Build GRCh38
rs544814205 Chr.11: 45772273-45772273 on Build GRCh38
rs980190792 Chr.11: 45772533-45772533 on Build GRCh38
rs920687412 Chr.11: 45772397-45772397 on Build GRCh38
rs76728937 Chr.11: 45772394-45772394 on Build GRCh38
rs948854643 Chr.11: 45772498-45772498 on Build GRCh38
rs900284196 Chr.11: 45772371-45772371 on Build GRCh38
rs887504327 Chr.11: 45772636-45772636 on Build GRCh38
rs137889556 Chr.11: 45772293-45772293 on Build GRCh38
rs759720717 Chr.11: 45772408-45772408 on Build GRCh38
rs935942269 Chr.11: 45772270-45772270 on Build GRCh38
rs115131816 Chr.11: 45772257-45772257 on Build GRCh38
rs867654221 Chr.11: 45772614-45772614 on Build GRCh38
rs570077473 Chr.11: 45772400-45772400 on Build GRCh38
rs900316538 Chr.11: 45772592-45772592 on Build GRCh38
rs543994559 Chr.11: 45772310-45772310 on Build GRCh38
rs1024870526 Chr.11: 45772391-45772391 on Build GRCh38
rs867837198 Chr.11: 45772492-45772492 on Build GRCh38
rs937772296 Chr.11: 45772616-45772616 on Build GRCh38
rs144604183 Chr.11: 45772686-45772686 on Build GRCh38
rs569368751 Chr.11: 45772620-45772620 on Build GRCh38
rs1043990203 Chr.11: 45772587-45772587 on Build GRCh38
rs1054765958 Chr.11: 45772648-45772648 on Build GRCh38
rs184329504 Chr.11: 45772444-45772444 on Build GRCh38
rs936015782 Chr.11: 45772539-45772539 on Build GRCh38
rs188220372 Chr.11: 45772445-45772445 on Build GRCh38
rs917415094 Chr.11: 45772494-45772494 on Build GRCh38
rs993410625 Chr.11: 45772479-45772479 on Build GRCh38
rs1040651690 Chr.11: 45772586-45772586 on Build GRCh38
rs11038647 Chr.11: 45772585-45772585 on Build GRCh38
rs1050676610 Chr.11: 45772439-45772439 on Build GRCh38
rs61884462 Chr.11: 45772581-45772581 on Build GRCh38
rs983088519 Chr.11: 45772435-45772435 on Build GRCh38
rs961614197 Chr.11: 45772454-45772454 on Build GRCh38

View Full Product Details