Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61757579-61758082 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DKFZP434K028
Entrez Gene ID
26070
Gene Name
uncharacterized LOC26070
Gene Aliases
-
UniGene
Hs.564254
Gene Chromosome Location
Chr.11: 61754029-61757664 on Build GRCh38
Transcripts
NR_026882.1 NM_001127392.2 NM_013279.3 XM_005274222.1 XM_005274223.1 XM_005274224.1 XM_005274225.1 XM_005274226.1 XM_005274227.1 XM_005274228.1 XM_011545234.2

Gene Symbol
MYRF
Entrez Gene ID
745
Gene Name
myelin regulatory factor
Gene Aliases
C11orf9, MRF, Ndt80, pqn-47
UniGene
Hs.473109
Gene Chromosome Location
Chr.11: 61752617-61788518 on Build GRCh38
Transcripts
NR_026882.1 NM_001127392.2 NM_013279.3 XM_005274222.1 XM_005274223.1 XM_005274224.1 XM_005274225.1 XM_005274226.1 XM_005274227.1 XM_005274228.1 XM_011545234.2

SNP Detail

SNP ID Chromosome Location
rs915908106 Chr.11: 61757815-61757815 on Build GRCh38
rs1023250498 Chr.11: 61758031-61758031 on Build GRCh38
rs889065096 Chr.11: 61757713-61757713 on Build GRCh38
rs1035877722 Chr.11: 61757732-61757732 on Build GRCh38
rs982630781 Chr.11: 61757835-61757835 on Build GRCh38
rs1043187814 Chr.11: 61757624-61757624 on Build GRCh38
rs896031760 Chr.11: 61757991-61757991 on Build GRCh38
rs951272133 Chr.11: 61757833-61757833 on Build GRCh38
rs866972392 Chr.11: 61757780-61757780 on Build GRCh38
rs557380012 Chr.11: 61757680-61757680 on Build GRCh38
rs1057236622 Chr.11: 61757989-61757989 on Build GRCh38
rs912455410 Chr.11: 61757888-61757888 on Build GRCh38
rs965836387 Chr.11: 61757764-61757764 on Build GRCh38
rs554632760 Chr.11: 61758024-61758024 on Build GRCh38
rs903344554 Chr.11: 61757688-61757688 on Build GRCh38
rs567571087 Chr.11: 61757666-61757666 on Build GRCh38
rs1010990380 Chr.11: 61757627-61757627 on Build GRCh38
rs971841579 Chr.11: 61757773-61757773 on Build GRCh38
rs1013170998 Chr.11: 61757779-61757779 on Build GRCh38
rs1043524953 Chr.11: 61757965-61757965 on Build GRCh38
rs904843821 Chr.11: 61758036-61758036 on Build GRCh38
rs1043823069 Chr.11: 61757996-61757996 on Build GRCh38
rs377576348 Chr.11: 61757683-61757683 on Build GRCh38
rs934829798 Chr.11: 61757953-61757953 on Build GRCh38
rs975374076 Chr.11: 61757915-61757915 on Build GRCh38
rs527260618 Chr.11: 61757601-61757601 on Build GRCh38
rs952125556 Chr.11: 61757808-61757808 on Build GRCh38
rs374163560 Chr.11: 61757966-61757966 on Build GRCh38
rs999000498 Chr.11: 61757698-61757698 on Build GRCh38
rs562605356 Chr.11: 61757602-61757602 on Build GRCh38
rs35290001 Chr.11: 61757950-61757950 on Build GRCh38
rs919716175 Chr.11: 61757832-61757832 on Build GRCh38
rs928818122 Chr.11: 61757839-61757839 on Build GRCh38
rs570927327 Chr.11: 61757614-61757614 on Build GRCh38
rs969119478 Chr.11: 61757788-61757788 on Build GRCh38
rs538791081 Chr.11: 61757921-61757921 on Build GRCh38
rs934799487 Chr.11: 61757978-61757978 on Build GRCh38
rs921262862 Chr.11: 61757916-61757916 on Build GRCh38
rs754871461 Chr.11: 61757618-61757618 on Build GRCh38
rs879636785 Chr.11: 61757652-61757652 on Build GRCh38
rs572136688 Chr.11: 61757976-61757976 on Build GRCh38
rs201312715 Chr.11: 61757692-61757692 on Build GRCh38
rs35607948 Chr.11: 61757686-61757686 on Build GRCh38
rs995207362 Chr.11: 61758047-61758047 on Build GRCh38
rs757792091 Chr.11: 61757599-61757599 on Build GRCh38
rs868152845 Chr.11: 61757846-61757846 on Build GRCh38
rs148069479 Chr.11: 61757985-61757985 on Build GRCh38
rs943944776 Chr.11: 61757902-61757902 on Build GRCh38
rs892217146 Chr.11: 61757990-61757990 on Build GRCh38
rs143440891 Chr.11: 61757809-61757809 on Build GRCh38
rs751536165 Chr.11: 61757934-61757934 on Build GRCh38
rs533691050 Chr.11: 61757651-61757651 on Build GRCh38
rs192071320 Chr.11: 61757745-61757745 on Build GRCh38
rs569749734 Chr.11: 61757824-61757824 on Build GRCh38

View Full Product Details