Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 74720111-74720629 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRDL2
Entrez Gene ID
25884
Gene Name
chordin like 2
Gene Aliases
BNF1, CHL2
UniGene
Hs.432379
Gene Chromosome Location
Chr.11: 74696428-74731385 on Build GRCh38
Transcripts
NM_001278473.2 NM_015424.5 NR_039845.1

Gene Symbol
MIR4696
Entrez Gene ID
100616402
Gene Name
microRNA 4696
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.11: 74720268-74720337 on Build GRCh38
Transcripts
NM_001278473.2 NM_015424.5 NR_039845.1

SNP Detail

SNP ID Chromosome Location
rs1035407604 Chr.11: 74720538-74720538 on Build GRCh38
rs1038491046 Chr.11: 74720523-74720523 on Build GRCh38
rs908810705 Chr.11: 74720317-74720317 on Build GRCh38
rs187355658 Chr.11: 74720537-74720537 on Build GRCh38
rs1014061089 Chr.11: 74720165-74720165 on Build GRCh38
rs932721871 Chr.11: 74720371-74720371 on Build GRCh38
rs992178965 Chr.11: 74720479-74720479 on Build GRCh38
rs190821107 Chr.11: 74720271-74720271 on Build GRCh38
rs367650769 Chr.11: 74720605-74720607 on Build GRCh38
rs561323185 Chr.11: 74720437-74720437 on Build GRCh38
rs751943726 Chr.11: 74720562-74720562 on Build GRCh38
rs974631042 Chr.11: 74720321-74720321 on Build GRCh38
rs760527381 Chr.11: 74720191-74720191 on Build GRCh38
rs1016720489 Chr.11: 74720281-74720281 on Build GRCh38
rs1021194549 Chr.11: 74720575-74720575 on Build GRCh38
rs886933506 Chr.11: 74720519-74720519 on Build GRCh38
rs955636028 Chr.11: 74720388-74720388 on Build GRCh38
rs1051222566 Chr.11: 74720397-74720397 on Build GRCh38
rs532993335 Chr.11: 74720524-74720524 on Build GRCh38
rs775237150 Chr.11: 74720176-74720176 on Build GRCh38
rs1011837983 Chr.11: 74720150-74720150 on Build GRCh38
rs541505753 Chr.11: 74720320-74720320 on Build GRCh38
rs1015904969 Chr.11: 74720584-74720584 on Build GRCh38
rs886966003 Chr.11: 74720233-74720233 on Build GRCh38
rs529540496 Chr.11: 74720275-74720275 on Build GRCh38
rs982959835 Chr.11: 74720310-74720310 on Build GRCh38
rs551362659 Chr.11: 74720548-74720548 on Build GRCh38
rs753552922 Chr.11: 74720255-74720255 on Build GRCh38
rs921473978 Chr.11: 74720346-74720346 on Build GRCh38
rs1035315352 Chr.11: 74720293-74720293 on Build GRCh38
rs11236230 Chr.11: 74720522-74720522 on Build GRCh38
rs764881598 Chr.11: 74720422-74720422 on Build GRCh38
rs895736251 Chr.11: 74720158-74720158 on Build GRCh38
rs904939643 Chr.11: 74720513-74720513 on Build GRCh38
rs1052996833 Chr.11: 74720424-74720424 on Build GRCh38
rs61239552 Chr.11: 74720508-74720510 on Build GRCh38
rs970198177 Chr.11: 74720257-74720257 on Build GRCh38
rs71977354 Chr.11: 74720503-74720505 on Build GRCh38
rs527613046 Chr.11: 74720491-74720491 on Build GRCh38
rs937969711 Chr.11: 74720475-74720475 on Build GRCh38
rs549940798 Chr.11: 74720534-74720534 on Build GRCh38
rs550875249 Chr.11: 74720306-74720306 on Build GRCh38
rs10581351 Chr.11: 74720603-74720605 on Build GRCh38
rs4944943 Chr.11: 74720232-74720232 on Build GRCh38
rs566786511 Chr.11: 74720181-74720181 on Build GRCh38
rs949931445 Chr.11: 74720507-74720508 on Build GRCh38
rs1056112782 Chr.11: 74720138-74720138 on Build GRCh38
rs926767294 Chr.11: 74720438-74720438 on Build GRCh38
rs968213945 Chr.11: 74720411-74720411 on Build GRCh38
rs780720522 Chr.11: 74720560-74720560 on Build GRCh38
rs890814032 Chr.11: 74720553-74720554 on Build GRCh38
rs891092812 Chr.11: 74720398-74720398 on Build GRCh38
rs1029836005 Chr.11: 74720356-74720356 on Build GRCh38
rs766033393 Chr.11: 74720602-74720603 on Build GRCh38
rs11236229 Chr.11: 74720507-74720507 on Build GRCh38

View Full Product Details