Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 111417898-111418397 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTG4
Entrez Gene ID
54766
Gene Name
BTG anti-proliferation factor 4
Gene Aliases
APRO3, PC3B
UniGene
Hs.128180
Gene Chromosome Location
Chr.11: 111383828-111512388 on Build GRCh38
Transcripts
XM_011542876.2 XM_011542878.2 NR_046085.1

Gene Symbol
LOC100132078
Entrez Gene ID
100132078
Gene Name
uncharacterized LOC100132078
Gene Aliases
-
UniGene
Hs.672662
Gene Chromosome Location
Chr.11: 111414242-111418186 on Build GRCh38
Transcripts
XM_011542876.2 XM_011542878.2 NR_046085.1

SNP Detail

SNP ID Chromosome Location
rs923148920 Chr.11: 111418039-111418039 on Build GRCh38
rs1054805050 Chr.11: 111418290-111418290 on Build GRCh38
rs964547967 Chr.11: 111418198-111418198 on Build GRCh38
rs1054774399 Chr.11: 111418096-111418096 on Build GRCh38
rs753530453 Chr.11: 111418335-111418335 on Build GRCh38
rs1040680696 Chr.11: 111418084-111418084 on Build GRCh38
rs553843185 Chr.11: 111418140-111418140 on Build GRCh38
rs1018329949 Chr.11: 111418197-111418197 on Build GRCh38
rs985558905 Chr.11: 111418195-111418195 on Build GRCh38
rs570946005 Chr.11: 111418268-111418268 on Build GRCh38
rs191389141 Chr.11: 111418289-111418289 on Build GRCh38
rs757023492 Chr.11: 111417950-111417952 on Build GRCh38
rs538127329 Chr.11: 111418035-111418035 on Build GRCh38
rs930209513 Chr.11: 111418091-111418091 on Build GRCh38
rs554640958 Chr.11: 111418171-111418171 on Build GRCh38
rs902665454 Chr.11: 111418362-111418362 on Build GRCh38
rs1005998939 Chr.11: 111418073-111418073 on Build GRCh38
rs752958506 Chr.11: 111418008-111418008 on Build GRCh38
rs956711286 Chr.11: 111418196-111418196 on Build GRCh38
rs949132597 Chr.11: 111418320-111418320 on Build GRCh38
rs887423142 Chr.11: 111418068-111418068 on Build GRCh38
rs543724030 Chr.11: 111417930-111417930 on Build GRCh38
rs150318260 Chr.11: 111418148-111418148 on Build GRCh38
rs547506493 Chr.11: 111417986-111417986 on Build GRCh38
rs1021552102 Chr.11: 111418256-111418256 on Build GRCh38
rs183579467 Chr.11: 111418294-111418294 on Build GRCh38
rs1024098703 Chr.11: 111418154-111418154 on Build GRCh38
rs140232682 Chr.11: 111417961-111417961 on Build GRCh38
rs943159194 Chr.11: 111418030-111418030 on Build GRCh38
rs375468844 Chr.11: 111417939-111417939 on Build GRCh38
rs531282457 Chr.11: 111418208-111418208 on Build GRCh38
rs188659781 Chr.11: 111418280-111418280 on Build GRCh38
rs556466627 Chr.11: 111418050-111418050 on Build GRCh38
rs138293345 Chr.11: 111418302-111418302 on Build GRCh38
rs976495416 Chr.11: 111418319-111418319 on Build GRCh38
rs538884390 Chr.11: 111418139-111418139 on Build GRCh38
rs910398303 Chr.11: 111418010-111418010 on Build GRCh38
rs1032371165 Chr.11: 111418173-111418173 on Build GRCh38
rs929206348 Chr.11: 111418042-111418042 on Build GRCh38
rs542326313 Chr.11: 111418118-111418118 on Build GRCh38
rs7106405 Chr.11: 111418243-111418243 on Build GRCh38
rs188662505 Chr.11: 111418115-111418115 on Build GRCh38
rs1025190691 Chr.11: 111418134-111418134 on Build GRCh38
rs914367988 Chr.11: 111417998-111417998 on Build GRCh38
rs543339380 Chr.11: 111418211-111418211 on Build GRCh38
rs929262763 Chr.11: 111418245-111418245 on Build GRCh38
rs369753929 Chr.11: 111417970-111417970 on Build GRCh38
rs771155077 Chr.11: 111417976-111417976 on Build GRCh38
rs529800525 Chr.11: 111417992-111417992 on Build GRCh38
rs569649371 Chr.11: 111418031-111418031 on Build GRCh38
rs547833512 Chr.11: 111418007-111418007 on Build GRCh38
rs1031045485 Chr.11: 111417943-111417943 on Build GRCh38
rs943825462 Chr.11: 111418059-111418059 on Build GRCh38
rs770008672 Chr.11: 111418090-111418090 on Build GRCh38
rs184109977 Chr.11: 111418174-111418174 on Build GRCh38
rs565828116 Chr.11: 111417989-111417989 on Build GRCh38
rs958190425 Chr.11: 111417991-111417991 on Build GRCh38
rs191416297 Chr.11: 111418170-111418170 on Build GRCh38
rs375259145 Chr.11: 111418158-111418158 on Build GRCh38
rs766047057 Chr.11: 111418283-111418283 on Build GRCh38
rs866810953 Chr.11: 111418307-111418307 on Build GRCh38
rs985308815 Chr.11: 111418276-111418276 on Build GRCh38

View Full Product Details