Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 2223228-2223730 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNA1C
Entrez Gene ID
775
Gene Name
calcium voltage-gated channel subunit alpha1 C
Gene Aliases
CACH2, CACN2, CACNL1A1, CCHL1A1, CaV1.2, LQT8, TS
UniGene
Hs.118262
Gene Chromosome Location
Chr.12: 1969677-2697949 on Build GRCh38
Transcripts
NM_000719.6 NM_001129827.1 NM_001129829.1 NM_001129830.2 NM_001129831.1 NM_001129832.1 NM_001129833.1 NM_001129834.1 NM_001129835.1 NM_001129836.1 NM_001129837.1 NM_001129838.1 NM_001129839.1 NM_001129840.1 NM_001129841.1 NM_001129842.1 NM_001129843.1 NM_001129844.1 NM_001129846.1 NM_001167623.1 NM_001167624.2 NM_001167625.1 NM_199460.3 XM_006719017.2 XM_011521020.2 XM_011521023.2 XM_017019926.1 XM_017019927.1 XM_017019928.1 XM_017019929.1 XM_017019930.1 XM_017019931.1 XM_017019932.1 XM_017019933.1 XM_017019934.1 XM_017019935.1 XM_017019936.1 XM_017019937.1 XM_017019938.1 XM_017019939.1 XM_017019940.1 XM_017019941.1 XM_017019942.1 XM_017019943.1 XM_017019944.1 XM_017019945.1 XM_017019946.1 XM_017019947.1 XM_017019948.1 XM_017019949.1 XM_017019950.1 XM_017019951.1 XM_017019952.1 XM_017019953.1 XM_017019954.1 XM_017019955.1 NR_046578.1

Gene Symbol
CACNA1C-AS4
Entrez Gene ID
100874234
Gene Name
CACNA1C antisense RNA 4
Gene Aliases
-
UniGene
Hs.372570
Gene Chromosome Location
Chr.12: 2220537-2223481 on Build GRCh38
Transcripts
NM_000719.6 NM_001129827.1 NM_001129829.1 NM_001129830.2 NM_001129831.1 NM_001129832.1 NM_001129833.1 NM_001129834.1 NM_001129835.1 NM_001129836.1 NM_001129837.1 NM_001129838.1 NM_001129839.1 NM_001129840.1 NM_001129841.1 NM_001129842.1 NM_001129843.1 NM_001129844.1 NM_001129846.1 NM_001167623.1 NM_001167624.2 NM_001167625.1 NM_199460.3 XM_006719017.2 XM_011521020.2 XM_011521023.2 XM_017019926.1 XM_017019927.1 XM_017019928.1 XM_017019929.1 XM_017019930.1 XM_017019931.1 XM_017019932.1 XM_017019933.1 XM_017019934.1 XM_017019935.1 XM_017019936.1 XM_017019937.1 XM_017019938.1 XM_017019939.1 XM_017019940.1 XM_017019941.1 XM_017019942.1 XM_017019943.1 XM_017019944.1 XM_017019945.1 XM_017019946.1 XM_017019947.1 XM_017019948.1 XM_017019949.1 XM_017019950.1 XM_017019951.1 XM_017019952.1 XM_017019953.1 XM_017019954.1 XM_017019955.1 NR_046578.1

SNP Detail

SNP ID Chromosome Location
rs1028664755 Chr.12: 2223477-2223477 on Build GRCh38
rs959535546 Chr.12: 2223444-2223444 on Build GRCh38
rs776563836 Chr.12: 2223405-2223405 on Build GRCh38
rs1025536002 Chr.12: 2223315-2223315 on Build GRCh38
rs529586696 Chr.12: 2223412-2223412 on Build GRCh38
rs758252373 Chr.12: 2223355-2223355 on Build GRCh38
rs746136774 Chr.12: 2223603-2223603 on Build GRCh38
rs1055027315 Chr.12: 2223642-2223642 on Build GRCh38
rs576458936 Chr.12: 2223287-2223287 on Build GRCh38
rs954761432 Chr.12: 2223478-2223478 on Build GRCh38
rs143162892 Chr.12: 2223645-2223645 on Build GRCh38
rs987417223 Chr.12: 2223498-2223498 on Build GRCh38
rs558008369 Chr.12: 2223333-2223333 on Build GRCh38
rs575896519 Chr.12: 2223641-2223641 on Build GRCh38
rs896818968 Chr.12: 2223298-2223298 on Build GRCh38
rs992362396 Chr.12: 2223445-2223445 on Build GRCh38
rs1108222 Chr.12: 2223690-2223690 on Build GRCh38
rs916594345 Chr.12: 2223644-2223644 on Build GRCh38
rs1032136009 Chr.12: 2223342-2223342 on Build GRCh38
rs993822475 Chr.12: 2223704-2223704 on Build GRCh38
rs993875019 Chr.12: 2223301-2223301 on Build GRCh38
rs112218179 Chr.12: 2223536-2223536 on Build GRCh38
rs978145238 Chr.12: 2223542-2223542 on Build GRCh38
rs150210050 Chr.12: 2223360-2223360 on Build GRCh38
rs1108221 Chr.12: 2223629-2223629 on Build GRCh38
rs995807023 Chr.12: 2223467-2223467 on Build GRCh38
rs895363744 Chr.12: 2223274-2223274 on Build GRCh38
rs2239029 Chr.12: 2223262-2223262 on Build GRCh38
rs75121418 Chr.12: 2223566-2223566 on Build GRCh38
rs1019553253 Chr.12: 2223291-2223291 on Build GRCh38
rs76269752 Chr.12: 2223565-2223565 on Build GRCh38
rs907687913 Chr.12: 2223499-2223499 on Build GRCh38
rs911026027 Chr.12: 2223651-2223651 on Build GRCh38
rs961927291 Chr.12: 2223535-2223535 on Build GRCh38
rs897099543 Chr.12: 2223701-2223701 on Build GRCh38
rs577799890 Chr.12: 2223338-2223338 on Build GRCh38
rs902681670 Chr.12: 2223700-2223700 on Build GRCh38

View Full Product Details